Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47623431-47623948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORIN
Entrez Gene ID
10699
Gene Name
corin, serine peptidase
Gene Aliases
ATC2, CRN, Lrp4, PEE5, TMPRSS10
UniGene
Hs.518618
Gene Chromosome Location
Chr.4: 47593998-47838106 on Build GRCh38
Transcripts
NM_001278585.1 NM_006587.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3604192 c.2345C>T p.S782F Chr.4: 47623919-47623919 on Build GRCh38
3604191 c.2403G>A p.R801R Chr.4: 47623708-47623708 on Build GRCh38
1202092 c.2340A>C p.K780N Chr.4: 47623924-47623924 on Build GRCh38
6634171 c.2421G>A p.T807T Chr.4: 47623690-47623690 on Build GRCh38
4962900 c.2446C>A p.Q816K Chr.4: 47623665-47623665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149965013 Chr.4: 47623631-47623631 on Build GRCh38
rs370501103 Chr.4: 47623524-47623524 on Build GRCh38
rs755376280 Chr.4: 47623874-47623874 on Build GRCh38
rs952765692 Chr.4: 47623706-47623706 on Build GRCh38
rs143333327 Chr.4: 47623733-47623733 on Build GRCh38
rs141053134 Chr.4: 47623694-47623694 on Build GRCh38
rs766007832 Chr.4: 47623569-47623570 on Build GRCh38
rs762760721 Chr.4: 47623719-47623719 on Build GRCh38
rs573873768 Chr.4: 47623652-47623652 on Build GRCh38
rs35604278 Chr.4: 47623837-47623837 on Build GRCh38
rs536003106 Chr.4: 47623691-47623691 on Build GRCh38
rs1034415334 Chr.4: 47623570-47623570 on Build GRCh38
rs781733056 Chr.4: 47623589-47623589 on Build GRCh38
rs752320935 Chr.4: 47623664-47623664 on Build GRCh38
rs189947013 Chr.4: 47623557-47623557 on Build GRCh38
rs750996817 Chr.4: 47623685-47623685 on Build GRCh38
rs768827492 Chr.4: 47623617-47623617 on Build GRCh38
rs915131051 Chr.4: 47623628-47623628 on Build GRCh38
rs554274347 Chr.4: 47623563-47623563 on Build GRCh38
rs760272638 Chr.4: 47623866-47623866 on Build GRCh38
rs76975373 Chr.4: 47623756-47623756 on Build GRCh38
rs551053223 Chr.4: 47623471-47623471 on Build GRCh38
rs766922887 Chr.4: 47623869-47623869 on Build GRCh38
rs747035467 Chr.4: 47623921-47623921 on Build GRCh38
rs757886805 Chr.4: 47623575-47623575 on Build GRCh38
rs989604895 Chr.4: 47623697-47623697 on Build GRCh38
rs758410173 Chr.4: 47623728-47623739 on Build GRCh38
rs921985295 Chr.4: 47623470-47623470 on Build GRCh38
rs767103061 Chr.4: 47623728-47623728 on Build GRCh38
rs748105735 Chr.4: 47623692-47623692 on Build GRCh38
rs762631726 Chr.4: 47623666-47623666 on Build GRCh38
rs777099502 Chr.4: 47623686-47623686 on Build GRCh38
rs757662978 Chr.4: 47623587-47623587 on Build GRCh38
rs914875635 Chr.4: 47623805-47623805 on Build GRCh38
rs780156009 Chr.4: 47623763-47623764 on Build GRCh38
rs749700191 Chr.4: 47623690-47623690 on Build GRCh38
rs754819585 Chr.4: 47623558-47623558 on Build GRCh38
rs185575139 Chr.4: 47623497-47623497 on Build GRCh38
rs770719777 Chr.4: 47623634-47623634 on Build GRCh38
rs761978477 Chr.4: 47623632-47623632 on Build GRCh38
rs746160285 Chr.4: 47623599-47623599 on Build GRCh38
rs759571237 Chr.4: 47623700-47623700 on Build GRCh38
rs754459418 Chr.4: 47623870-47623870 on Build GRCh38
rs750529501 Chr.4: 47623639-47623640 on Build GRCh38
rs748464470 Chr.4: 47623742-47623742 on Build GRCh38
rs983083810 Chr.4: 47623493-47623493 on Build GRCh38
rs777952463 Chr.4: 47623751-47623751 on Build GRCh38
rs755154812 Chr.4: 47623913-47623917 on Build GRCh38
rs397729701 Chr.4: 47623797-47623797 on Build GRCh38
rs771493157 Chr.4: 47623606-47623606 on Build GRCh38
rs761224967 Chr.4: 47623723-47623723 on Build GRCh38
rs1057205030 Chr.4: 47623681-47623681 on Build GRCh38
rs764807663 Chr.4: 47623559-47623559 on Build GRCh38
rs752408919 Chr.4: 47623560-47623560 on Build GRCh38
rs781484683 Chr.4: 47623684-47623684 on Build GRCh38
rs751229050 Chr.4: 47623722-47623722 on Build GRCh38
rs773852508 Chr.4: 47623850-47623850 on Build GRCh38
rs965612395 Chr.4: 47623517-47623517 on Build GRCh38
rs553392138 Chr.4: 47623556-47623556 on Build GRCh38
rs778994562 Chr.4: 47623737-47623737 on Build GRCh38
rs202055180 Chr.4: 47623849-47623849 on Build GRCh38
rs763680668 Chr.4: 47623721-47623721 on Build GRCh38
rs146408075 Chr.4: 47623881-47623881 on Build GRCh38
rs760023375 Chr.4: 47623698-47623698 on Build GRCh38
rs75141391 Chr.4: 47623736-47623736 on Build GRCh38
rs2292104 Chr.4: 47623853-47623853 on Build GRCh38
rs967260783 Chr.4: 47623877-47623877 on Build GRCh38
rs763764889 Chr.4: 47623671-47623671 on Build GRCh38
rs760571452 Chr.4: 47623537-47623537 on Build GRCh38
rs759242885 Chr.4: 47623657-47623657 on Build GRCh38
rs754133364 Chr.4: 47623891-47623891 on Build GRCh38
rs371632457 Chr.4: 47623919-47623919 on Build GRCh38
rs766373399 Chr.4: 47623551-47623551 on Build GRCh38
rs1004868234 Chr.4: 47623716-47623716 on Build GRCh38
rs772727213 Chr.4: 47623861-47623861 on Build GRCh38
rs199949182 Chr.4: 47623788-47623788 on Build GRCh38
rs775227959 Chr.4: 47623715-47623715 on Build GRCh38
rs770199212 Chr.4: 47623608-47623608 on Build GRCh38
rs375618006 Chr.4: 47623734-47623734 on Build GRCh38
rs945103650 Chr.4: 47623818-47623818 on Build GRCh38
rs370687466 Chr.4: 47623531-47623531 on Build GRCh38
rs780410217 Chr.4: 47623610-47623610 on Build GRCh38
rs749572458 Chr.4: 47623615-47623615 on Build GRCh38
rs775420735 Chr.4: 47623772-47623772 on Build GRCh38
rs761565961 Chr.4: 47623852-47623852 on Build GRCh38
rs758608893 Chr.4: 47623901-47623901 on Build GRCh38
rs750983513 Chr.4: 47623673-47623673 on Build GRCh38
rs3215139 Chr.4: 47623795-47623795 on Build GRCh38
rs145089913 Chr.4: 47623663-47623663 on Build GRCh38
rs1033241784 Chr.4: 47623896-47623896 on Build GRCh38
rs765540392 Chr.4: 47623792-47623792 on Build GRCh38
rs780604194 Chr.4: 47623688-47623688 on Build GRCh38
rs758389169 Chr.4: 47623744-47623744 on Build GRCh38
rs769410001 Chr.4: 47623850-47623850 on Build GRCh38
rs769548762 Chr.4: 47623712-47623712 on Build GRCh38
rs868081324 Chr.4: 47623783-47623783 on Build GRCh38
rs367732949 Chr.4: 47623855-47623855 on Build GRCh38
rs747125498 Chr.4: 47623754-47623754 on Build GRCh38
rs756803885 Chr.4: 47623682-47623682 on Build GRCh38
rs1005022207 Chr.4: 47623538-47623538 on Build GRCh38
rs200808483 Chr.4: 47623576-47623576 on Build GRCh38
rs142240381 Chr.4: 47623777-47623777 on Build GRCh38
rs958387461 Chr.4: 47623601-47623601 on Build GRCh38

View Full Product Details