Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47641796-47642333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORIN
Entrez Gene ID
10699
Gene Name
corin, serine peptidase
Gene Aliases
ATC2, CRN, Lrp4, PEE5, TMPRSS10
UniGene
Hs.518618
Gene Chromosome Location
Chr.4: 47593998-47838106 on Build GRCh38
Transcripts
NM_001278585.1 NM_006587.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5866460 c.2132A>G p.H711R Chr.4: 47641986-47641986 on Build GRCh38
6529222 c.2078C>G p.S693C Chr.4: 47642040-47642040 on Build GRCh38
1235993 c.2192G>A p.G731D Chr.4: 47641926-47641926 on Build GRCh38
4843133 c.2091C>T p.N697N Chr.4: 47642027-47642027 on Build GRCh38
3373500 c.2088G>T p.V696V Chr.4: 47642030-47642030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748677860 Chr.4: 47642046-47642046 on Build GRCh38
rs201847788 Chr.4: 47641992-47641992 on Build GRCh38
rs551419755 Chr.4: 47642072-47642072 on Build GRCh38
rs1045571976 Chr.4: 47642038-47642038 on Build GRCh38
rs767658082 Chr.4: 47641896-47641896 on Build GRCh38
rs143992388 Chr.4: 47641948-47641948 on Build GRCh38
rs774841632 Chr.4: 47641977-47641977 on Build GRCh38
rs17462783 Chr.4: 47641846-47641846 on Build GRCh38
rs757124949 Chr.4: 47641932-47641932 on Build GRCh38
rs544560178 Chr.4: 47641996-47641996 on Build GRCh38
rs749902880 Chr.4: 47642098-47642098 on Build GRCh38
rs577151724 Chr.4: 47641914-47641914 on Build GRCh38
rs769093722 Chr.4: 47641971-47641971 on Build GRCh38
rs533707594 Chr.4: 47642001-47642001 on Build GRCh38
rs147292219 Chr.4: 47641981-47641981 on Build GRCh38
rs374332011 Chr.4: 47641893-47641893 on Build GRCh38
rs761428760 Chr.4: 47642054-47642054 on Build GRCh38
rs751614868 Chr.4: 47642004-47642004 on Build GRCh38
rs913130723 Chr.4: 47642287-47642287 on Build GRCh38
rs1003534299 Chr.4: 47641844-47641844 on Build GRCh38
rs534281692 Chr.4: 47641860-47641860 on Build GRCh38
rs373734354 Chr.4: 47642222-47642223 on Build GRCh38
rs1055097552 Chr.4: 47642091-47642091 on Build GRCh38
rs773729668 Chr.4: 47642098-47642099 on Build GRCh38
rs879745456 Chr.4: 47642223-47642223 on Build GRCh38
rs750061020 Chr.4: 47642022-47642022 on Build GRCh38
rs542026992 Chr.4: 47642118-47642118 on Build GRCh38
rs1013882735 Chr.4: 47641851-47641851 on Build GRCh38
rs527431682 Chr.4: 47642277-47642277 on Build GRCh38
rs965821044 Chr.4: 47642197-47642197 on Build GRCh38
rs1005040280 Chr.4: 47642186-47642186 on Build GRCh38
rs906518682 Chr.4: 47641823-47641823 on Build GRCh38
rs974269321 Chr.4: 47642141-47642141 on Build GRCh38
rs560305643 Chr.4: 47642274-47642274 on Build GRCh38
rs774649418 Chr.4: 47641881-47641881 on Build GRCh38
rs935894161 Chr.4: 47641974-47641974 on Build GRCh38
rs61759686 Chr.4: 47642188-47642188 on Build GRCh38
rs771105672 Chr.4: 47641873-47641873 on Build GRCh38
rs750674980 Chr.4: 47641897-47641897 on Build GRCh38
rs1004078130 Chr.4: 47642075-47642075 on Build GRCh38
rs760792402 Chr.4: 47641911-47641911 on Build GRCh38
rs201948411 Chr.4: 47641982-47641982 on Build GRCh38
rs764059963 Chr.4: 47641999-47641999 on Build GRCh38
rs767265119 Chr.4: 47642018-47642018 on Build GRCh38
rs754548296 Chr.4: 47642050-47642050 on Build GRCh38
rs757106498 Chr.4: 47642009-47642009 on Build GRCh38
rs191176929 Chr.4: 47641913-47641913 on Build GRCh38
rs920105790 Chr.4: 47642143-47642143 on Build GRCh38
rs950969277 Chr.4: 47642172-47642172 on Build GRCh38
rs1028718309 Chr.4: 47642011-47642011 on Build GRCh38
rs775273225 Chr.4: 47641915-47641915 on Build GRCh38
rs138303275 Chr.4: 47641926-47641926 on Build GRCh38
rs921578463 Chr.4: 47642266-47642266 on Build GRCh38
rs957480180 Chr.4: 47641972-47641972 on Build GRCh38
rs950874688 Chr.4: 47641904-47641904 on Build GRCh38
rs575029439 Chr.4: 47642044-47642044 on Build GRCh38
rs748852470 Chr.4: 47641968-47641968 on Build GRCh38
rs762725395 Chr.4: 47642133-47642133 on Build GRCh38
rs758232899 Chr.4: 47641919-47641919 on Build GRCh38
rs770530407 Chr.4: 47642097-47642098 on Build GRCh38
rs760842408 Chr.4: 47641985-47641985 on Build GRCh38
rs755628483 Chr.4: 47641949-47641949 on Build GRCh38
rs896107818 Chr.4: 47642220-47642220 on Build GRCh38
rs780988498 Chr.4: 47641941-47641941 on Build GRCh38
rs779471659 Chr.4: 47641966-47641966 on Build GRCh38
rs146079372 Chr.4: 47642286-47642286 on Build GRCh38
rs751344276 Chr.4: 47641928-47641928 on Build GRCh38
rs895146077 Chr.4: 47642111-47642111 on Build GRCh38
rs1027928500 Chr.4: 47642155-47642155 on Build GRCh38
rs139160376 Chr.4: 47642026-47642026 on Build GRCh38
rs1055172238 Chr.4: 47641821-47641821 on Build GRCh38
rs769181782 Chr.4: 47641877-47641877 on Build GRCh38
rs974901331 Chr.4: 47642255-47642255 on Build GRCh38

View Full Product Details