Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48994534-48995036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWH43
Entrez Gene ID
80157
Gene Name
cell wall biogenesis 43 C-terminal homolog
Gene Aliases
CWH43-C
UniGene
Hs.479703
Gene Chromosome Location
Chr.4: 48986247-49062079 on Build GRCh38
Transcripts
NM_001286791.1 NM_025087.2 XM_011513755.1 XM_011513756.2 XM_011513757.2 XM_011513758.1 XM_011513759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6167488 c.526A>G p.K176E Chr.4: 48994633-48994633 on Build GRCh38
5642819 c.650C>G p.S217C Chr.4: 48994757-48994757 on Build GRCh38
7003375 c.570C>T p.A190A Chr.4: 48994677-48994677 on Build GRCh38
3604337 c.540G>A p.K180K Chr.4: 48994647-48994647 on Build GRCh38
6545404 c.552G>T p.E184D Chr.4: 48994659-48994659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781562748 Chr.4: 48994668-48994668 on Build GRCh38
rs776181703 Chr.4: 48994807-48994807 on Build GRCh38
rs368666678 Chr.4: 48994851-48994851 on Build GRCh38
rs757666593 Chr.4: 48994751-48994751 on Build GRCh38
rs369675204 Chr.4: 48994567-48994567 on Build GRCh38
rs776312160 Chr.4: 48994704-48994704 on Build GRCh38
rs759013519 Chr.4: 48994708-48994708 on Build GRCh38
rs542406303 Chr.4: 48994693-48994693 on Build GRCh38
rs147719750 Chr.4: 48994882-48994882 on Build GRCh38
rs753030703 Chr.4: 48994566-48994566 on Build GRCh38
rs771141886 Chr.4: 48994598-48994598 on Build GRCh38
rs756179636 Chr.4: 48994755-48994755 on Build GRCh38
rs774313478 Chr.4: 48994608-48994608 on Build GRCh38
rs748048156 Chr.4: 48994773-48994773 on Build GRCh38
rs1049534757 Chr.4: 48994885-48994885 on Build GRCh38
rs916867861 Chr.4: 48994735-48994735 on Build GRCh38
rs374917398 Chr.4: 48994665-48994665 on Build GRCh38
rs745948351 Chr.4: 48994601-48994601 on Build GRCh38
rs369801723 Chr.4: 48994641-48994641 on Build GRCh38
rs774232549 Chr.4: 48994727-48994727 on Build GRCh38
rs371832961 Chr.4: 48994657-48994657 on Build GRCh38
rs752039087 Chr.4: 48994734-48994734 on Build GRCh38
rs563132282 Chr.4: 48994604-48994604 on Build GRCh38
rs759252470 Chr.4: 48994968-48994968 on Build GRCh38
rs759078197 Chr.4: 48994644-48994644 on Build GRCh38
rs377675320 Chr.4: 48994700-48994700 on Build GRCh38
rs762978917 Chr.4: 48994636-48994636 on Build GRCh38
rs543442731 Chr.4: 48994583-48994583 on Build GRCh38
rs760252951 Chr.4: 48994623-48994623 on Build GRCh38
rs769203270 Chr.4: 48994821-48994821 on Build GRCh38
rs370975619 Chr.4: 48994853-48994853 on Build GRCh38
rs745461380 Chr.4: 48994815-48994815 on Build GRCh38
rs758385628 Chr.4: 48994779-48994779 on Build GRCh38
rs552225134 Chr.4: 48994909-48994909 on Build GRCh38
rs866694562 Chr.4: 48994790-48994790 on Build GRCh38
rs762549873 Chr.4: 48994837-48994837 on Build GRCh38
rs369636597 Chr.4: 48994800-48994800 on Build GRCh38
rs757613110 Chr.4: 48994667-48994667 on Build GRCh38
rs772932772 Chr.4: 48994695-48994695 on Build GRCh38
rs774042614 Chr.4: 48994760-48994761 on Build GRCh38
rs374066982 Chr.4: 48994806-48994806 on Build GRCh38
rs767900405 Chr.4: 48994753-48994753 on Build GRCh38
rs369610807 Chr.4: 48994675-48994675 on Build GRCh38
rs780112691 Chr.4: 48994766-48994766 on Build GRCh38
rs765699849 Chr.4: 48994650-48994650 on Build GRCh38
rs934434038 Chr.4: 48994846-48994846 on Build GRCh38
rs775869448 Chr.4: 48994731-48994731 on Build GRCh38
rs889729647 Chr.4: 48994964-48994964 on Build GRCh38
rs745975176 Chr.4: 48994669-48994669 on Build GRCh38
rs778167519 Chr.4: 48994580-48994580 on Build GRCh38
rs746842057 Chr.4: 48994805-48994805 on Build GRCh38
rs771804892 Chr.4: 48994621-48994621 on Build GRCh38
rs774976213 Chr.4: 48994852-48994852 on Build GRCh38
rs750614604 Chr.4: 48994867-48994867 on Build GRCh38
rs758725626 Chr.4: 48994656-48994656 on Build GRCh38
rs767882350 Chr.4: 48994862-48994862 on Build GRCh38
rs942078636 Chr.4: 48994896-48994896 on Build GRCh38
rs775126332 Chr.4: 48994823-48994823 on Build GRCh38
rs776461107 Chr.4: 48994949-48994949 on Build GRCh38
rs184009054 Chr.4: 48994655-48994655 on Build GRCh38
rs868810538 Chr.4: 48994796-48994796 on Build GRCh38
rs764768646 Chr.4: 48994717-48994717 on Build GRCh38
rs763498154 Chr.4: 48994733-48994733 on Build GRCh38
rs749392071 Chr.4: 48994561-48994561 on Build GRCh38
rs778737042 Chr.4: 48994676-48994676 on Build GRCh38
rs373628138 Chr.4: 48994686-48994686 on Build GRCh38
rs147167780 Chr.4: 48994698-48994698 on Build GRCh38
rs781366446 Chr.4: 48994600-48994600 on Build GRCh38
rs1008253384 Chr.4: 48994605-48994605 on Build GRCh38
rs750611314 Chr.4: 48994754-48994754 on Build GRCh38
rs201613779 Chr.4: 48994613-48994613 on Build GRCh38
rs772880873 Chr.4: 48994622-48994622 on Build GRCh38

View Full Product Details