Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 49007011-49007432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWH43
Entrez Gene ID
80157
Gene Name
cell wall biogenesis 43 C-terminal homolog
Gene Aliases
CWH43-C
UniGene
Hs.479703
Gene Chromosome Location
Chr.4: 48986247-49062079 on Build GRCh38
Transcripts
NM_001286791.1 NM_025087.2 XM_011513755.1 XM_011513756.2 XM_011513757.2 XM_011513758.1 XM_011513759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7326476 c.1081G>C p.G361R Chr.4: 49007221-49007221 on Build GRCh38
4521335 c.1107G>A p.K369K Chr.4: 49007247-49007247 on Build GRCh38
7209840 c.1122G>T p.L374F Chr.4: 49007262-49007262 on Build GRCh38
7192643 c.1071G>C p.M357I Chr.4: 49007211-49007211 on Build GRCh38
3604344 c.1108A>C p.K370Q Chr.4: 49007248-49007248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756630342 Chr.4: 49007349-49007349 on Build GRCh38
rs201462278 Chr.4: 49007196-49007196 on Build GRCh38
rs772227117 Chr.4: 49007190-49007190 on Build GRCh38
rs750022722 Chr.4: 49007190-49007194 on Build GRCh38
rs375380080 Chr.4: 49007281-49007281 on Build GRCh38
rs769722314 Chr.4: 49007220-49007220 on Build GRCh38
rs370189018 Chr.4: 49007177-49007177 on Build GRCh38
rs768825227 Chr.4: 49007173-49007173 on Build GRCh38
rs751179545 Chr.4: 49007237-49007237 on Build GRCh38
rs781405037 Chr.4: 49007286-49007286 on Build GRCh38
rs1018573427 Chr.4: 49007168-49007168 on Build GRCh38
rs1028599407 Chr.4: 49007088-49007088 on Build GRCh38
rs773997932 Chr.4: 49007378-49007378 on Build GRCh38
rs779025162 Chr.4: 49007320-49007320 on Build GRCh38
rs755411433 Chr.4: 49007201-49007202 on Build GRCh38
rs377060143 Chr.4: 49007174-49007174 on Build GRCh38
rs753314035 Chr.4: 49007315-49007315 on Build GRCh38
rs747084090 Chr.4: 49007354-49007354 on Build GRCh38
rs1053218822 Chr.4: 49007397-49007397 on Build GRCh38
rs756673155 Chr.4: 49007293-49007293 on Build GRCh38
rs775480232 Chr.4: 49007363-49007363 on Build GRCh38
rs373362250 Chr.4: 49007167-49007167 on Build GRCh38
rs893338846 Chr.4: 49007059-49007059 on Build GRCh38
rs532968698 Chr.4: 49007401-49007401 on Build GRCh38
rs35035590 Chr.4: 49007199-49007200 on Build GRCh38
rs772280407 Chr.4: 49007343-49007343 on Build GRCh38
rs370474877 Chr.4: 49007123-49007123 on Build GRCh38
rs755347963 Chr.4: 49007308-49007308 on Build GRCh38
rs370222566 Chr.4: 49007161-49007161 on Build GRCh38
rs145501408 Chr.4: 49007066-49007066 on Build GRCh38
rs1011735208 Chr.4: 49007081-49007081 on Build GRCh38
rs146581575 Chr.4: 49007203-49007203 on Build GRCh38
rs143983446 Chr.4: 49007287-49007287 on Build GRCh38
rs756589309 Chr.4: 49007118-49007118 on Build GRCh38
rs139130649 Chr.4: 49007178-49007178 on Build GRCh38
rs535528683 Chr.4: 49007049-49007049 on Build GRCh38
rs768458618 Chr.4: 49007373-49007373 on Build GRCh38
rs749064162 Chr.4: 49007364-49007364 on Build GRCh38
rs777939824 Chr.4: 49007347-49007347 on Build GRCh38
rs112484785 Chr.4: 49007043-49007043 on Build GRCh38
rs748337177 Chr.4: 49007339-49007339 on Build GRCh38
rs780605224 Chr.4: 49007304-49007304 on Build GRCh38
rs754193083 Chr.4: 49007306-49007306 on Build GRCh38
rs954293847 Chr.4: 49007145-49007145 on Build GRCh38
rs145043160 Chr.4: 49007271-49007271 on Build GRCh38
rs749650251 Chr.4: 49007171-49007171 on Build GRCh38
rs141315226 Chr.4: 49007221-49007221 on Build GRCh38
rs769663465 Chr.4: 49007357-49007357 on Build GRCh38

View Full Product Details