Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55430514-55431054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLOCK
Entrez Gene ID
9575
Gene Name
clock circadian regulator
Gene Aliases
KAT13D, bHLHe8
UniGene
Hs.436975
Gene Chromosome Location
Chr.4: 55427901-55547138 on Build GRCh38
Transcripts
NM_001267843.1 NM_004898.3 XM_005265787.2 XM_011534409.2 XM_011534410.2 XM_011534411.2 XM_017008854.1 XM_017008855.1 XM_011534394.2

Gene Symbol
TMEM165
Entrez Gene ID
55858
Gene Name
transmembrane protein 165
Gene Aliases
CDG2K, FT27, GDT1, TMPT27, TPARL
UniGene
Hs.479766
Gene Chromosome Location
Chr.4: 55395913-55453397 on Build GRCh38
Transcripts
NM_001267843.1 NM_004898.3 XM_005265787.2 XM_011534409.2 XM_011534410.2 XM_011534411.2 XM_017008854.1 XM_017008855.1 XM_011534394.2

SNP Detail

SNP ID Chromosome Location
rs897758506 Chr.4: 55430908-55430908 on Build GRCh38
rs748162607 Chr.4: 55430768-55430768 on Build GRCh38
rs939031986 Chr.4: 55430864-55430864 on Build GRCh38
rs747285489 Chr.4: 55430598-55430598 on Build GRCh38
rs956680093 Chr.4: 55430652-55430652 on Build GRCh38
rs1044857843 Chr.4: 55430840-55430840 on Build GRCh38
rs531668311 Chr.4: 55430723-55430723 on Build GRCh38
rs922881823 Chr.4: 55430755-55430755 on Build GRCh38
rs897800416 Chr.4: 55430849-55430849 on Build GRCh38
rs562524531 Chr.4: 55430707-55430707 on Build GRCh38
rs1025708977 Chr.4: 55430634-55430634 on Build GRCh38
rs956388721 Chr.4: 55430986-55430986 on Build GRCh38
rs763393632 Chr.4: 55430976-55430976 on Build GRCh38
rs904874604 Chr.4: 55430590-55430590 on Build GRCh38
rs780139675 Chr.4: 55430925-55430925 on Build GRCh38
rs1056545 Chr.4: 55430730-55430730 on Build GRCh38
rs559139302 Chr.4: 55430988-55430988 on Build GRCh38
rs5863 Chr.4: 55430740-55430740 on Build GRCh38
rs961894580 Chr.4: 55430601-55430601 on Build GRCh38
rs547125910 Chr.4: 55430775-55430775 on Build GRCh38
rs952935341 Chr.4: 55430665-55430665 on Build GRCh38
rs1018087246 Chr.4: 55430931-55430931 on Build GRCh38
rs981031476 Chr.4: 55430762-55430762 on Build GRCh38
rs993775984 Chr.4: 55430619-55430619 on Build GRCh38
rs946432287 Chr.4: 55430838-55430838 on Build GRCh38
rs904824077 Chr.4: 55430842-55430842 on Build GRCh38
rs899753802 Chr.4: 55430949-55430952 on Build GRCh38
rs552318026 Chr.4: 55430606-55430606 on Build GRCh38
rs770399717 Chr.4: 55430954-55430954 on Build GRCh38
rs929394023 Chr.4: 55430854-55430854 on Build GRCh38
rs536603494 Chr.4: 55430758-55430758 on Build GRCh38
rs1018206737 Chr.4: 55430712-55430712 on Build GRCh38
rs890988186 Chr.4: 55430895-55430895 on Build GRCh38
rs1056547 Chr.4: 55430596-55430596 on Build GRCh38
rs538883364 Chr.4: 55430970-55430970 on Build GRCh38
rs1051843556 Chr.4: 55430871-55430871 on Build GRCh38
rs1056390357 Chr.4: 55430879-55430879 on Build GRCh38
rs987881543 Chr.4: 55430667-55430667 on Build GRCh38
rs73151851 Chr.4: 55430963-55430963 on Build GRCh38
rs575737744 Chr.4: 55431029-55431029 on Build GRCh38
rs942250262 Chr.4: 55430839-55430839 on Build GRCh38

View Full Product Details