Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55430935-55431462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLOCK
Entrez Gene ID
9575
Gene Name
clock circadian regulator
Gene Aliases
KAT13D, bHLHe8
UniGene
Hs.436975
Gene Chromosome Location
Chr.4: 55427901-55547138 on Build GRCh38
Transcripts
NM_001267843.1 NM_004898.3 XM_005265787.2 XM_011534409.2 XM_011534410.2 XM_011534411.2 XM_017008854.1 XM_017008855.1 XM_011534394.2

Gene Symbol
TMEM165
Entrez Gene ID
55858
Gene Name
transmembrane protein 165
Gene Aliases
CDG2K, FT27, GDT1, TMPT27, TPARL
UniGene
Hs.479766
Gene Chromosome Location
Chr.4: 55395913-55453397 on Build GRCh38
Transcripts
NM_001267843.1 NM_004898.3 XM_005265787.2 XM_011534409.2 XM_011534410.2 XM_011534411.2 XM_017008854.1 XM_017008855.1 XM_011534394.2

SNP Detail

SNP ID Chromosome Location
rs369707775 Chr.4: 55431150-55431150 on Build GRCh38
rs980911971 Chr.4: 55431079-55431080 on Build GRCh38
rs915373651 Chr.4: 55431129-55431129 on Build GRCh38
rs759277202 Chr.4: 55431276-55431276 on Build GRCh38
rs559139302 Chr.4: 55430988-55430988 on Build GRCh38
rs926398458 Chr.4: 55431186-55431186 on Build GRCh38
rs78795192 Chr.4: 55431434-55431434 on Build GRCh38
rs938979640 Chr.4: 55431195-55431195 on Build GRCh38
rs539807043 Chr.4: 55431270-55431270 on Build GRCh38
rs897501629 Chr.4: 55431274-55431274 on Build GRCh38
rs554795137 Chr.4: 55431184-55431184 on Build GRCh38
rs976761842 Chr.4: 55431095-55431095 on Build GRCh38
rs890552937 Chr.4: 55431197-55431198 on Build GRCh38
rs1029897372 Chr.4: 55431098-55431098 on Build GRCh38
rs988321668 Chr.4: 55431054-55431054 on Build GRCh38
rs576900799 Chr.4: 55431273-55431273 on Build GRCh38
rs957208003 Chr.4: 55431099-55431099 on Build GRCh38
rs1005553301 Chr.4: 55431042-55431042 on Build GRCh38
rs73151851 Chr.4: 55430963-55430963 on Build GRCh38
rs1032243387 Chr.4: 55431342-55431342 on Build GRCh38
rs973651170 Chr.4: 55431163-55431163 on Build GRCh38
rs191647866 Chr.4: 55431332-55431332 on Build GRCh38
rs560010915 Chr.4: 55431272-55431272 on Build GRCh38
rs1001220704 Chr.4: 55431277-55431277 on Build GRCh38
rs956388721 Chr.4: 55430986-55430986 on Build GRCh38
rs763393632 Chr.4: 55430976-55430976 on Build GRCh38
rs189091341 Chr.4: 55431221-55431221 on Build GRCh38
rs942304120 Chr.4: 55431132-55431132 on Build GRCh38
rs1007381761 Chr.4: 55431395-55431395 on Build GRCh38
rs35962723 Chr.4: 55431186-55431187 on Build GRCh38
rs779685802 Chr.4: 55431240-55431241 on Build GRCh38
rs1048974886 Chr.4: 55431380-55431380 on Build GRCh38
rs538193827 Chr.4: 55431178-55431178 on Build GRCh38
rs771308065 Chr.4: 55431092-55431092 on Build GRCh38
rs1024726580 Chr.4: 55431064-55431064 on Build GRCh38
rs752312373 Chr.4: 55431412-55431412 on Build GRCh38
rs970902142 Chr.4: 55431075-55431075 on Build GRCh38
rs1018321476 Chr.4: 55431062-55431062 on Build GRCh38
rs946874118 Chr.4: 55431139-55431139 on Build GRCh38
rs887573515 Chr.4: 55431394-55431394 on Build GRCh38
rs35186124 Chr.4: 55431068-55431069 on Build GRCh38
rs774661794 Chr.4: 55431250-55431250 on Build GRCh38
rs988426918 Chr.4: 55431119-55431119 on Build GRCh38
rs538883364 Chr.4: 55430970-55430970 on Build GRCh38
rs892167947 Chr.4: 55431402-55431402 on Build GRCh38
rs919279453 Chr.4: 55431164-55431164 on Build GRCh38
rs899952502 Chr.4: 55431413-55431413 on Build GRCh38
rs767056747 Chr.4: 55431369-55431369 on Build GRCh38
rs575737744 Chr.4: 55431029-55431029 on Build GRCh38

View Full Product Details