Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55870560-55871062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC1
Entrez Gene ID
55763
Gene Name
exocyst complex component 1
Gene Aliases
BM-102, SEC3, SEC3L1, SEC3P
UniGene
Hs.269665
Gene Chromosome Location
Chr.4: 55853616-55905078 on Build GRCh38
Transcripts
NM_001024924.1 NM_018261.3 NM_178237.2 XM_005265747.2 XM_005265748.1 XM_005265750.2 XM_017008405.1 XM_017008406.1 XM_017008407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5736975 c.677A>C p.K226T Chr.4: 55870751-55870751 on Build GRCh38
4125236 c.753G>A p.Q251Q Chr.4: 55870827-55870827 on Build GRCh38
1731804 c.765C>A p.S255R Chr.4: 55870839-55870839 on Build GRCh38
1284790 c.741A>G p.E247E Chr.4: 55870815-55870815 on Build GRCh38
6221631 c.604-8C>A p.? Chr.4: 55870670-55870670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182283845 Chr.4: 55870789-55870789 on Build GRCh38
rs769944790 Chr.4: 55870831-55870831 on Build GRCh38
rs755975266 Chr.4: 55870914-55870914 on Build GRCh38
rs951800849 Chr.4: 55870961-55870961 on Build GRCh38
rs1019009183 Chr.4: 55870631-55870631 on Build GRCh38
rs773180012 Chr.4: 55870720-55870720 on Build GRCh38
rs753183532 Chr.4: 55870721-55870721 on Build GRCh38
rs747423589 Chr.4: 55870643-55870643 on Build GRCh38
rs377516805 Chr.4: 55870738-55870738 on Build GRCh38
rs754646510 Chr.4: 55870745-55870745 on Build GRCh38
rs369775108 Chr.4: 55870661-55870661 on Build GRCh38
rs746190725 Chr.4: 55870824-55870824 on Build GRCh38
rs751278702 Chr.4: 55870930-55870930 on Build GRCh38
rs764903178 Chr.4: 55870764-55870764 on Build GRCh38
rs761482435 Chr.4: 55870629-55870636 on Build GRCh38
rs372578105 Chr.4: 55870917-55870917 on Build GRCh38
rs759039846 Chr.4: 55870910-55870910 on Build GRCh38
rs899452936 Chr.4: 55871036-55871036 on Build GRCh38
rs563534569 Chr.4: 55870628-55870628 on Build GRCh38
rs766247298 Chr.4: 55870870-55870870 on Build GRCh38
rs762433095 Chr.4: 55870903-55870903 on Build GRCh38
rs775458425 Chr.4: 55870683-55870683 on Build GRCh38
rs573897321 Chr.4: 55870652-55870652 on Build GRCh38
rs766130509 Chr.4: 55870595-55870595 on Build GRCh38
rs111685222 Chr.4: 55870630-55870631 on Build GRCh38
rs149053656 Chr.4: 55870841-55870841 on Build GRCh38
rs753684117 Chr.4: 55870629-55870632 on Build GRCh38
rs1055191755 Chr.4: 55871003-55871003 on Build GRCh38
rs767397918 Chr.4: 55870704-55870704 on Build GRCh38
rs557931911 Chr.4: 55870931-55870931 on Build GRCh38
rs770003187 Chr.4: 55870668-55870668 on Build GRCh38
rs756658541 Chr.4: 55870763-55870763 on Build GRCh38
rs752377692 Chr.4: 55870898-55870898 on Build GRCh38
rs757930898 Chr.4: 55870660-55870661 on Build GRCh38
rs751053841 Chr.4: 55870911-55870911 on Build GRCh38
rs765286057 Chr.4: 55870632-55870633 on Build GRCh38
rs764845637 Chr.4: 55870893-55870893 on Build GRCh38
rs749380037 Chr.4: 55870840-55870840 on Build GRCh38
rs757768400 Chr.4: 55870820-55870820 on Build GRCh38
rs905048273 Chr.4: 55870608-55870608 on Build GRCh38
rs771991374 Chr.4: 55870627-55870628 on Build GRCh38
rs375609735 Chr.4: 55870918-55870918 on Build GRCh38
rs763051613 Chr.4: 55870698-55870698 on Build GRCh38
rs897754285 Chr.4: 55870751-55870751 on Build GRCh38
rs77829116 Chr.4: 55870607-55870607 on Build GRCh38
rs756482120 Chr.4: 55870927-55870927 on Build GRCh38
rs752186950 Chr.4: 55870777-55870777 on Build GRCh38
rs781697685 Chr.4: 55870822-55870822 on Build GRCh38
rs200303000 Chr.4: 55870930-55870931 on Build GRCh38
rs34281712 Chr.4: 55870659-55870660 on Build GRCh38
rs747511391 Chr.4: 55870794-55870794 on Build GRCh38
rs150749988 Chr.4: 55870628-55870629 on Build GRCh38
rs369619617 Chr.4: 55870949-55870949 on Build GRCh38
rs200395524 Chr.4: 55870884-55870884 on Build GRCh38
rs746089509 Chr.4: 55870664-55870664 on Build GRCh38
rs773202822 Chr.4: 55870852-55870852 on Build GRCh38
rs757902363 Chr.4: 55870787-55870787 on Build GRCh38
rs937720360 Chr.4: 55870985-55870985 on Build GRCh38
rs750269553 Chr.4: 55870648-55870649 on Build GRCh38
rs906272964 Chr.4: 55870945-55870945 on Build GRCh38
rs1007753387 Chr.4: 55871005-55871005 on Build GRCh38
rs760612698 Chr.4: 55870859-55870859 on Build GRCh38
rs763662120 Chr.4: 55870909-55870909 on Build GRCh38
rs72152902 Chr.4: 55870629-55870630 on Build GRCh38
rs373536097 Chr.4: 55870674-55870674 on Build GRCh38
rs775752378 Chr.4: 55870834-55870834 on Build GRCh38
rs778199203 Chr.4: 55870791-55870791 on Build GRCh38

View Full Product Details