Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 67738553-67739066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNRHR
Entrez Gene ID
2798
Gene Name
gonadotropin releasing hormone receptor
Gene Aliases
GNRHR1, GRHR, HH7, LHRHR, LRHR
UniGene
Hs.407587
Gene Chromosome Location
Chr.4: 67737375-67756086 on Build GRCh38
Transcripts
NM_000406.2 NM_001012763.1

SNP Detail

SNP ID Chromosome Location
rs910861296 Chr.4: 67738719-67738719 on Build GRCh38
rs559826470 Chr.4: 67738625-67738625 on Build GRCh38
rs946430136 Chr.4: 67738669-67738670 on Build GRCh38
rs930727189 Chr.4: 67738737-67738737 on Build GRCh38
rs888347260 Chr.4: 67738838-67738838 on Build GRCh38
rs544007778 Chr.4: 67738905-67738905 on Build GRCh38
rs956192537 Chr.4: 67738958-67738958 on Build GRCh38
rs895131473 Chr.4: 67738865-67738865 on Build GRCh38
rs188539752 Chr.4: 67738859-67738859 on Build GRCh38
rs901017165 Chr.4: 67738815-67738815 on Build GRCh38
rs116200291 Chr.4: 67738836-67738836 on Build GRCh38
rs370116715 Chr.4: 67738721-67738721 on Build GRCh38
rs910922936 Chr.4: 67739012-67739012 on Build GRCh38
rs570095578 Chr.4: 67738867-67738867 on Build GRCh38
rs748061618 Chr.4: 67738662-67738662 on Build GRCh38
rs376543667 Chr.4: 67738697-67738697 on Build GRCh38
rs191511938 Chr.4: 67738946-67738946 on Build GRCh38
rs990891263 Chr.4: 67738581-67738581 on Build GRCh38
rs548462213 Chr.4: 67738789-67738789 on Build GRCh38
rs184230255 Chr.4: 67738964-67738964 on Build GRCh38
rs1025022848 Chr.4: 67738879-67738879 on Build GRCh38
rs184206874 Chr.4: 67738826-67738826 on Build GRCh38
rs771216443 Chr.4: 67738898-67738898 on Build GRCh38
rs188359092 Chr.4: 67738614-67738614 on Build GRCh38
rs912173198 Chr.4: 67738647-67738647 on Build GRCh38
rs978340988 Chr.4: 67738623-67738623 on Build GRCh38
rs191961829 Chr.4: 67738743-67738743 on Build GRCh38
rs990124825 Chr.4: 67739010-67739010 on Build GRCh38
rs557933290 Chr.4: 67738924-67738925 on Build GRCh38
rs12508464 Chr.4: 67738589-67738589 on Build GRCh38
rs978224882 Chr.4: 67738925-67738925 on Build GRCh38
rs989136388 Chr.4: 67738683-67738683 on Build GRCh38
rs974734835 Chr.4: 67739038-67739038 on Build GRCh38
rs747357066 Chr.4: 67738872-67738872 on Build GRCh38
rs528286579 Chr.4: 67738948-67738948 on Build GRCh38
rs1047883846 Chr.4: 67738774-67738774 on Build GRCh38
rs780997275 Chr.4: 67738618-67738618 on Build GRCh38
rs573018491 Chr.4: 67738986-67738986 on Build GRCh38
rs773416944 Chr.4: 67738847-67738847 on Build GRCh38
rs989503394 Chr.4: 67739006-67739006 on Build GRCh38
rs1005866651 Chr.4: 67738849-67738849 on Build GRCh38
rs886266241 Chr.4: 67738784-67738784 on Build GRCh38
rs183815234 Chr.4: 67738588-67738588 on Build GRCh38
rs1040763333 Chr.4: 67738788-67738788 on Build GRCh38
rs942480884 Chr.4: 67738772-67738772 on Build GRCh38
rs1003405979 Chr.4: 67738804-67738804 on Build GRCh38

View Full Product Details