Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70055256-70055731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTN1
Entrez Gene ID
3346
Gene Name
histatin 1
Gene Aliases
HIS1
UniGene
Hs.250959
Gene Chromosome Location
Chr.4: 70050415-70058848 on Build GRCh38
Transcripts
NM_002159.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671055 c.140G>A p.G47E Chr.4: 70055535-70055535 on Build GRCh38
5532458 c.149G>A p.G50E Chr.4: 70055544-70055544 on Build GRCh38
234253 c.121C>T p.R41* Chr.4: 70055516-70055516 on Build GRCh38
5823854 c.173G>A p.*58* Chr.4: 70055568-70055568 on Build GRCh38
6168175 c.112C>A p.H38N Chr.4: 70055507-70055507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375127098 Chr.4: 70055517-70055517 on Build GRCh38
rs61744465 Chr.4: 70055510-70055510 on Build GRCh38
rs370623231 Chr.4: 70055607-70055607 on Build GRCh38
rs746129703 Chr.4: 70055576-70055576 on Build GRCh38
rs989243511 Chr.4: 70055677-70055677 on Build GRCh38
rs558595582 Chr.4: 70055305-70055305 on Build GRCh38
rs199580841 Chr.4: 70055494-70055494 on Build GRCh38
rs774023728 Chr.4: 70055422-70055422 on Build GRCh38
rs770158535 Chr.4: 70055459-70055459 on Build GRCh38
rs561872744 Chr.4: 70055363-70055363 on Build GRCh38
rs189040461 Chr.4: 70055493-70055493 on Build GRCh38
rs749628232 Chr.4: 70055593-70055593 on Build GRCh38
rs368031080 Chr.4: 70055557-70055557 on Build GRCh38
rs11249473 Chr.4: 70055657-70055657 on Build GRCh38
rs763515608 Chr.4: 70055534-70055534 on Build GRCh38
rs556373181 Chr.4: 70055377-70055377 on Build GRCh38
rs769118208 Chr.4: 70055536-70055536 on Build GRCh38
rs6818370 Chr.4: 70055352-70055352 on Build GRCh38
rs775869475 Chr.4: 70055467-70055467 on Build GRCh38
rs2271644 Chr.4: 70055620-70055620 on Build GRCh38
rs761278274 Chr.4: 70055428-70055428 on Build GRCh38
rs367871166 Chr.4: 70055573-70055573 on Build GRCh38
rs181485765 Chr.4: 70055506-70055506 on Build GRCh38
rs184620440 Chr.4: 70055455-70055455 on Build GRCh38
rs762725571 Chr.4: 70055376-70055376 on Build GRCh38
rs537961920 Chr.4: 70055369-70055369 on Build GRCh38
rs371488004 Chr.4: 70055481-70055481 on Build GRCh38
rs1007649363 Chr.4: 70055541-70055541 on Build GRCh38
rs767076366 Chr.4: 70055433-70055433 on Build GRCh38
rs545712957 Chr.4: 70055703-70055703 on Build GRCh38
rs372745567 Chr.4: 70055574-70055574 on Build GRCh38
rs774868284 Chr.4: 70055606-70055606 on Build GRCh38
rs150657741 Chr.4: 70055565-70055565 on Build GRCh38
rs756351093 Chr.4: 70055587-70055587 on Build GRCh38
rs750042189 Chr.4: 70055504-70055504 on Build GRCh38
rs549359904 Chr.4: 70055680-70055680 on Build GRCh38
rs553938845 Chr.4: 70055675-70055675 on Build GRCh38
rs116978549 Chr.4: 70055524-70055524 on Build GRCh38
rs1022488378 Chr.4: 70055697-70055697 on Build GRCh38
rs767012944 Chr.4: 70055542-70055542 on Build GRCh38
rs779831486 Chr.4: 70055507-70055507 on Build GRCh38
rs759843819 Chr.4: 70055457-70055457 on Build GRCh38
rs746039493 Chr.4: 70055523-70055523 on Build GRCh38
rs377686351 Chr.4: 70055604-70055604 on Build GRCh38
rs763389433 Chr.4: 70055469-70055469 on Build GRCh38
rs745458170 Chr.4: 70055688-70055688 on Build GRCh38
rs374729257 Chr.4: 70055490-70055490 on Build GRCh38
rs542097009 Chr.4: 70055410-70055410 on Build GRCh38
rs750543129 Chr.4: 70055363-70055363 on Build GRCh38
rs72651134 Chr.4: 70055448-70055448 on Build GRCh38
rs770419218 Chr.4: 70055608-70055608 on Build GRCh38
rs775815012 Chr.4: 70055526-70055526 on Build GRCh38
rs778040971 Chr.4: 70055515-70055515 on Build GRCh38
rs868775954 Chr.4: 70055583-70055583 on Build GRCh38
rs758661576 Chr.4: 70055397-70055397 on Build GRCh38
rs560233461 Chr.4: 70055415-70055415 on Build GRCh38
rs771496456 Chr.4: 70055612-70055612 on Build GRCh38
rs13353559 Chr.4: 70055474-70055474 on Build GRCh38
rs201645043 Chr.4: 70055509-70055509 on Build GRCh38
rs750047560 Chr.4: 70055552-70055552 on Build GRCh38
rs997498344 Chr.4: 70055319-70055319 on Build GRCh38
rs2271643 Chr.4: 70055447-70055447 on Build GRCh38
rs35823441 Chr.4: 70055328-70055329 on Build GRCh38
rs73824787 Chr.4: 70055405-70055405 on Build GRCh38
rs371461805 Chr.4: 70055516-70055516 on Build GRCh38
rs753656464 Chr.4: 70055558-70055558 on Build GRCh38
rs760362084 Chr.4: 70055554-70055554 on Build GRCh38
rs570491653 Chr.4: 70055537-70055537 on Build GRCh38
rs780433839 Chr.4: 70055589-70055589 on Build GRCh38
rs535248155 Chr.4: 70055670-70055670 on Build GRCh38

View Full Product Details