Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70244732-70245243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN3
Entrez Gene ID
1448
Gene Name
casein kappa
Gene Aliases
CNS10, CSN10, CSNK, KCA
UniGene
Hs.54415
Gene Chromosome Location
Chr.4: 70238370-70251432 on Build GRCh38
Transcripts
NM_005212.2 XM_011531620.2 XM_017007761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3826121 c.54G>T p.L18F Chr.4: 70244873-70244873 on Build GRCh38
1753813 c.15T>A p.L5L Chr.4: 70244834-70244834 on Build GRCh38
341402 c.15T>C p.L5L Chr.4: 70244834-70244834 on Build GRCh38
7449331 c.34G>A p.A12T Chr.4: 70244853-70244853 on Build GRCh38
1057047 c.8G>T p.S3I Chr.4: 70244827-70244827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17850701 Chr.4: 70244825-70244825 on Build GRCh38
rs148200686 Chr.4: 70244845-70244845 on Build GRCh38
rs112561516 Chr.4: 70245130-70245130 on Build GRCh38
rs1012856422 Chr.4: 70244804-70244804 on Build GRCh38
rs969770408 Chr.4: 70244821-70244821 on Build GRCh38
rs549252375 Chr.4: 70244854-70244854 on Build GRCh38
rs537803187 Chr.4: 70244805-70244805 on Build GRCh38
rs746727688 Chr.4: 70244874-70244874 on Build GRCh38
rs1035464232 Chr.4: 70245105-70245105 on Build GRCh38
rs140151261 Chr.4: 70245047-70245047 on Build GRCh38
rs974584047 Chr.4: 70244951-70244951 on Build GRCh38
rs1002623531 Chr.4: 70245074-70245074 on Build GRCh38
rs757193660 Chr.4: 70244879-70244879 on Build GRCh38
rs758198366 Chr.4: 70244869-70244869 on Build GRCh38
rs974958730 Chr.4: 70244930-70244930 on Build GRCh38
rs147959002 Chr.4: 70245017-70245017 on Build GRCh38
rs965860510 Chr.4: 70244946-70244946 on Build GRCh38
rs188502316 Chr.4: 70245064-70245064 on Build GRCh38
rs939963671 Chr.4: 70245028-70245028 on Build GRCh38
rs952023913 Chr.4: 70244983-70244983 on Build GRCh38
rs192633784 Chr.4: 70244797-70244797 on Build GRCh38
rs777634156 Chr.4: 70244777-70244777 on Build GRCh38
rs576809215 Chr.4: 70244798-70244798 on Build GRCh38
rs933373203 Chr.4: 70244960-70244960 on Build GRCh38
rs906704876 Chr.4: 70245063-70245063 on Build GRCh38
rs1002623613 Chr.4: 70244848-70244848 on Build GRCh38
rs566182103 Chr.4: 70245137-70245137 on Build GRCh38
rs747902934 Chr.4: 70244772-70244772 on Build GRCh38
rs41292331 Chr.4: 70244771-70244771 on Build GRCh38
rs950110722 Chr.4: 70245044-70245044 on Build GRCh38
rs779067000 Chr.4: 70244788-70244789 on Build GRCh38
rs375124537 Chr.4: 70244863-70244863 on Build GRCh38
rs907877566 Chr.4: 70244987-70244987 on Build GRCh38
rs768868991 Chr.4: 70244823-70244823 on Build GRCh38
rs374280097 Chr.4: 70244865-70244865 on Build GRCh38
rs753400244 Chr.4: 70244861-70244861 on Build GRCh38
rs1029235352 Chr.4: 70245208-70245208 on Build GRCh38
rs368102586 Chr.4: 70244775-70244775 on Build GRCh38
rs777614391 Chr.4: 70244870-70244870 on Build GRCh38
rs13103289 Chr.4: 70244922-70244922 on Build GRCh38
rs1055093846 Chr.4: 70245176-70245176 on Build GRCh38
rs768701343 Chr.4: 70244910-70244910 on Build GRCh38
rs774574281 Chr.4: 70244839-70244839 on Build GRCh38
rs13122200 Chr.4: 70245135-70245135 on Build GRCh38
rs41292335 Chr.4: 70245030-70245030 on Build GRCh38
rs41292333 Chr.4: 70244963-70244963 on Build GRCh38
rs1023816636 Chr.4: 70244811-70244811 on Build GRCh38
rs891456437 Chr.4: 70245118-70245118 on Build GRCh38
rs746929413 Chr.4: 70244787-70244787 on Build GRCh38
rs781069696 Chr.4: 70244886-70244886 on Build GRCh38
rs560126838 Chr.4: 70244897-70244897 on Build GRCh38
rs184144813 Chr.4: 70244945-70244945 on Build GRCh38
rs940718743 Chr.4: 70245010-70245010 on Build GRCh38
rs762142502 Chr.4: 70244841-70244841 on Build GRCh38
rs140888032 Chr.4: 70244789-70244789 on Build GRCh38
rs749393487 Chr.4: 70244893-70244893 on Build GRCh38
rs534294695 Chr.4: 70244928-70244928 on Build GRCh38
rs746098050 Chr.4: 70244798-70244798 on Build GRCh38
rs62308380 Chr.4: 70245149-70245149 on Build GRCh38
rs974695700 Chr.4: 70245154-70245154 on Build GRCh38
rs1044593298 Chr.4: 70245119-70245119 on Build GRCh38
rs1009960689 Chr.4: 70244939-70244939 on Build GRCh38
rs531604175 Chr.4: 70245020-70245020 on Build GRCh38
rs556310390 Chr.4: 70244849-70244849 on Build GRCh38
rs985082449 Chr.4: 70245016-70245016 on Build GRCh38
rs761242108 Chr.4: 70244853-70244853 on Build GRCh38
rs921791817 Chr.4: 70244932-70244932 on Build GRCh38
rs41292337 Chr.4: 70245036-70245036 on Build GRCh38

View Full Product Details