Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70666202-70666722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JCHAIN
Entrez Gene ID
3512
Gene Name
joining chain of multimeric IgA and IgM
Gene Aliases
IGCJ, IGJ, JCH
UniGene
Hs.643431
Gene Chromosome Location
Chr.4: 70655541-70676901 on Build GRCh38
Transcripts
NM_144646.3 XM_006714211.2 XM_006714212.2 XM_011531925.1 XM_011531926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6101050 c.64G>C p.A22P Chr.4: 70666427-70666427 on Build GRCh38
4125764 c.35C>T p.A12V Chr.4: 70666456-70666456 on Build GRCh38
6168150 c.30C>A p.V10V Chr.4: 70666461-70666461 on Build GRCh38
5619982 c.38T>G p.V13G Chr.4: 70666453-70666453 on Build GRCh38
3918098 c.3G>A p.M1I Chr.4: 70666488-70666488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766259333 Chr.4: 70666482-70666482 on Build GRCh38
rs1048512312 Chr.4: 70666518-70666518 on Build GRCh38
rs1053893536 Chr.4: 70666237-70666237 on Build GRCh38
rs985649940 Chr.4: 70666411-70666411 on Build GRCh38
rs954254598 Chr.4: 70666328-70666328 on Build GRCh38
rs1017093956 Chr.4: 70666696-70666696 on Build GRCh38
rs756669364 Chr.4: 70666455-70666455 on Build GRCh38
rs375452642 Chr.4: 70666522-70666522 on Build GRCh38
rs980058063 Chr.4: 70666500-70666500 on Build GRCh38
rs760620807 Chr.4: 70666470-70666470 on Build GRCh38
rs770363099 Chr.4: 70666439-70666439 on Build GRCh38
rs972452200 Chr.4: 70666420-70666420 on Build GRCh38
rs746378954 Chr.4: 70666435-70666435 on Build GRCh38
rs373866287 Chr.4: 70666454-70666454 on Build GRCh38
rs758475498 Chr.4: 70666492-70666492 on Build GRCh38
rs757058522 Chr.4: 70666639-70666639 on Build GRCh38
rs767802564 Chr.4: 70666243-70666243 on Build GRCh38
rs752755197 Chr.4: 70666483-70666483 on Build GRCh38
rs959286505 Chr.4: 70666248-70666248 on Build GRCh38
rs186205050 Chr.4: 70666320-70666320 on Build GRCh38
rs746182777 Chr.4: 70666531-70666531 on Build GRCh38
rs377277594 Chr.4: 70666456-70666456 on Build GRCh38
rs377627799 Chr.4: 70666697-70666697 on Build GRCh38
rs543140564 Chr.4: 70666287-70666287 on Build GRCh38
rs1028305789 Chr.4: 70666617-70666617 on Build GRCh38
rs750348058 Chr.4: 70666379-70666379 on Build GRCh38
rs190949649 Chr.4: 70666346-70666346 on Build GRCh38
rs1055030307 Chr.4: 70666611-70666611 on Build GRCh38
rs1005749379 Chr.4: 70666673-70666673 on Build GRCh38
rs761635518 Chr.4: 70666377-70666377 on Build GRCh38
rs966520541 Chr.4: 70666315-70666315 on Build GRCh38
rs182852639 Chr.4: 70666428-70666428 on Build GRCh38
rs901186587 Chr.4: 70666559-70666559 on Build GRCh38
rs925417746 Chr.4: 70666337-70666337 on Build GRCh38
rs373220622 Chr.4: 70666508-70666508 on Build GRCh38
rs781697571 Chr.4: 70666511-70666511 on Build GRCh38
rs868038348 Chr.4: 70666473-70666473 on Build GRCh38
rs780578818 Chr.4: 70666534-70666534 on Build GRCh38
rs573278772 Chr.4: 70666343-70666343 on Build GRCh38
rs767490447 Chr.4: 70666378-70666378 on Build GRCh38
rs767675649 Chr.4: 70666228-70666228 on Build GRCh38
rs567738132 Chr.4: 70666638-70666638 on Build GRCh38
rs530769246 Chr.4: 70666558-70666558 on Build GRCh38
rs1009643501 Chr.4: 70666281-70666281 on Build GRCh38
rs377548643 Chr.4: 70666529-70666529 on Build GRCh38
rs145363741 Chr.4: 70666451-70666451 on Build GRCh38
rs558935845 Chr.4: 70666352-70666352 on Build GRCh38
rs991863788 Chr.4: 70666266-70666266 on Build GRCh38
rs998456031 Chr.4: 70666319-70666319 on Build GRCh38
rs186360969 Chr.4: 70666657-70666657 on Build GRCh38
rs755119530 Chr.4: 70666382-70666382 on Build GRCh38
rs373731911 Chr.4: 70666629-70666629 on Build GRCh38
rs749949804 Chr.4: 70666554-70666554 on Build GRCh38
rs936861272 Chr.4: 70666390-70666390 on Build GRCh38
rs763679732 Chr.4: 70666501-70666503 on Build GRCh38
rs945483669 Chr.4: 70666557-70666557 on Build GRCh38
rs747375918 Chr.4: 70666418-70666418 on Build GRCh38
rs76178812 Chr.4: 70666448-70666448 on Build GRCh38
rs575841104 Chr.4: 70666270-70666270 on Build GRCh38
rs1006070484 Chr.4: 70666571-70666571 on Build GRCh38
rs112770439 Chr.4: 70666426-70666426 on Build GRCh38
rs551654637 Chr.4: 70666446-70666446 on Build GRCh38
rs750151829 Chr.4: 70666459-70666459 on Build GRCh38
rs758574444 Chr.4: 70666403-70666403 on Build GRCh38
rs1013663077 Chr.4: 70666684-70666684 on Build GRCh38
rs998568430 Chr.4: 70666598-70666598 on Build GRCh38
rs753151015 Chr.4: 70666441-70666441 on Build GRCh38
rs893919511 Chr.4: 70666267-70666267 on Build GRCh38
rs1041089077 Chr.4: 70666461-70666461 on Build GRCh38
rs752765737 Chr.4: 70666398-70666398 on Build GRCh38
rs373482441 Chr.4: 70666434-70666434 on Build GRCh38
rs763752131 Chr.4: 70666376-70666376 on Build GRCh38
rs369615126 Chr.4: 70666493-70666493 on Build GRCh38
rs778969334 Chr.4: 70666384-70666384 on Build GRCh38
rs778020112 Chr.4: 70666410-70666410 on Build GRCh38
rs901167397 Chr.4: 70666307-70666307 on Build GRCh38

View Full Product Details