Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88725812-88726356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
UniGene
Hs.714536
Gene Chromosome Location
Chr.4: 88725954-89111398 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1 NR_002806.2

Gene Symbol
FAM13A-AS1
Entrez Gene ID
285512
Gene Name
FAM13A antisense RNA 1
Gene Aliases
FAM13A1OS, FAM13AOS, NCRNA00039
UniGene
Hs.744957
Gene Chromosome Location
Chr.4: 88709789-88730103 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1 NR_002806.2

SNP Detail

SNP ID Chromosome Location
rs999613873 Chr.4: 88725995-88725995 on Build GRCh38
rs2860500 Chr.4: 88725919-88725919 on Build GRCh38
rs553071071 Chr.4: 88726047-88726047 on Build GRCh38
rs66508874 Chr.4: 88726300-88726301 on Build GRCh38
rs538461993 Chr.4: 88726256-88726256 on Build GRCh38
rs926032293 Chr.4: 88726161-88726161 on Build GRCh38
rs749381526 Chr.4: 88725903-88725904 on Build GRCh38
rs779930352 Chr.4: 88725882-88725884 on Build GRCh38
rs190336833 Chr.4: 88726325-88726325 on Build GRCh38
rs116068244 Chr.4: 88725917-88725917 on Build GRCh38
rs552184553 Chr.4: 88725926-88725926 on Build GRCh38
rs908981245 Chr.4: 88725902-88725902 on Build GRCh38
rs555198100 Chr.4: 88726113-88726113 on Build GRCh38
rs879579629 Chr.4: 88726070-88726071 on Build GRCh38
rs979297236 Chr.4: 88726138-88726138 on Build GRCh38
rs548007490 Chr.4: 88726205-88726208 on Build GRCh38
rs965503288 Chr.4: 88726242-88726242 on Build GRCh38
rs913250680 Chr.4: 88726117-88726118 on Build GRCh38
rs940440071 Chr.4: 88725906-88725906 on Build GRCh38
rs985602435 Chr.4: 88725950-88725950 on Build GRCh38
rs377444925 Chr.4: 88726211-88726211 on Build GRCh38
rs997318851 Chr.4: 88726084-88726084 on Build GRCh38
rs1052532160 Chr.4: 88726330-88726330 on Build GRCh38
rs34123244 Chr.4: 88725981-88725983 on Build GRCh38
rs916857981 Chr.4: 88726292-88726292 on Build GRCh38
rs538206852 Chr.4: 88726304-88726305 on Build GRCh38
rs1026573640 Chr.4: 88726216-88726216 on Build GRCh38
rs2904163 Chr.4: 88725918-88725918 on Build GRCh38
rs566287256 Chr.4: 88726061-88726061 on Build GRCh38
rs1006570261 Chr.4: 88726228-88726228 on Build GRCh38
rs1056650738 Chr.4: 88725920-88725920 on Build GRCh38
rs1043718608 Chr.4: 88725962-88725962 on Build GRCh38
rs953301367 Chr.4: 88726128-88726128 on Build GRCh38
rs952425848 Chr.4: 88725947-88725947 on Build GRCh38
rs533872390 Chr.4: 88726301-88726304 on Build GRCh38
rs57101865 Chr.4: 88726302-88726303 on Build GRCh38
rs140733471 Chr.4: 88726299-88726300 on Build GRCh38
rs376949935 Chr.4: 88726188-88726188 on Build GRCh38
rs999898135 Chr.4: 88726324-88726324 on Build GRCh38
rs946058690 Chr.4: 88726133-88726133 on Build GRCh38
rs1018419826 Chr.4: 88726234-88726235 on Build GRCh38
rs757046149 Chr.4: 88725939-88725939 on Build GRCh38
rs1043135446 Chr.4: 88725861-88725864 on Build GRCh38
rs72119891 Chr.4: 88725983-88725985 on Build GRCh38
rs931647326 Chr.4: 88725886-88725886 on Build GRCh38
rs748548826 Chr.4: 88725971-88725971 on Build GRCh38
rs984448279 Chr.4: 88725888-88725888 on Build GRCh38
rs948321658 Chr.4: 88726293-88726293 on Build GRCh38
rs74629576 Chr.4: 88726220-88726220 on Build GRCh38
rs894958968 Chr.4: 88726200-88726200 on Build GRCh38
rs796387492 Chr.4: 88726250-88726251 on Build GRCh38
rs767419469 Chr.4: 88725989-88725990 on Build GRCh38
rs1026733107 Chr.4: 88725942-88725943 on Build GRCh38
rs946148404 Chr.4: 88725845-88725845 on Build GRCh38
rs184410907 Chr.4: 88726079-88726079 on Build GRCh38
rs971843556 Chr.4: 88726219-88726219 on Build GRCh38
rs577179887 Chr.4: 88726303-88726303 on Build GRCh38
rs939536718 Chr.4: 88726257-88726257 on Build GRCh38
rs749373624 Chr.4: 88725980-88725981 on Build GRCh38
rs570330382 Chr.4: 88725977-88725977 on Build GRCh38
rs937376574 Chr.4: 88726189-88726189 on Build GRCh38
rs370500675 Chr.4: 88726251-88726252 on Build GRCh38
rs967996468 Chr.4: 88726301-88726302 on Build GRCh38
rs903719642 Chr.4: 88725885-88725887 on Build GRCh38
rs376495470 Chr.4: 88725925-88725925 on Build GRCh38
rs1046299339 Chr.4: 88726204-88726205 on Build GRCh38
rs781015843 Chr.4: 88726201-88726201 on Build GRCh38
rs1031051960 Chr.4: 88726017-88726017 on Build GRCh38
rs576715751 Chr.4: 88726209-88726209 on Build GRCh38
rs200852518 Chr.4: 88725980-88725982 on Build GRCh38
rs1044155470 Chr.4: 88726299-88726299 on Build GRCh38
rs752584908 Chr.4: 88725997-88725998 on Build GRCh38
rs541134670 Chr.4: 88726304-88726304 on Build GRCh38
rs895374914 Chr.4: 88725927-88725927 on Build GRCh38
rs903968569 Chr.4: 88725966-88725966 on Build GRCh38
rs1059122 Chr.4: 88726273-88726273 on Build GRCh38
rs534736882 Chr.4: 88726044-88726044 on Build GRCh38
rs1054979664 Chr.4: 88726192-88726192 on Build GRCh38

View Full Product Details