Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88726247-88726758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
UniGene
Hs.714536
Gene Chromosome Location
Chr.4: 88725954-89111398 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1 NR_002806.2

Gene Symbol
FAM13A-AS1
Entrez Gene ID
285512
Gene Name
FAM13A antisense RNA 1
Gene Aliases
FAM13A1OS, FAM13AOS, NCRNA00039
UniGene
Hs.744957
Gene Chromosome Location
Chr.4: 88709789-88730103 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1 NR_002806.2

SNP Detail

SNP ID Chromosome Location
rs66508874 Chr.4: 88726300-88726301 on Build GRCh38
rs201337586 Chr.4: 88726371-88726372 on Build GRCh38
rs552158797 Chr.4: 88726622-88726622 on Build GRCh38
rs541660167 Chr.4: 88726396-88726396 on Build GRCh38
rs190336833 Chr.4: 88726325-88726325 on Build GRCh38
rs1052415806 Chr.4: 88726676-88726676 on Build GRCh38
rs75415222 Chr.4: 88726649-88726649 on Build GRCh38
rs779541998 Chr.4: 88726393-88726393 on Build GRCh38
rs1048066976 Chr.4: 88726491-88726491 on Build GRCh38
rs971676461 Chr.4: 88726521-88726521 on Build GRCh38
rs58657905 Chr.4: 88726369-88726370 on Build GRCh38
rs935361905 Chr.4: 88726651-88726651 on Build GRCh38
rs918517657 Chr.4: 88726441-88726441 on Build GRCh38
rs769173472 Chr.4: 88726538-88726538 on Build GRCh38
rs530127451 Chr.4: 88726363-88726363 on Build GRCh38
rs978795401 Chr.4: 88726350-88726350 on Build GRCh38
rs1052532160 Chr.4: 88726330-88726330 on Build GRCh38
rs763386420 Chr.4: 88726487-88726491 on Build GRCh38
rs916857981 Chr.4: 88726292-88726292 on Build GRCh38
rs1043991184 Chr.4: 88726628-88726628 on Build GRCh38
rs538206852 Chr.4: 88726304-88726305 on Build GRCh38
rs958682915 Chr.4: 88726721-88726721 on Build GRCh38
rs533872390 Chr.4: 88726301-88726304 on Build GRCh38
rs57101865 Chr.4: 88726302-88726303 on Build GRCh38
rs3067631 Chr.4: 88726651-88726651 on Build GRCh38
rs1006622543 Chr.4: 88726540-88726540 on Build GRCh38
rs953455107 Chr.4: 88726381-88726381 on Build GRCh38
rs140733471 Chr.4: 88726299-88726300 on Build GRCh38
rs901955170 Chr.4: 88726694-88726694 on Build GRCh38
rs202105612 Chr.4: 88726652-88726653 on Build GRCh38
rs999898135 Chr.4: 88726324-88726324 on Build GRCh38
rs992319827 Chr.4: 88726541-88726541 on Build GRCh38
rs397797371 Chr.4: 88726377-88726378 on Build GRCh38
rs530862508 Chr.4: 88726573-88726573 on Build GRCh38
rs375236629 Chr.4: 88726477-88726479 on Build GRCh38
rs916446710 Chr.4: 88726372-88726372 on Build GRCh38
rs997373108 Chr.4: 88726352-88726352 on Build GRCh38
rs4210 Chr.4: 88726648-88726649 on Build GRCh38
rs752196019 Chr.4: 88726488-88726489 on Build GRCh38
rs888094261 Chr.4: 88726504-88726504 on Build GRCh38
rs779352056 Chr.4: 88726589-88726589 on Build GRCh38
rs557340270 Chr.4: 88726623-88726627 on Build GRCh38
rs901902887 Chr.4: 88726347-88726347 on Build GRCh38
rs948321658 Chr.4: 88726293-88726293 on Build GRCh38
rs201689335 Chr.4: 88726378-88726378 on Build GRCh38
rs933386575 Chr.4: 88726705-88726705 on Build GRCh38
rs11323 Chr.4: 88726423-88726423 on Build GRCh38
rs925496534 Chr.4: 88726635-88726635 on Build GRCh38
rs748736524 Chr.4: 88726522-88726522 on Build GRCh38
rs869205212 Chr.4: 88726651-88726652 on Build GRCh38
rs577179887 Chr.4: 88726303-88726303 on Build GRCh38
rs1039419354 Chr.4: 88726559-88726559 on Build GRCh38
rs967996468 Chr.4: 88726301-88726302 on Build GRCh38
rs916750768 Chr.4: 88726591-88726591 on Build GRCh38
rs1044155470 Chr.4: 88726299-88726299 on Build GRCh38
rs774643000 Chr.4: 88726611-88726612 on Build GRCh38
rs948180950 Chr.4: 88726616-88726616 on Build GRCh38
rs541134670 Chr.4: 88726304-88726304 on Build GRCh38
rs57157914 Chr.4: 88726480-88726482 on Build GRCh38
rs1059122 Chr.4: 88726273-88726273 on Build GRCh38
rs570442099 Chr.4: 88726642-88726642 on Build GRCh38
rs1020840242 Chr.4: 88726641-88726642 on Build GRCh38

View Full Product Details