Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 89245309-89245803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRIN3
Entrez Gene ID
285513
Gene Name
GPRIN family member 3
Gene Aliases
GRIN3
UniGene
Hs.100912
Gene Chromosome Location
Chr.4: 89237666-89308010 on Build GRCh38
Transcripts
NM_198281.2 XM_005262936.3 XM_005262937.4 XM_017008043.1 XM_017008044.1

SNP Detail

SNP ID Chromosome Location
rs6532145 Chr.4: 89245622-89245622 on Build GRCh38
rs534842553 Chr.4: 89245486-89245486 on Build GRCh38
rs6532146 Chr.4: 89245627-89245627 on Build GRCh38
rs910121408 Chr.4: 89245453-89245453 on Build GRCh38
rs767301757 Chr.4: 89245602-89245602 on Build GRCh38
rs1031557414 Chr.4: 89245765-89245765 on Build GRCh38
rs1028409018 Chr.4: 89245406-89245406 on Build GRCh38
rs904384080 Chr.4: 89245681-89245681 on Build GRCh38
rs6532148 Chr.4: 89245770-89245770 on Build GRCh38
rs768464391 Chr.4: 89245394-89245394 on Build GRCh38
rs985171197 Chr.4: 89245552-89245552 on Build GRCh38
rs35653127 Chr.4: 89245769-89245770 on Build GRCh38
rs190249229 Chr.4: 89245598-89245598 on Build GRCh38
rs879488997 Chr.4: 89245769-89245770 on Build GRCh38
rs570110138 Chr.4: 89245338-89245338 on Build GRCh38
rs940276397 Chr.4: 89245472-89245472 on Build GRCh38
rs972996698 Chr.4: 89245518-89245518 on Build GRCh38
rs731926 Chr.4: 89245528-89245528 on Build GRCh38
rs147008255 Chr.4: 89245435-89245435 on Build GRCh38
rs536935284 Chr.4: 89245372-89245372 on Build GRCh38
rs528878743 Chr.4: 89245349-89245349 on Build GRCh38
rs984338227 Chr.4: 89245448-89245448 on Build GRCh38
rs6532147 Chr.4: 89245769-89245769 on Build GRCh38
rs938365656 Chr.4: 89245550-89245550 on Build GRCh38
rs952339177 Chr.4: 89245532-89245532 on Build GRCh38
rs558821865 Chr.4: 89245384-89245384 on Build GRCh38
rs547555243 Chr.4: 89245715-89245715 on Build GRCh38
rs761692633 Chr.4: 89245531-89245531 on Build GRCh38
rs910954656 Chr.4: 89245579-89245579 on Build GRCh38
rs895263058 Chr.4: 89245388-89245388 on Build GRCh38
rs961585897 Chr.4: 89245506-89245506 on Build GRCh38
rs913076557 Chr.4: 89245665-89245665 on Build GRCh38
rs769176614 Chr.4: 89245411-89245411 on Build GRCh38
rs1042894688 Chr.4: 89245672-89245672 on Build GRCh38
rs1055602495 Chr.4: 89245355-89245355 on Build GRCh38

View Full Product Details