Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 93224522-93225023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRID2
Entrez Gene ID
2895
Gene Name
glutamate ionotropic receptor delta type subunit 2
Gene Aliases
GluD2, SCAR18
UniGene
Hs.162727
Gene Chromosome Location
Chr.4: 92304249-93810330 on Build GRCh38
Transcripts
NM_001286838.1 NM_001510.3 XM_011531893.2 XM_011531894.2 XM_011531895.2 XM_017008118.1 XM_017008119.1 XM_017008120.1 XM_017008121.1 XM_017008122.1 XM_017008123.1 XM_017008124.1 XM_017008125.1 XM_017008126.1 XM_017008127.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
281538 c.1031A>G p.D344G Chr.4: 93224681-93224681 on Build GRCh38
1694683 c.968C>T p.S323F Chr.4: 93224618-93224618 on Build GRCh38
5002570 c.1004C>A p.A335D Chr.4: 93224654-93224654 on Build GRCh38
3607056 c.1103C>T p.S368F Chr.4: 93224753-93224753 on Build GRCh38
735294 c.1092G>T p.Q364H Chr.4: 93224742-93224742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370239617 Chr.4: 93224996-93224997 on Build GRCh38
rs1002043071 Chr.4: 93224823-93224823 on Build GRCh38
rs542465122 Chr.4: 93224649-93224649 on Build GRCh38
rs36090230 Chr.4: 93224797-93224797 on Build GRCh38
rs778277476 Chr.4: 93224569-93224569 on Build GRCh38
rs771190813 Chr.4: 93224851-93224851 on Build GRCh38
rs761727838 Chr.4: 93224631-93224631 on Build GRCh38
rs529275890 Chr.4: 93224549-93224549 on Build GRCh38
rs757742604 Chr.4: 93224694-93224694 on Build GRCh38
rs191438581 Chr.4: 93224760-93224760 on Build GRCh38
rs752197003 Chr.4: 93224827-93224827 on Build GRCh38
rs923547251 Chr.4: 93224939-93224939 on Build GRCh38
rs1047863100 Chr.4: 93224950-93224950 on Build GRCh38
rs1047980661 Chr.4: 93224958-93224958 on Build GRCh38
rs762975898 Chr.4: 93224604-93224604 on Build GRCh38
rs747484049 Chr.4: 93224573-93224573 on Build GRCh38
rs772088564 Chr.4: 93224793-93224793 on Build GRCh38
rs3796675 Chr.4: 93224942-93224942 on Build GRCh38
rs771313188 Chr.4: 93224574-93224574 on Build GRCh38
rs1038124259 Chr.4: 93224937-93224937 on Build GRCh38
rs781512821 Chr.4: 93224707-93224707 on Build GRCh38
rs143138111 Chr.4: 93224696-93224696 on Build GRCh38
rs569567401 Chr.4: 93224974-93224974 on Build GRCh38
rs546802285 Chr.4: 93224884-93224884 on Build GRCh38
rs750295354 Chr.4: 93224640-93224640 on Build GRCh38
rs373866971 Chr.4: 93224639-93224639 on Build GRCh38
rs748097566 Chr.4: 93224779-93224779 on Build GRCh38
rs950314780 Chr.4: 93224963-93224963 on Build GRCh38
rs910870299 Chr.4: 93224922-93224923 on Build GRCh38
rs1046177660 Chr.4: 93224969-93224969 on Build GRCh38
rs898326674 Chr.4: 93224801-93224801 on Build GRCh38
rs564359823 Chr.4: 93224811-93224811 on Build GRCh38
rs752101789 Chr.4: 93224676-93224676 on Build GRCh38
rs533433373 Chr.4: 93224841-93224841 on Build GRCh38
rs754477454 Chr.4: 93224566-93224566 on Build GRCh38
rs544360335 Chr.4: 93224749-93224749 on Build GRCh38
rs759177123 Chr.4: 93224804-93224804 on Build GRCh38
rs1033058700 Chr.4: 93224989-93224989 on Build GRCh38
rs770047659 Chr.4: 93224583-93224583 on Build GRCh38
rs148758653 Chr.4: 93224748-93224748 on Build GRCh38
rs768618901 Chr.4: 93224606-93224606 on Build GRCh38
rs560690289 Chr.4: 93224896-93224896 on Build GRCh38
rs549639581 Chr.4: 93224971-93224971 on Build GRCh38
rs746058310 Chr.4: 93224581-93224581 on Build GRCh38
rs886711512 Chr.4: 93224956-93224956 on Build GRCh38
rs1020593988 Chr.4: 93224870-93224870 on Build GRCh38
rs774338834 Chr.4: 93224774-93224774 on Build GRCh38
rs199766182 Chr.4: 93224795-93224795 on Build GRCh38
rs930806497 Chr.4: 93224948-93224948 on Build GRCh38
rs778353741 Chr.4: 93224829-93224829 on Build GRCh38
rs746179603 Chr.4: 93224727-93224727 on Build GRCh38
rs775708984 Chr.4: 93224603-93224603 on Build GRCh38
rs760480753 Chr.4: 93224797-93224797 on Build GRCh38
rs367665351 Chr.4: 93224579-93224579 on Build GRCh38
rs575224950 Chr.4: 93224750-93224750 on Build GRCh38
rs943571386 Chr.4: 93224922-93224922 on Build GRCh38
rs776408851 Chr.4: 93224798-93224798 on Build GRCh38
rs774103181 Chr.4: 93224821-93224821 on Build GRCh38
rs974929725 Chr.4: 93224933-93224933 on Build GRCh38
rs371885400 Chr.4: 93224644-93224644 on Build GRCh38
rs150053332 Chr.4: 93224628-93224628 on Build GRCh38
rs955288704 Chr.4: 93224840-93224840 on Build GRCh38
rs1014686370 Chr.4: 93224594-93224594 on Build GRCh38
rs754454093 Chr.4: 93224658-93224658 on Build GRCh38
rs144350770 Chr.4: 93224666-93224666 on Build GRCh38
rs960602232 Chr.4: 93224842-93224842 on Build GRCh38
rs562865308 Chr.4: 93224767-93224767 on Build GRCh38
rs778344399 Chr.4: 93224669-93224669 on Build GRCh38
rs925015656 Chr.4: 93224905-93224905 on Build GRCh38
rs760570729 Chr.4: 93224641-93224641 on Build GRCh38
rs555553103 Chr.4: 93224988-93224989 on Build GRCh38
rs562449934 Chr.4: 93224673-93224673 on Build GRCh38

View Full Product Details