Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99088611-99089112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH5
Entrez Gene ID
128
Gene Name
alcohol dehydrogenase 5 (class III), chi polypeptide
Gene Aliases
ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p
UniGene
Hs.78989
Gene Chromosome Location
Chr.4: 99070978-99088788 on Build GRCh38
Transcripts
NM_000671.4 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_000671.4 NR_037884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163188 c.10G>A p.E4K Chr.4: 99088691-99088691 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760608338 Chr.4: 99088652-99088652 on Build GRCh38
rs570148536 Chr.4: 99088758-99088758 on Build GRCh38
rs901073928 Chr.4: 99088928-99088928 on Build GRCh38
rs752570947 Chr.4: 99088715-99088715 on Build GRCh38
rs776488021 Chr.4: 99088657-99088657 on Build GRCh38
rs758146818 Chr.4: 99088680-99088680 on Build GRCh38
rs760941592 Chr.4: 99088696-99088696 on Build GRCh38
rs550931476 Chr.4: 99088681-99088681 on Build GRCh38
rs2851301 Chr.4: 99088976-99088976 on Build GRCh38
rs759597389 Chr.4: 99088705-99088705 on Build GRCh38
rs749858663 Chr.4: 99088685-99088685 on Build GRCh38
rs879180132 Chr.4: 99088976-99088977 on Build GRCh38
rs762060028 Chr.4: 99088643-99088643 on Build GRCh38
rs2602899 Chr.4: 99088977-99088977 on Build GRCh38
rs759893611 Chr.4: 99088713-99088713 on Build GRCh38
rs1042867061 Chr.4: 99088786-99088786 on Build GRCh38
rs869301375 Chr.4: 99088810-99088811 on Build GRCh38
rs753780632 Chr.4: 99088658-99088658 on Build GRCh38
rs748530011 Chr.4: 99088726-99088726 on Build GRCh38
rs938973931 Chr.4: 99088877-99088877 on Build GRCh38
rs569053250 Chr.4: 99088683-99088683 on Build GRCh38
rs78551514 Chr.4: 99088944-99088944 on Build GRCh38
rs1007662826 Chr.4: 99088781-99088781 on Build GRCh38
rs909405693 Chr.4: 99088707-99088707 on Build GRCh38
rs928891600 Chr.4: 99088866-99088866 on Build GRCh38
rs771247299 Chr.4: 99088686-99088686 on Build GRCh38
rs1024472137 Chr.4: 99088950-99088950 on Build GRCh38
rs553928451 Chr.4: 99088857-99088857 on Build GRCh38
rs201289075 Chr.4: 99088649-99088649 on Build GRCh38
rs774690450 Chr.4: 99088639-99088639 on Build GRCh38
rs948949880 Chr.4: 99088717-99088717 on Build GRCh38
rs926404755 Chr.4: 99088862-99088862 on Build GRCh38
rs918908293 Chr.4: 99088883-99088883 on Build GRCh38
rs372642917 Chr.4: 99088718-99088718 on Build GRCh38
rs928924362 Chr.4: 99089061-99089061 on Build GRCh38
rs369658800 Chr.4: 99088648-99088648 on Build GRCh38
rs1044668238 Chr.4: 99088794-99088794 on Build GRCh38
rs139641644 Chr.4: 99089044-99089044 on Build GRCh38
rs770985624 Chr.4: 99088697-99088697 on Build GRCh38
rs891775760 Chr.4: 99088941-99088941 on Build GRCh38
rs548369237 Chr.4: 99088931-99088931 on Build GRCh38
rs536167515 Chr.4: 99088845-99088845 on Build GRCh38
rs766223562 Chr.4: 99088891-99088891 on Build GRCh38
rs980662840 Chr.4: 99088997-99088997 on Build GRCh38
rs185048668 Chr.4: 99088901-99088901 on Build GRCh38
rs397698549 Chr.4: 99088809-99088810 on Build GRCh38
rs890303182 Chr.4: 99088900-99088900 on Build GRCh38
rs994123522 Chr.4: 99088937-99088937 on Build GRCh38
rs1005274112 Chr.4: 99088967-99088967 on Build GRCh38
rs888148213 Chr.4: 99088950-99088951 on Build GRCh38
rs11568816 Chr.4: 99088772-99088772 on Build GRCh38
rs1154400 Chr.4: 99088859-99088859 on Build GRCh38
rs546023363 Chr.4: 99088875-99088875 on Build GRCh38
rs779296852 Chr.4: 99088691-99088691 on Build GRCh38
rs370136358 Chr.4: 99088700-99088700 on Build GRCh38
rs759544558 Chr.4: 99088669-99088669 on Build GRCh38
rs778197850 Chr.4: 99088728-99088728 on Build GRCh38
rs1000646028 Chr.4: 99088821-99088821 on Build GRCh38
rs114145805 Chr.4: 99088898-99088898 on Build GRCh38
rs769859421 Chr.4: 99088747-99088747 on Build GRCh38
rs968753380 Chr.4: 99088808-99088808 on Build GRCh38
rs761372030 Chr.4: 99088663-99088663 on Build GRCh38
rs776874897 Chr.4: 99088740-99088740 on Build GRCh38
rs29001342 Chr.4: 99088893-99088893 on Build GRCh38
rs533696123 Chr.4: 99088750-99088750 on Build GRCh38
rs574654425 Chr.4: 99088802-99088802 on Build GRCh38
rs751886471 Chr.4: 99088699-99088700 on Build GRCh38
rs1031664875 Chr.4: 99088823-99088823 on Build GRCh38
rs772309402 Chr.4: 99088695-99088695 on Build GRCh38
rs759474707 Chr.4: 99088907-99088907 on Build GRCh38
rs762970413 Chr.4: 99088748-99088748 on Build GRCh38
rs913273016 Chr.4: 99088897-99088897 on Build GRCh38
rs960562444 Chr.4: 99089006-99089006 on Build GRCh38
rs753167271 Chr.4: 99088722-99088723 on Build GRCh38
rs992991917 Chr.4: 99088693-99088693 on Build GRCh38
rs1018073716 Chr.4: 99088789-99088789 on Build GRCh38
rs987389844 Chr.4: 99089079-99089079 on Build GRCh38
rs747204106 Chr.4: 99088865-99088865 on Build GRCh38
rs751227485 Chr.4: 99088720-99088720 on Build GRCh38
rs199747738 Chr.4: 99088724-99088724 on Build GRCh38
rs765219080 Chr.4: 99088708-99088708 on Build GRCh38
rs768038825 Chr.4: 99088722-99088722 on Build GRCh38
rs755493701 Chr.4: 99088725-99088725 on Build GRCh38
rs780738152 Chr.4: 99088684-99088684 on Build GRCh38
rs956518848 Chr.4: 99088842-99088842 on Build GRCh38
rs994253161 Chr.4: 99088812-99088812 on Build GRCh38
rs376403212 Chr.4: 99088723-99088723 on Build GRCh38
rs749127384 Chr.4: 99088888-99088888 on Build GRCh38
rs1035884585 Chr.4: 99089000-99089000 on Build GRCh38
rs1011639114 Chr.4: 99088759-99088759 on Build GRCh38
rs29001343 Chr.4: 99088755-99088755 on Build GRCh38
rs754124595 Chr.4: 99088735-99088735 on Build GRCh38
rs564461243 Chr.4: 99088884-99088884 on Build GRCh38
rs764880455 Chr.4: 99088695-99088695 on Build GRCh38
rs200326308 Chr.4: 99088672-99088672 on Build GRCh38
rs575931116 Chr.4: 99088871-99088871 on Build GRCh38
rs941780401 Chr.4: 99088872-99088872 on Build GRCh38
rs760252745 Chr.4: 99088861-99088861 on Build GRCh38
rs766357116 Chr.4: 99088657-99088657 on Build GRCh38
rs1017634186 Chr.4: 99088664-99088664 on Build GRCh38
rs756235376 Chr.4: 99088723-99088724 on Build GRCh38
rs989062438 Chr.4: 99089031-99089031 on Build GRCh38
rs748458712 Chr.4: 99088692-99088692 on Build GRCh38
rs771163901 Chr.4: 99088739-99088739 on Build GRCh38
rs529875815 Chr.4: 99088909-99088909 on Build GRCh38
rs868672962 Chr.4: 99088721-99088721 on Build GRCh38
rs1000272646 Chr.4: 99088927-99088927 on Build GRCh38
rs752519844 Chr.4: 99088673-99088673 on Build GRCh38
rs980711964 Chr.4: 99088958-99088958 on Build GRCh38
rs777817524 Chr.4: 99088730-99088731 on Build GRCh38
rs1006773658 Chr.4: 99088808-99088809 on Build GRCh38
rs973151758 Chr.4: 99088984-99088984 on Build GRCh38
rs745903552 Chr.4: 99088743-99088743 on Build GRCh38
rs562329399 Chr.4: 99088801-99088802 on Build GRCh38
rs942546019 Chr.4: 99088906-99088907 on Build GRCh38
rs1016434769 Chr.4: 99088864-99088864 on Build GRCh38
rs970280494 Chr.4: 99088957-99088957 on Build GRCh38
rs35638332 Chr.4: 99088976-99088977 on Build GRCh38
rs374663436 Chr.4: 99088727-99088727 on Build GRCh38
rs11434442 Chr.4: 99088802-99088803 on Build GRCh38
rs1040939099 Chr.4: 99088918-99088918 on Build GRCh38
rs747238832 Chr.4: 99088738-99088738 on Build GRCh38
rs950833310 Chr.4: 99088688-99088688 on Build GRCh38
rs538127712 Chr.4: 99088763-99088763 on Build GRCh38

View Full Product Details