Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99141449-99141950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH4
Entrez Gene ID
127
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Gene Aliases
ADH-2, HEL-S-4
UniGene
Hs.1219
Gene Chromosome Location
Chr.4: 99123657-99144330 on Build GRCh38
Transcripts
NM_000670.4 NM_001306171.1 NM_001306172.1 XM_017007713.1 XM_017007714.1 XM_017007715.1 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_000670.4 NM_001306171.1 NM_001306172.1 XM_017007713.1 XM_017007714.1 XM_017007715.1 NR_037884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5924316 c.236G>A p.G79E Chr.4: 99141567-99141567 on Build GRCh38
7354585 c.158T>A p.V53D Chr.4: 99141645-99141645 on Build GRCh38
3598517 c.198C>T p.I66I Chr.4: 99141605-99141605 on Build GRCh38
1173008 c.133T>A p.S45T Chr.4: 99141670-99141670 on Build GRCh38
3598518 c.162C>T p.I54I Chr.4: 99141641-99141641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748709323 Chr.4: 99141604-99141604 on Build GRCh38
rs758309728 Chr.4: 99141576-99141576 on Build GRCh38
rs745518809 Chr.4: 99141597-99141597 on Build GRCh38
rs747548647 Chr.4: 99141733-99141733 on Build GRCh38
rs768030752 Chr.4: 99141523-99141523 on Build GRCh38
rs759594450 Chr.4: 99141563-99141563 on Build GRCh38
rs1048518636 Chr.4: 99141480-99141480 on Build GRCh38
rs61734619 Chr.4: 99141570-99141570 on Build GRCh38
rs61734620 Chr.4: 99141663-99141663 on Build GRCh38
rs762402595 Chr.4: 99141517-99141517 on Build GRCh38
rs550356850 Chr.4: 99141541-99141541 on Build GRCh38
rs61754857 Chr.4: 99141595-99141595 on Build GRCh38
rs369997257 Chr.4: 99141605-99141605 on Build GRCh38
rs939926780 Chr.4: 99141567-99141567 on Build GRCh38
rs750079516 Chr.4: 99141680-99141680 on Build GRCh38
rs752672320 Chr.4: 99141572-99141572 on Build GRCh38
rs772492403 Chr.4: 99141583-99141584 on Build GRCh38
rs1040894360 Chr.4: 99141569-99141569 on Build GRCh38
rs781034184 Chr.4: 99141679-99141679 on Build GRCh38
rs769492124 Chr.4: 99141599-99141599 on Build GRCh38
rs557732938 Chr.4: 99141640-99141640 on Build GRCh38
rs748956727 Chr.4: 99141566-99141566 on Build GRCh38
rs1004168452 Chr.4: 99141761-99141761 on Build GRCh38
rs997791542 Chr.4: 99141496-99141496 on Build GRCh38
rs572462879 Chr.4: 99141603-99141603 on Build GRCh38
rs765350444 Chr.4: 99141657-99141657 on Build GRCh38
rs1016722732 Chr.4: 99141544-99141544 on Build GRCh38
rs772710044 Chr.4: 99141707-99141707 on Build GRCh38
rs777572180 Chr.4: 99141577-99141577 on Build GRCh38
rs959503113 Chr.4: 99141830-99141830 on Build GRCh38
rs778082667 Chr.4: 99141713-99141713 on Build GRCh38
rs1022694591 Chr.4: 99141855-99141855 on Build GRCh38
rs1026300819 Chr.4: 99141504-99141504 on Build GRCh38
rs1035170668 Chr.4: 99141780-99141780 on Build GRCh38
rs775835319 Chr.4: 99141587-99141596 on Build GRCh38
rs13138294 Chr.4: 99141524-99141524 on Build GRCh38
rs2032349 Chr.4: 99141668-99141668 on Build GRCh38
rs574551057 Chr.4: 99141849-99141849 on Build GRCh38
rs568631109 Chr.4: 99141548-99141548 on Build GRCh38
rs373527102 Chr.4: 99141649-99141649 on Build GRCh38
rs370952138 Chr.4: 99141681-99141681 on Build GRCh38
rs771196498 Chr.4: 99141639-99141639 on Build GRCh38
rs752725480 Chr.4: 99141661-99141661 on Build GRCh38
rs751276332 Chr.4: 99141582-99141582 on Build GRCh38
rs952568126 Chr.4: 99141884-99141884 on Build GRCh38
rs757056129 Chr.4: 99141677-99141677 on Build GRCh38
rs779732857 Chr.4: 99141689-99141689 on Build GRCh38
rs902094291 Chr.4: 99141481-99141481 on Build GRCh38
rs761811968 Chr.4: 99141675-99141675 on Build GRCh38
rs748766354 Chr.4: 99141698-99141698 on Build GRCh38
rs756942645 Chr.4: 99141588-99141588 on Build GRCh38
rs376943095 Chr.4: 99141543-99141543 on Build GRCh38
rs902040245 Chr.4: 99141845-99141845 on Build GRCh38
rs201022290 Chr.4: 99141619-99141619 on Build GRCh38
rs111700010 Chr.4: 99141555-99141555 on Build GRCh38
rs760880924 Chr.4: 99141620-99141620 on Build GRCh38
rs556229640 Chr.4: 99141805-99141805 on Build GRCh38
rs1010930242 Chr.4: 99141646-99141646 on Build GRCh38
rs769297720 Chr.4: 99141510-99141510 on Build GRCh38
rs775053540 Chr.4: 99141515-99141515 on Build GRCh38
rs766470523 Chr.4: 99141550-99141550 on Build GRCh38

View Full Product Details