Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 110475985-110476479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPEP
Entrez Gene ID
2028
Gene Name
glutamyl aminopeptidase
Gene Aliases
APA, CD249, gp160
UniGene
Hs.435765
Gene Chromosome Location
Chr.4: 110476073-110563337 on Build GRCh38
Transcripts
NM_001977.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671206 c.43C>A p.Q15K Chr.4: 110476457-110476457 on Build GRCh38
6237976 c.4A>T p.N2Y Chr.4: 110476418-110476418 on Build GRCh38
7257391 c.1A>C p.M1L Chr.4: 110476415-110476415 on Build GRCh38
5351033 c.36C>A p.Y12* Chr.4: 110476450-110476450 on Build GRCh38
5420278 c.47C>T p.T16M Chr.4: 110476461-110476461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149344736 Chr.4: 110476435-110476435 on Build GRCh38
rs542297020 Chr.4: 110476330-110476330 on Build GRCh38
rs773576095 Chr.4: 110476381-110476381 on Build GRCh38
rs138856188 Chr.4: 110476188-110476188 on Build GRCh38
rs554249336 Chr.4: 110476138-110476138 on Build GRCh38
rs931084843 Chr.4: 110476131-110476131 on Build GRCh38
rs1021857378 Chr.4: 110476245-110476245 on Build GRCh38
rs759416446 Chr.4: 110476389-110476389 on Build GRCh38
rs767450534 Chr.4: 110476406-110476406 on Build GRCh38
rs911419380 Chr.4: 110476172-110476172 on Build GRCh38
rs565382435 Chr.4: 110476125-110476125 on Build GRCh38
rs558506119 Chr.4: 110476189-110476189 on Build GRCh38
rs563793554 Chr.4: 110476332-110476332 on Build GRCh38
rs555937038 Chr.4: 110476148-110476150 on Build GRCh38
rs975529604 Chr.4: 110476239-110476239 on Build GRCh38
rs201390327 Chr.4: 110476438-110476438 on Build GRCh38
rs571137443 Chr.4: 110476046-110476046 on Build GRCh38
rs893894184 Chr.4: 110476024-110476024 on Build GRCh38
rs574999527 Chr.4: 110476091-110476091 on Build GRCh38
rs754934683 Chr.4: 110476298-110476298 on Build GRCh38
rs535905667 Chr.4: 110476137-110476137 on Build GRCh38
rs762680348 Chr.4: 110476378-110476378 on Build GRCh38
rs1036031427 Chr.4: 110476073-110476074 on Build GRCh38
rs569488927 Chr.4: 110476170-110476170 on Build GRCh38
rs758858347 Chr.4: 110476453-110476453 on Build GRCh38
rs774266553 Chr.4: 110476386-110476386 on Build GRCh38
rs113262412 Chr.4: 110476207-110476207 on Build GRCh38
rs553323124 Chr.4: 110476240-110476240 on Build GRCh38
rs972397442 Chr.4: 110476095-110476104 on Build GRCh38
rs753448958 Chr.4: 110476197-110476197 on Build GRCh38
rs750747961 Chr.4: 110476450-110476450 on Build GRCh38
rs754174177 Chr.4: 110476203-110476204 on Build GRCh38
rs765939558 Chr.4: 110476180-110476180 on Build GRCh38
rs181365915 Chr.4: 110476052-110476052 on Build GRCh38
rs775439998 Chr.4: 110476410-110476410 on Build GRCh38
rs201919062 Chr.4: 110476426-110476426 on Build GRCh38
rs3756249 Chr.4: 110476059-110476059 on Build GRCh38
rs147278588 Chr.4: 110476440-110476440 on Build GRCh38
rs542215949 Chr.4: 110476311-110476311 on Build GRCh38
rs1041270520 Chr.4: 110476182-110476183 on Build GRCh38
rs564550794 Chr.4: 110476443-110476443 on Build GRCh38
rs1035569340 Chr.4: 110476347-110476347 on Build GRCh38
rs918308502 Chr.4: 110476112-110476112 on Build GRCh38
rs764559241 Chr.4: 110476278-110476279 on Build GRCh38
rs868419087 Chr.4: 110476357-110476357 on Build GRCh38
rs770890629 Chr.4: 110476381-110476381 on Build GRCh38
rs552566252 Chr.4: 110476016-110476016 on Build GRCh38
rs780541765 Chr.4: 110476454-110476454 on Build GRCh38
rs185786013 Chr.4: 110476204-110476204 on Build GRCh38
rs61735573 Chr.4: 110476429-110476429 on Build GRCh38
rs1011029484 Chr.4: 110476235-110476235 on Build GRCh38
rs574893945 Chr.4: 110476300-110476300 on Build GRCh38
rs1021490315 Chr.4: 110476042-110476042 on Build GRCh38

View Full Product Details