Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 110515250-110515653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPEP
Entrez Gene ID
2028
Gene Name
glutamyl aminopeptidase
Gene Aliases
APA, CD249, gp160
UniGene
Hs.435765
Gene Chromosome Location
Chr.4: 110476073-110563337 on Build GRCh38
Transcripts
NM_001977.3 XM_017007877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6654144 c.1488G>T p.E496D Chr.4: 110515421-110515421 on Build GRCh38
3599141 c.1495C>T p.Q499* Chr.4: 110515428-110515428 on Build GRCh38
3599140 c.1445G>A p.G482E Chr.4: 110515378-110515378 on Build GRCh38
6098677 c.1451C>T p.S484F Chr.4: 110515384-110515384 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770852366 Chr.4: 110515591-110515591 on Build GRCh38
rs550635643 Chr.4: 110515285-110515285 on Build GRCh38
rs925164614 Chr.4: 110515510-110515510 on Build GRCh38
rs937923612 Chr.4: 110515538-110515538 on Build GRCh38
rs560750527 Chr.4: 110515335-110515335 on Build GRCh38
rs368535852 Chr.4: 110515321-110515321 on Build GRCh38
rs773900635 Chr.4: 110515441-110515441 on Build GRCh38
rs770967742 Chr.4: 110515333-110515333 on Build GRCh38
rs745773539 Chr.4: 110515359-110515359 on Build GRCh38
rs768692044 Chr.4: 110515376-110515376 on Build GRCh38
rs183646371 Chr.4: 110515356-110515356 on Build GRCh38
rs267599982 Chr.4: 110515421-110515421 on Build GRCh38
rs777069598 Chr.4: 110515381-110515381 on Build GRCh38
rs762144919 Chr.4: 110515382-110515382 on Build GRCh38
rs955633204 Chr.4: 110515560-110515560 on Build GRCh38
rs990731910 Chr.4: 110515572-110515572 on Build GRCh38
rs759265756 Chr.4: 110515527-110515527 on Build GRCh38
rs752874654 Chr.4: 110515618-110515618 on Build GRCh38
rs753522403 Chr.4: 110515466-110515466 on Build GRCh38
rs755798053 Chr.4: 110515462-110515462 on Build GRCh38
rs7356493 Chr.4: 110515352-110515352 on Build GRCh38
rs887217473 Chr.4: 110515319-110515319 on Build GRCh38
rs777657155 Chr.4: 110515339-110515339 on Build GRCh38
rs765990529 Chr.4: 110515312-110515312 on Build GRCh38
rs556616281 Chr.4: 110515537-110515537 on Build GRCh38
rs975552095 Chr.4: 110515456-110515456 on Build GRCh38
rs954657351 Chr.4: 110515351-110515351 on Build GRCh38
rs549071605 Chr.4: 110515406-110515406 on Build GRCh38
rs567632611 Chr.4: 110515453-110515453 on Build GRCh38
rs749893030 Chr.4: 110515429-110515429 on Build GRCh38
rs763710383 Chr.4: 110515465-110515465 on Build GRCh38
rs143320654 Chr.4: 110515322-110515322 on Build GRCh38
rs747260253 Chr.4: 110515374-110515374 on Build GRCh38
rs752215235 Chr.4: 110515455-110515455 on Build GRCh38
rs562273706 Chr.4: 110515297-110515297 on Build GRCh38
rs3733596 Chr.4: 110515585-110515585 on Build GRCh38
rs763532538 Chr.4: 110515442-110515442 on Build GRCh38
rs757041909 Chr.4: 110515478-110515478 on Build GRCh38
rs201144470 Chr.4: 110515482-110515482 on Build GRCh38
rs979664571 Chr.4: 110515459-110515459 on Build GRCh38
rs1030348055 Chr.4: 110515501-110515501 on Build GRCh38
rs539193039 Chr.4: 110515617-110515617 on Build GRCh38
rs772184787 Chr.4: 110515366-110515366 on Build GRCh38
rs532960993 Chr.4: 110515304-110515304 on Build GRCh38
rs752447949 Chr.4: 110515334-110515334 on Build GRCh38
rs775496097 Chr.4: 110515368-110515368 on Build GRCh38
rs1047397270 Chr.4: 110515621-110515621 on Build GRCh38

View Full Product Details