Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 113455646-113456147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
UniGene
Hs.144114
Gene Chromosome Location
Chr.4: 113451032-113761927 on Build GRCh38
Transcripts
NM_001221.3 NM_001321566.1 NM_001321567.1 NM_001321568.1 NM_001321569.1 NM_001321570.1 NM_001321571.1 NM_001321572.1 NM_001321573.1 NM_001321574.1 NM_001321575.1 NM_001321576.1 NM_001321577.1 NM_001321578.1 NM_001321579.1 NM_001321580.1 NM_001321581.1 NM_001321582.1 NM_001321583.1 NM_001321584.1 NM_001321585.1 NM_001321586.1 NM_001321587.1 NM_001321588.1 NM_001321589.1 NM_001321590.1 NM_001321591.1 NM_172127.2 NM_172128.2 XM_005263253.3 XM_005263254.3 XM_006714330.2 XM_011532289.1 XM_011532290.1 XM_011532291.1 XM_011532292.2 XM_011532295.1 XM_017008672.1 XM_017008673.1 XM_017008674.1 XM_017008675.1 XM_017008676.1 XM_017008677.1 XM_017008678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1050259 c.1496T>C p.I499T Chr.4: 113455759-113455759 on Build GRCh38
5499680 c.1464C>G p.N488K Chr.4: 113455791-113455791 on Build GRCh38
3599302 c.1438C>T p.P480S Chr.4: 113455817-113455817 on Build GRCh38
6845668 c.1488G>A p.W496* Chr.4: 113455767-113455767 on Build GRCh38
1694851 c.1453G>A p.G485R Chr.4: 113455802-113455802 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748351908 Chr.4: 113455872-113455872 on Build GRCh38
rs777310167 Chr.4: 113455765-113455765 on Build GRCh38
rs779484703 Chr.4: 113456031-113456031 on Build GRCh38
rs760458481 Chr.4: 113455805-113455805 on Build GRCh38
rs201528950 Chr.4: 113455862-113455862 on Build GRCh38
rs755794963 Chr.4: 113455749-113455749 on Build GRCh38
rs773613193 Chr.4: 113455797-113455797 on Build GRCh38
rs768862917 Chr.4: 113455779-113455779 on Build GRCh38
rs893486517 Chr.4: 113456096-113456096 on Build GRCh38
rs201504913 Chr.4: 113455785-113455785 on Build GRCh38
rs969953638 Chr.4: 113455795-113455795 on Build GRCh38
rs1041595557 Chr.4: 113456003-113456003 on Build GRCh38
rs532171360 Chr.4: 113455974-113455974 on Build GRCh38
rs568850894 Chr.4: 113455847-113455847 on Build GRCh38
rs911460007 Chr.4: 113455938-113455938 on Build GRCh38
rs897677145 Chr.4: 113455701-113455701 on Build GRCh38
rs764121809 Chr.4: 113455690-113455690 on Build GRCh38
rs761548065 Chr.4: 113455825-113455825 on Build GRCh38
rs35765784 Chr.4: 113455777-113455777 on Build GRCh38
rs763492401 Chr.4: 113455801-113455801 on Build GRCh38
rs909904696 Chr.4: 113455966-113455966 on Build GRCh38
rs757014510 Chr.4: 113455766-113455766 on Build GRCh38
rs919886262 Chr.4: 113455910-113455910 on Build GRCh38
rs775133200 Chr.4: 113455803-113455803 on Build GRCh38
rs771745152 Chr.4: 113455802-113455802 on Build GRCh38
rs1022923278 Chr.4: 113456113-113456113 on Build GRCh38
rs919157749 Chr.4: 113455998-113455998 on Build GRCh38
rs1029784287 Chr.4: 113455880-113455880 on Build GRCh38
rs748840215 Chr.4: 113456076-113456076 on Build GRCh38
rs753479517 Chr.4: 113455819-113455819 on Build GRCh38
rs560042777 Chr.4: 113455987-113455987 on Build GRCh38
rs373513807 Chr.4: 113455706-113455706 on Build GRCh38
rs752444156 Chr.4: 113455735-113455735 on Build GRCh38
rs530429179 Chr.4: 113456007-113456007 on Build GRCh38
rs1031314380 Chr.4: 113455732-113455732 on Build GRCh38
rs2293323 Chr.4: 113456014-113456014 on Build GRCh38
rs77774463 Chr.4: 113455984-113455984 on Build GRCh38
rs773701603 Chr.4: 113455724-113455724 on Build GRCh38
rs148075816 Chr.4: 113455761-113455761 on Build GRCh38
rs751627200 Chr.4: 113455849-113455849 on Build GRCh38
rs763847621 Chr.4: 113455811-113455811 on Build GRCh38
rs750067756 Chr.4: 113455928-113455928 on Build GRCh38
rs765036496 Chr.4: 113455835-113455835 on Build GRCh38
rs1050043498 Chr.4: 113455925-113455925 on Build GRCh38
rs762375183 Chr.4: 113455707-113455707 on Build GRCh38
rs1011130609 Chr.4: 113455843-113455843 on Build GRCh38
rs758327554 Chr.4: 113455839-113455839 on Build GRCh38
rs141794471 Chr.4: 113455776-113455776 on Build GRCh38
rs770154380 Chr.4: 113455791-113455791 on Build GRCh38
rs941316366 Chr.4: 113455979-113455979 on Build GRCh38
rs750338554 Chr.4: 113455769-113455769 on Build GRCh38
rs758998985 Chr.4: 113455725-113455725 on Build GRCh38
rs937760230 Chr.4: 113456071-113456071 on Build GRCh38
rs754994642 Chr.4: 113455853-113455853 on Build GRCh38
rs192100024 Chr.4: 113456072-113456072 on Build GRCh38
rs954260609 Chr.4: 113455924-113455924 on Build GRCh38
rs116016548 Chr.4: 113456108-113456108 on Build GRCh38
rs765648920 Chr.4: 113455713-113455713 on Build GRCh38
rs767002825 Chr.4: 113455731-113455731 on Build GRCh38
rs1037601845 Chr.4: 113455681-113455681 on Build GRCh38
rs781596266 Chr.4: 113455780-113455780 on Build GRCh38
rs1052773896 Chr.4: 113456021-113456021 on Build GRCh38
rs764043866 Chr.4: 113455753-113455753 on Build GRCh38
rs931570120 Chr.4: 113455916-113455916 on Build GRCh38
rs778724654 Chr.4: 113455767-113455767 on Build GRCh38
rs868277844 Chr.4: 113456090-113456090 on Build GRCh38

View Full Product Details