Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 127673895-127674441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTU
Entrez Gene ID
27152
Gene Name
inturned planar cell polarity protein
Gene Aliases
INT, PDZD6, PDZK6
UniGene
Hs.391481
Gene Chromosome Location
Chr.4: 127623271-127716779 on Build GRCh38
Transcripts
NM_015693.3 XM_011531844.2 XM_011531845.2 XM_011531846.1 XM_011531847.1 XM_011531849.2 XM_011531850.2 XM_011531851.2 XM_017008025.1 XM_017008026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1051073 c.1122T>G p.I374M Chr.4: 127674154-127674154 on Build GRCh38
1051074 c.1127T>C p.V376A Chr.4: 127674159-127674159 on Build GRCh38
6862769 c.1104T>A p.L368L Chr.4: 127674136-127674136 on Build GRCh38
6384330 c.1115A>G p.K372R Chr.4: 127674147-127674147 on Build GRCh38
3917153 c.1098C>T p.S366S Chr.4: 127674130-127674130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749756216 Chr.4: 127674237-127674238 on Build GRCh38
rs555724966 Chr.4: 127674233-127674233 on Build GRCh38
rs372029639 Chr.4: 127673921-127673922 on Build GRCh38
rs536587691 Chr.4: 127674197-127674197 on Build GRCh38
rs960425320 Chr.4: 127673921-127673921 on Build GRCh38
rs546379232 Chr.4: 127674009-127674009 on Build GRCh38
rs577782537 Chr.4: 127674396-127674396 on Build GRCh38
rs759862557 Chr.4: 127674093-127674093 on Build GRCh38
rs550864021 Chr.4: 127674040-127674040 on Build GRCh38
rs376158810 Chr.4: 127674307-127674307 on Build GRCh38
rs937196180 Chr.4: 127673988-127673988 on Build GRCh38
rs553201652 Chr.4: 127674342-127674342 on Build GRCh38
rs1033208414 Chr.4: 127674252-127674252 on Build GRCh38
rs369024378 Chr.4: 127674102-127674102 on Build GRCh38
rs536016344 Chr.4: 127674245-127674245 on Build GRCh38
rs143095951 Chr.4: 127674141-127674141 on Build GRCh38
rs950133203 Chr.4: 127673958-127673958 on Build GRCh38
rs771467316 Chr.4: 127674217-127674217 on Build GRCh38
rs560761838 Chr.4: 127673940-127673940 on Build GRCh38
rs1045803205 Chr.4: 127673974-127673974 on Build GRCh38
rs780119950 Chr.4: 127674142-127674142 on Build GRCh38
rs780801040 Chr.4: 127674190-127674190 on Build GRCh38
rs957836438 Chr.4: 127674262-127674262 on Build GRCh38
rs981470511 Chr.4: 127674293-127674293 on Build GRCh38
rs764781959 Chr.4: 127674241-127674241 on Build GRCh38
rs889934577 Chr.4: 127674234-127674234 on Build GRCh38
rs200425250 Chr.4: 127674099-127674100 on Build GRCh38
rs769842168 Chr.4: 127674196-127674196 on Build GRCh38
rs112194826 Chr.4: 127673992-127673992 on Build GRCh38
rs761157376 Chr.4: 127673961-127673961 on Build GRCh38
rs1056253682 Chr.4: 127674272-127674272 on Build GRCh38
rs1020671972 Chr.4: 127674286-127674286 on Build GRCh38
rs911863734 Chr.4: 127674034-127674034 on Build GRCh38
rs896131506 Chr.4: 127674281-127674281 on Build GRCh38
rs542586728 Chr.4: 127673924-127673924 on Build GRCh38
rs759987323 Chr.4: 127674221-127674221 on Build GRCh38
rs913254434 Chr.4: 127673922-127673922 on Build GRCh38
rs535072104 Chr.4: 127674292-127674292 on Build GRCh38
rs762521624 Chr.4: 127674248-127674248 on Build GRCh38
rs930775567 Chr.4: 127674165-127674165 on Build GRCh38
rs1000673047 Chr.4: 127674349-127674349 on Build GRCh38
rs901543401 Chr.4: 127674045-127674045 on Build GRCh38
rs532361153 Chr.4: 127674020-127674020 on Build GRCh38
rs944766155 Chr.4: 127674059-127674064 on Build GRCh38
rs1036436077 Chr.4: 127674030-127674030 on Build GRCh38
rs764805056 Chr.4: 127674121-127674121 on Build GRCh38
rs927397629 Chr.4: 127674321-127674321 on Build GRCh38
rs761590413 Chr.4: 127674237-127674237 on Build GRCh38
rs767933500 Chr.4: 127674223-127674223 on Build GRCh38
rs749346211 Chr.4: 127674200-127674200 on Build GRCh38
rs775646634 Chr.4: 127674229-127674229 on Build GRCh38
rs749894129 Chr.4: 127674124-127674124 on Build GRCh38
rs748417096 Chr.4: 127674195-127674195 on Build GRCh38
rs753267930 Chr.4: 127674324-127674324 on Build GRCh38
rs937456197 Chr.4: 127674339-127674339 on Build GRCh38
rs754855833 Chr.4: 127674185-127674185 on Build GRCh38
rs972720161 Chr.4: 127674061-127674061 on Build GRCh38
rs375562605 Chr.4: 127674306-127674306 on Build GRCh38
rs775199992 Chr.4: 127674047-127674047 on Build GRCh38
rs763686576 Chr.4: 127674328-127674328 on Build GRCh38
rs377048616 Chr.4: 127674263-127674263 on Build GRCh38
rs184601561 Chr.4: 127674154-127674154 on Build GRCh38
rs756798673 Chr.4: 127674120-127674120 on Build GRCh38
rs181472716 Chr.4: 127674013-127674013 on Build GRCh38
rs1049230900 Chr.4: 127674213-127674213 on Build GRCh38
rs746844789 Chr.4: 127674166-127674166 on Build GRCh38
rs1020917005 Chr.4: 127673931-127673931 on Build GRCh38
rs11098934 Chr.4: 127673975-127673975 on Build GRCh38
rs767828979 Chr.4: 127674094-127674094 on Build GRCh38

View Full Product Details