Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145109034-145109574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCE1
Entrez Gene ID
6059
Gene Name
ATP binding cassette subfamily E member 1
Gene Aliases
ABC38, OABP, RLI, RLI1, RNASEL1, RNASELI, RNS4I
UniGene
Hs.12013
Gene Chromosome Location
Chr.4: 145098004-145129524 on Build GRCh38
Transcripts
NM_001040876.1 NM_002940.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3917220 c.298C>T p.P100S Chr.4: 145109142-145109142 on Build GRCh38
6504298 c.332C>T p.T111I Chr.4: 145109176-145109176 on Build GRCh38
1051703 c.404A>T p.D135V Chr.4: 145109248-145109248 on Build GRCh38
390236 c.320G>C p.G107A Chr.4: 145109164-145109164 on Build GRCh38
4658417 c.365T>A p.I122N Chr.4: 145109209-145109209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758091592 Chr.4: 145109122-145109122 on Build GRCh38
rs59601167 Chr.4: 145109480-145109480 on Build GRCh38
rs999770659 Chr.4: 145109488-145109488 on Build GRCh38
rs776684959 Chr.4: 145109295-145109295 on Build GRCh38
rs752453579 Chr.4: 145109089-145109089 on Build GRCh38
rs756855136 Chr.4: 145109108-145109108 on Build GRCh38
rs904319421 Chr.4: 145109339-145109339 on Build GRCh38
rs199627300 Chr.4: 145109308-145109308 on Build GRCh38
rs775165612 Chr.4: 145109273-145109273 on Build GRCh38
rs200833361 Chr.4: 145109150-145109150 on Build GRCh38
rs778550166 Chr.4: 145109118-145109118 on Build GRCh38
rs916886497 Chr.4: 145109505-145109505 on Build GRCh38
rs138109779 Chr.4: 145109223-145109223 on Build GRCh38
rs910009351 Chr.4: 145109465-145109465 on Build GRCh38
rs781545712 Chr.4: 145109425-145109425 on Build GRCh38
rs139255095 Chr.4: 145109174-145109174 on Build GRCh38
rs760555226 Chr.4: 145109096-145109096 on Build GRCh38
rs531628085 Chr.4: 145109349-145109349 on Build GRCh38
rs148851976 Chr.4: 145109498-145109498 on Build GRCh38
rs375908638 Chr.4: 145109119-145109119 on Build GRCh38
rs569240022 Chr.4: 145109329-145109329 on Build GRCh38
rs1056823649 Chr.4: 145109317-145109317 on Build GRCh38
rs926849807 Chr.4: 145109493-145109493 on Build GRCh38
rs750892477 Chr.4: 145109291-145109291 on Build GRCh38
rs34888298 Chr.4: 145109536-145109536 on Build GRCh38
rs769048001 Chr.4: 145109167-145109167 on Build GRCh38
rs966239659 Chr.4: 145109495-145109495 on Build GRCh38
rs763605265 Chr.4: 145109288-145109289 on Build GRCh38
rs372732238 Chr.4: 145109535-145109535 on Build GRCh38
rs145492249 Chr.4: 145109396-145109396 on Build GRCh38
rs567524504 Chr.4: 145109539-145109539 on Build GRCh38
rs753288718 Chr.4: 145109101-145109101 on Build GRCh38
rs951391580 Chr.4: 145109445-145109445 on Build GRCh38
rs895502068 Chr.4: 145109327-145109327 on Build GRCh38
rs10023133 Chr.4: 145109459-145109459 on Build GRCh38
rs57541361 Chr.4: 145109313-145109313 on Build GRCh38
rs540418957 Chr.4: 145109177-145109177 on Build GRCh38
rs764030883 Chr.4: 145109100-145109100 on Build GRCh38
rs767343035 Chr.4: 145109088-145109088 on Build GRCh38
rs1022692180 Chr.4: 145109059-145109059 on Build GRCh38
rs1045721635 Chr.4: 145109262-145109262 on Build GRCh38
rs945699149 Chr.4: 145109531-145109531 on Build GRCh38
rs749546565 Chr.4: 145109246-145109246 on Build GRCh38
rs747484163 Chr.4: 145109334-145109334 on Build GRCh38
rs141006109 Chr.4: 145109282-145109282 on Build GRCh38
rs921038891 Chr.4: 145109195-145109195 on Build GRCh38
rs779225094 Chr.4: 145109380-145109380 on Build GRCh38
rs201224966 Chr.4: 145109162-145109162 on Build GRCh38
rs1010920322 Chr.4: 145109058-145109058 on Build GRCh38
rs201583929 Chr.4: 145109249-145109249 on Build GRCh38
rs983806796 Chr.4: 145109265-145109265 on Build GRCh38
rs769912189 Chr.4: 145109182-145109182 on Build GRCh38
rs113847438 Chr.4: 145109458-145109458 on Build GRCh38
rs780596034 Chr.4: 145109127-145109127 on Build GRCh38
rs922658131 Chr.4: 145109070-145109070 on Build GRCh38
rs953828282 Chr.4: 145109542-145109542 on Build GRCh38
rs763788349 Chr.4: 145109286-145109286 on Build GRCh38
rs773247727 Chr.4: 145109194-145109194 on Build GRCh38
rs980992902 Chr.4: 145109485-145109485 on Build GRCh38
rs760447748 Chr.4: 145109275-145109275 on Build GRCh38
rs745983335 Chr.4: 145109494-145109494 on Build GRCh38
rs982842205 Chr.4: 145109153-145109153 on Build GRCh38
rs535696856 Chr.4: 145109358-145109358 on Build GRCh38
rs998527178 Chr.4: 145109398-145109398 on Build GRCh38
rs371757081 Chr.4: 145109432-145109432 on Build GRCh38

View Full Product Details