Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145109970-145110451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCE1
Entrez Gene ID
6059
Gene Name
ATP binding cassette subfamily E member 1
Gene Aliases
ABC38, OABP, RLI, RLI1, RNASEL1, RNASELI, RNS4I
UniGene
Hs.12013
Gene Chromosome Location
Chr.4: 145098004-145129524 on Build GRCh38
Transcripts
NM_001040876.1 NM_002940.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6362846 c.523C>T p.Q175* Chr.4: 145110220-145110220 on Build GRCh38
7155441 c.569G>A p.R190Q Chr.4: 145110400-145110400 on Build GRCh38
1427347 c.547A>G p.T183A Chr.4: 145110378-145110378 on Build GRCh38
7176087 c.539C>A p.A180E Chr.4: 145110236-145110236 on Build GRCh38
1051704 c.450T>G p.S150S Chr.4: 145110147-145110147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758144917 Chr.4: 145110083-145110083 on Build GRCh38
rs756576892 Chr.4: 145110130-145110130 on Build GRCh38
rs757381481 Chr.4: 145110336-145110336 on Build GRCh38
rs772861686 Chr.4: 145110243-145110243 on Build GRCh38
rs886960802 Chr.4: 145110254-145110254 on Build GRCh38
rs1013692992 Chr.4: 145110358-145110358 on Build GRCh38
rs887980231 Chr.4: 145110003-145110003 on Build GRCh38
rs751275913 Chr.4: 145110263-145110263 on Build GRCh38
rs770516086 Chr.4: 145110401-145110401 on Build GRCh38
rs781061209 Chr.4: 145110366-145110366 on Build GRCh38
rs773340939 Chr.4: 145110396-145110396 on Build GRCh38
rs35207405 Chr.4: 145110244-145110244 on Build GRCh38
rs931963728 Chr.4: 145110389-145110389 on Build GRCh38
rs774698791 Chr.4: 145110177-145110177 on Build GRCh38
rs754335126 Chr.4: 145110282-145110282 on Build GRCh38
rs748731932 Chr.4: 145110124-145110124 on Build GRCh38
rs752957708 Chr.4: 145110269-145110269 on Build GRCh38
rs755437792 Chr.4: 145110085-145110086 on Build GRCh38
rs896594251 Chr.4: 145110306-145110306 on Build GRCh38
rs527933023 Chr.4: 145110016-145110016 on Build GRCh38
rs764877677 Chr.4: 145110060-145110060 on Build GRCh38
rs761631688 Chr.4: 145110055-145110055 on Build GRCh38
rs756630197 Chr.4: 145110272-145110272 on Build GRCh38
rs746140153 Chr.4: 145110342-145110342 on Build GRCh38
rs993881544 Chr.4: 145110109-145110109 on Build GRCh38
rs35152106 Chr.4: 145110275-145110275 on Build GRCh38
rs752978600 Chr.4: 145110085-145110087 on Build GRCh38
rs4148238 Chr.4: 145110085-145110085 on Build GRCh38
rs761511353 Chr.4: 145110027-145110027 on Build GRCh38
rs979691282 Chr.4: 145109993-145109993 on Build GRCh38
rs376877949 Chr.4: 145110187-145110187 on Build GRCh38
rs763421923 Chr.4: 145110058-145110058 on Build GRCh38
rs769859219 Chr.4: 145110386-145110386 on Build GRCh38
rs756238387 Chr.4: 145110085-145110085 on Build GRCh38
rs761871608 Chr.4: 145110211-145110211 on Build GRCh38
rs751911710 Chr.4: 145110084-145110085 on Build GRCh38
rs777856523 Chr.4: 145110131-145110131 on Build GRCh38
rs759258481 Chr.4: 145110426-145110426 on Build GRCh38
rs747893024 Chr.4: 145110378-145110378 on Build GRCh38
rs147976617 Chr.4: 145110299-145110299 on Build GRCh38
rs755429648 Chr.4: 145110106-145110106 on Build GRCh38
rs773696855 Chr.4: 145110419-145110419 on Build GRCh38
rs762962860 Chr.4: 145110397-145110397 on Build GRCh38
rs778028674 Chr.4: 145110413-145110414 on Build GRCh38
rs765216926 Chr.4: 145110234-145110234 on Build GRCh38
rs766071306 Chr.4: 145110100-145110100 on Build GRCh38
rs776257863 Chr.4: 145110195-145110195 on Build GRCh38
rs746061584 Chr.4: 145110183-145110183 on Build GRCh38
rs781390456 Chr.4: 145110122-145110122 on Build GRCh38
rs370360581 Chr.4: 145110051-145110051 on Build GRCh38
rs749312212 Chr.4: 145110132-145110132 on Build GRCh38
rs771142522 Chr.4: 145110169-145110169 on Build GRCh38
rs1010298094 Chr.4: 145110134-145110134 on Build GRCh38
rs1015076045 Chr.4: 145110049-145110050 on Build GRCh38
rs898458248 Chr.4: 145110276-145110276 on Build GRCh38
rs1006813164 Chr.4: 145110004-145110012 on Build GRCh38
rs767958181 Chr.4: 145110268-145110268 on Build GRCh38
rs556371325 Chr.4: 145110350-145110350 on Build GRCh38
rs975186068 Chr.4: 145110078-145110078 on Build GRCh38
rs375439532 Chr.4: 145110267-145110267 on Build GRCh38
rs935452858 Chr.4: 145110323-145110323 on Build GRCh38

View Full Product Details