Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152911150-152911664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP1
Entrez Gene ID
27236
Gene Name
ADP ribosylation factor interacting protein 1
Gene Aliases
HSU52521
UniGene
Hs.416089
Gene Chromosome Location
Chr.4: 152779937-152911912 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531868.2 XM_011531869.2 XM_011531870.2 XM_011531871.2 XM_011531872.1 XM_011532388.1

Gene Symbol
FHDC1
Entrez Gene ID
85462
Gene Name
FH2 domain containing 1
Gene Aliases
-
UniGene
Hs.132629
Gene Chromosome Location
Chr.4: 152911570-152979696 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531868.2 XM_011531869.2 XM_011531870.2 XM_011531871.2 XM_011531872.1 XM_011532388.1

SNP Detail

SNP ID Chromosome Location
rs186384501 Chr.4: 152911609-152911609 on Build GRCh38
rs966255370 Chr.4: 152911469-152911469 on Build GRCh38
rs944941855 Chr.4: 152911239-152911239 on Build GRCh38
rs555485755 Chr.4: 152911185-152911185 on Build GRCh38
rs902401505 Chr.4: 152911358-152911358 on Build GRCh38
rs775099943 Chr.4: 152911176-152911176 on Build GRCh38
rs543541797 Chr.4: 152911608-152911608 on Build GRCh38
rs1014742341 Chr.4: 152911275-152911275 on Build GRCh38
rs946139929 Chr.4: 152911576-152911576 on Build GRCh38
rs1027396871 Chr.4: 152911428-152911428 on Build GRCh38
rs545740586 Chr.4: 152911530-152911530 on Build GRCh38
rs1003424773 Chr.4: 152911373-152911373 on Build GRCh38
rs990853283 Chr.4: 152911360-152911360 on Build GRCh38
rs894958572 Chr.4: 152911416-152911416 on Build GRCh38
rs996809920 Chr.4: 152911547-152911547 on Build GRCh38
rs1053924744 Chr.4: 152911534-152911534 on Build GRCh38
rs568227336 Chr.4: 152911594-152911598 on Build GRCh38
rs763960511 Chr.4: 152911616-152911618 on Build GRCh38
rs894077658 Chr.4: 152911539-152911539 on Build GRCh38
rs189238423 Chr.4: 152911429-152911429 on Build GRCh38
rs953559616 Chr.4: 152911545-152911545 on Build GRCh38
rs1056369032 Chr.4: 152911621-152911621 on Build GRCh38
rs759036707 Chr.4: 152911610-152911614 on Build GRCh38
rs533466861 Chr.4: 152911638-152911638 on Build GRCh38
rs770541449 Chr.4: 152911454-152911454 on Build GRCh38
rs541657931 Chr.4: 152911237-152911237 on Build GRCh38
rs983148973 Chr.4: 152911207-152911207 on Build GRCh38
rs995865244 Chr.4: 152911205-152911205 on Build GRCh38
rs932035988 Chr.4: 152911289-152911289 on Build GRCh38
rs756034804 Chr.4: 152911498-152911498 on Build GRCh38
rs563937940 Chr.4: 152911578-152911578 on Build GRCh38
rs79985890 Chr.4: 152911637-152911637 on Build GRCh38
rs182326281 Chr.4: 152911595-152911595 on Build GRCh38
rs747432595 Chr.4: 152911222-152911222 on Build GRCh38
rs1041742538 Chr.4: 152911344-152911344 on Build GRCh38
rs946044987 Chr.4: 152911327-152911327 on Build GRCh38
rs752482904 Chr.4: 152911420-152911420 on Build GRCh38
rs916549904 Chr.4: 152911409-152911409 on Build GRCh38
rs773362426 Chr.4: 152911255-152911255 on Build GRCh38
rs914661376 Chr.4: 152911568-152911568 on Build GRCh38
rs928656446 Chr.4: 152911584-152911584 on Build GRCh38
rs1014605975 Chr.4: 152911462-152911462 on Build GRCh38
rs759507681 Chr.4: 152911283-152911283 on Build GRCh38
rs990760690 Chr.4: 152911566-152911566 on Build GRCh38
rs976381006 Chr.4: 152911506-152911506 on Build GRCh38
rs951814883 Chr.4: 152911436-152911436 on Build GRCh38
rs1056675983 Chr.4: 152911393-152911393 on Build GRCh38
rs776316763 Chr.4: 152911475-152911475 on Build GRCh38
rs139895520 Chr.4: 152911634-152911634 on Build GRCh38
rs190489005 Chr.4: 152911633-152911633 on Build GRCh38
rs185966767 Chr.4: 152911301-152911301 on Build GRCh38
rs893240038 Chr.4: 152911314-152911314 on Build GRCh38
rs11929781 Chr.4: 152911213-152911213 on Build GRCh38
rs529555163 Chr.4: 152911500-152911500 on Build GRCh38

View Full Product Details