Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168178982-168179512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA10
Entrez Gene ID
11199
Gene Name
annexin A10
Gene Aliases
ANX14
UniGene
Hs.188401
Gene Chromosome Location
Chr.4: 168092525-168187742 on Build GRCh38
Transcripts
NM_007193.4 XM_011531571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6469398 c.661G>A p.D221N Chr.4: 168179249-168179249 on Build GRCh38
1695229 c.637G>A p.E213K Chr.4: 168179225-168179225 on Build GRCh38
6243122 c.698A>G p.Y233C Chr.4: 168179286-168179286 on Build GRCh38
5975049 c.629-1delG p.? Chr.4: 168179216-168179216 on Build GRCh38
6601293 c.653C>A p.S218Y Chr.4: 168179241-168179241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540383303 Chr.4: 168179219-168179219 on Build GRCh38
rs980048862 Chr.4: 168179135-168179135 on Build GRCh38
rs771236167 Chr.4: 168179188-168179188 on Build GRCh38
rs888701647 Chr.4: 168179412-168179412 on Build GRCh38
rs200616323 Chr.4: 168179275-168179275 on Build GRCh38
rs766070039 Chr.4: 168179283-168179283 on Build GRCh38
rs775991175 Chr.4: 168179245-168179245 on Build GRCh38
rs776091128 Chr.4: 168179258-168179258 on Build GRCh38
rs777976843 Chr.4: 168179174-168179174 on Build GRCh38
rs369459720 Chr.4: 168179280-168179280 on Build GRCh38
rs773800184 Chr.4: 168179454-168179454 on Build GRCh38
rs968103746 Chr.4: 168179322-168179322 on Build GRCh38
rs1049967412 Chr.4: 168179395-168179395 on Build GRCh38
rs1039208914 Chr.4: 168179042-168179042 on Build GRCh38
rs769143203 Chr.4: 168179277-168179277 on Build GRCh38
rs1030970863 Chr.4: 168179347-168179347 on Build GRCh38
rs975589526 Chr.4: 168179485-168179485 on Build GRCh38
rs754934546 Chr.4: 168179307-168179307 on Build GRCh38
rs779125919 Chr.4: 168179338-168179338 on Build GRCh38
rs749419189 Chr.4: 168179176-168179176 on Build GRCh38
rs769204340 Chr.4: 168179255-168179255 on Build GRCh38
rs189224821 Chr.4: 168179326-168179326 on Build GRCh38
rs761346677 Chr.4: 168179271-168179271 on Build GRCh38
rs551867650 Chr.4: 168179411-168179411 on Build GRCh38
rs772328727 Chr.4: 168179237-168179237 on Build GRCh38
rs764884583 Chr.4: 168179278-168179278 on Build GRCh38
rs1005188515 Chr.4: 168179413-168179413 on Build GRCh38
rs936646209 Chr.4: 168179154-168179154 on Build GRCh38
rs59482770 Chr.4: 168179417-168179417 on Build GRCh38
rs761156738 Chr.4: 168179254-168179254 on Build GRCh38
rs896888746 Chr.4: 168179426-168179426 on Build GRCh38
rs368739904 Chr.4: 168179323-168179323 on Build GRCh38
rs191641911 Chr.4: 168179113-168179113 on Build GRCh38
rs900236651 Chr.4: 168179083-168179083 on Build GRCh38
rs775975576 Chr.4: 168179282-168179282 on Build GRCh38
rs932899200 Chr.4: 168179369-168179369 on Build GRCh38
rs572956680 Chr.4: 168179216-168179216 on Build GRCh38
rs114097925 Chr.4: 168179289-168179289 on Build GRCh38
rs368827822 Chr.4: 168179339-168179339 on Build GRCh38
rs140456301 Chr.4: 168179310-168179310 on Build GRCh38
rs114021300 Chr.4: 168179462-168179462 on Build GRCh38
rs761241437 Chr.4: 168179459-168179459 on Build GRCh38
rs193095344 Chr.4: 168179469-168179469 on Build GRCh38
rs926614185 Chr.4: 168179153-168179153 on Build GRCh38
rs1048755607 Chr.4: 168179175-168179175 on Build GRCh38
rs969949392 Chr.4: 168179119-168179119 on Build GRCh38
rs772396480 Chr.4: 168179345-168179345 on Build GRCh38
rs779654853 Chr.4: 168179221-168179221 on Build GRCh38

View Full Product Details