Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168357285-168357802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60L
Entrez Gene ID
91351
Gene Name
DEAD-box helicase 60-like
Gene Aliases
-
UniGene
Hs.535011
Gene Chromosome Location
Chr.4: 168356735-168480514 on Build GRCh38
Transcripts
NM_001012967.2 XM_005263341.4 XM_005263342.2 XM_005263343.2 XM_005263345.4 XM_006714408.2 XM_011532402.2 XM_011532403.2 XM_011532404.1 XM_011532405.1 XM_011532406.1 XM_011532409.2 XM_017008829.1 XM_017008830.1 XM_017008831.1

SNP Detail

SNP ID Chromosome Location
rs983836456 Chr.4: 168357597-168357597 on Build GRCh38
rs921852661 Chr.4: 168357771-168357771 on Build GRCh38
rs1038925501 Chr.4: 168357736-168357736 on Build GRCh38
rs961210909 Chr.4: 168357682-168357682 on Build GRCh38
rs555578772 Chr.4: 168357559-168357559 on Build GRCh38
rs1019844216 Chr.4: 168357507-168357507 on Build GRCh38
rs923910521 Chr.4: 168357628-168357628 on Build GRCh38
rs746478651 Chr.4: 168357612-168357612 on Build GRCh38
rs986844283 Chr.4: 168357720-168357720 on Build GRCh38
rs890271784 Chr.4: 168357335-168357335 on Build GRCh38
rs1027622021 Chr.4: 168357462-168357462 on Build GRCh38
rs917929060 Chr.4: 168357347-168357347 on Build GRCh38
rs3172538 Chr.4: 168357742-168357742 on Build GRCh38
rs375635748 Chr.4: 168357513-168357513 on Build GRCh38
rs1005010073 Chr.4: 168357447-168357447 on Build GRCh38
rs577983068 Chr.4: 168357426-168357426 on Build GRCh38
rs936882273 Chr.4: 168357437-168357437 on Build GRCh38
rs540678806 Chr.4: 168357517-168357517 on Build GRCh38
rs912659944 Chr.4: 168357727-168357727 on Build GRCh38
rs775101725 Chr.4: 168357506-168357506 on Build GRCh38
rs553898360 Chr.4: 168357492-168357492 on Build GRCh38
rs556227243 Chr.4: 168357330-168357330 on Build GRCh38
rs1034749066 Chr.4: 168357474-168357474 on Build GRCh38
rs189107444 Chr.4: 168357504-168357504 on Build GRCh38
rs1027757678 Chr.4: 168357399-168357399 on Build GRCh38
rs777615901 Chr.4: 168357404-168357404 on Build GRCh38
rs944526973 Chr.4: 168357345-168357345 on Build GRCh38
rs558304954 Chr.4: 168357381-168357381 on Build GRCh38
rs952430346 Chr.4: 168357550-168357550 on Build GRCh38
rs573838818 Chr.4: 168357729-168357729 on Build GRCh38
rs935366593 Chr.4: 168357663-168357663 on Build GRCh38
rs778447386 Chr.4: 168357365-168357365 on Build GRCh38
rs900850366 Chr.4: 168357509-168357509 on Build GRCh38
rs1010236271 Chr.4: 168357498-168357498 on Build GRCh38
rs979446249 Chr.4: 168357543-168357543 on Build GRCh38
rs764046479 Chr.4: 168357353-168357354 on Build GRCh38
rs115747239 Chr.4: 168357314-168357314 on Build GRCh38
rs917161105 Chr.4: 168357769-168357769 on Build GRCh38
rs1140067 Chr.4: 168357373-168357373 on Build GRCh38
rs538957690 Chr.4: 168357490-168357490 on Build GRCh38
rs370256874 Chr.4: 168357757-168357757 on Build GRCh38
rs994510449 Chr.4: 168357393-168357393 on Build GRCh38
rs758333939 Chr.4: 168357389-168357389 on Build GRCh38
rs1028101929 Chr.4: 168357537-168357537 on Build GRCh38

View Full Product Details