Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31534047-31534542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf22
Entrez Gene ID
55322
Gene Name
chromosome 5 open reading frame 22
Gene Aliases
-
UniGene
Hs.519246
Gene Chromosome Location
Chr.5: 31532086-31555058 on Build GRCh38
Transcripts
NM_018356.2 NR_134298.1 XM_006714479.2 XM_006714480.3 XM_011514062.2 XM_017009607.1 XM_017009608.1 XM_017009609.1 XM_017009610.1 XM_017009611.1 XM_017009612.1 XM_017009613.1 XM_017009614.1 XM_017009615.1 XM_017009616.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1436996 c.137T>C p.V46A Chr.5: 31534327-31534327 on Build GRCh38
5651438 c.186T>C p.N62N Chr.5: 31534376-31534376 on Build GRCh38
360009 c.201C>T p.T67T Chr.5: 31534391-31534391 on Build GRCh38
738147 c.127G>A p.A43T Chr.5: 31534317-31534317 on Build GRCh38
738148 c.103G>T p.A35S Chr.5: 31534293-31534293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868631976 Chr.5: 31534070-31534070 on Build GRCh38
rs565212422 Chr.5: 31534090-31534090 on Build GRCh38
rs754710898 Chr.5: 31534142-31534142 on Build GRCh38
rs947769722 Chr.5: 31534502-31534502 on Build GRCh38
rs770875845 Chr.5: 31534433-31534433 on Build GRCh38
rs968287985 Chr.5: 31534486-31534486 on Build GRCh38
rs1035582092 Chr.5: 31534284-31534284 on Build GRCh38
rs751873975 Chr.5: 31534262-31534262 on Build GRCh38
rs201547431 Chr.5: 31534290-31534290 on Build GRCh38
rs764095889 Chr.5: 31534392-31534392 on Build GRCh38
rs749237993 Chr.5: 31534366-31534366 on Build GRCh38
rs7737168 Chr.5: 31534183-31534183 on Build GRCh38
rs768335741 Chr.5: 31534325-31534325 on Build GRCh38
rs999815331 Chr.5: 31534155-31534155 on Build GRCh38
rs954947790 Chr.5: 31534371-31534371 on Build GRCh38
rs113953500 Chr.5: 31534216-31534216 on Build GRCh38
rs760288902 Chr.5: 31534221-31534221 on Build GRCh38
rs779888296 Chr.5: 31534307-31534307 on Build GRCh38
rs973065181 Chr.5: 31534472-31534472 on Build GRCh38
rs73747444 Chr.5: 31534180-31534180 on Build GRCh38
rs766831369 Chr.5: 31534256-31534256 on Build GRCh38
rs75374942 Chr.5: 31534249-31534249 on Build GRCh38
rs150791229 Chr.5: 31534467-31534467 on Build GRCh38
rs768812196 Chr.5: 31534405-31534405 on Build GRCh38
rs540272241 Chr.5: 31534149-31534149 on Build GRCh38
rs752975087 Chr.5: 31534379-31534379 on Build GRCh38
rs763449182 Chr.5: 31534328-31534328 on Build GRCh38
rs934102344 Chr.5: 31534178-31534178 on Build GRCh38
rs906464820 Chr.5: 31534154-31534154 on Build GRCh38
rs921323297 Chr.5: 31534175-31534175 on Build GRCh38
rs757172451 Chr.5: 31534409-31534409 on Build GRCh38
rs771353594 Chr.5: 31534337-31534337 on Build GRCh38
rs565243950 Chr.5: 31534395-31534395 on Build GRCh38
rs904912568 Chr.5: 31534270-31534270 on Build GRCh38
rs551664112 Chr.5: 31534434-31534434 on Build GRCh38
rs750197579 Chr.5: 31534413-31534413 on Build GRCh38
rs374568303 Chr.5: 31534229-31534229 on Build GRCh38
rs745675913 Chr.5: 31534295-31534295 on Build GRCh38
rs528009766 Chr.5: 31534421-31534421 on Build GRCh38
rs767704970 Chr.5: 31534368-31534368 on Build GRCh38
rs916351185 Chr.5: 31534473-31534473 on Build GRCh38
rs1030465177 Chr.5: 31534317-31534317 on Build GRCh38
rs200237808 Chr.5: 31534269-31534269 on Build GRCh38
rs747149196 Chr.5: 31534330-31534331 on Build GRCh38
rs145934075 Chr.5: 31534182-31534182 on Build GRCh38
rs1022470969 Chr.5: 31534479-31534479 on Build GRCh38
rs199731432 Chr.5: 31534297-31534297 on Build GRCh38
rs560316811 Chr.5: 31534160-31534160 on Build GRCh38
rs780693302 Chr.5: 31534465-31534465 on Build GRCh38
rs141326993 Chr.5: 31534263-31534263 on Build GRCh38
rs746491521 Chr.5: 31534324-31534324 on Build GRCh38
rs899088127 Chr.5: 31534204-31534204 on Build GRCh38
rs1003323570 Chr.5: 31534272-31534272 on Build GRCh38
rs774052435 Chr.5: 31534492-31534492 on Build GRCh38
rs757148459 Chr.5: 31534291-31534291 on Build GRCh38
rs777676452 Chr.5: 31534287-31534287 on Build GRCh38
rs760741460 Chr.5: 31534391-31534391 on Build GRCh38
rs916311629 Chr.5: 31534136-31534136 on Build GRCh38
rs941694484 Chr.5: 31534119-31534119 on Build GRCh38
rs531970192 Chr.5: 31534414-31534414 on Build GRCh38
rs79734107 Chr.5: 31534303-31534303 on Build GRCh38
rs774727680 Chr.5: 31534361-31534361 on Build GRCh38
rs775831828 Chr.5: 31534296-31534297 on Build GRCh38
rs964760453 Chr.5: 31534234-31534234 on Build GRCh38
rs1009768438 Chr.5: 31534476-31534476 on Build GRCh38
rs540103558 Chr.5: 31534124-31534124 on Build GRCh38
rs756237388 Chr.5: 31534275-31534275 on Build GRCh38
rs527576352 Chr.5: 31534145-31534145 on Build GRCh38
rs770389873 Chr.5: 31534226-31534226 on Build GRCh38

View Full Product Details