Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33546037-33546561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS12
Entrez Gene ID
81792
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 12
Gene Aliases
PRO4389
UniGene
Hs.12680
Gene Chromosome Location
Chr.5: 33523535-33892180 on Build GRCh38
Transcripts
NM_001324512.1 NM_030955.3 XM_017009905.1 XM_017009906.1 XM_017009907.1 XM_017009908.1 XM_017009909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6103916 c.4443C>A p.D1481E Chr.5: 33546062-33546062 on Build GRCh38
7354254 c.4403A>G p.N1468S Chr.5: 33546102-33546102 on Build GRCh38
6103915 c.4370G>A p.W1457* Chr.5: 33546135-33546135 on Build GRCh38
4659449 c.4437C>T p.N1479N Chr.5: 33546068-33546068 on Build GRCh38
6103914 c.4342G>T p.V1448L Chr.5: 33546163-33546163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61753559 Chr.5: 33546102-33546102 on Build GRCh38
rs76061577 Chr.5: 33546132-33546132 on Build GRCh38
rs997231048 Chr.5: 33546406-33546406 on Build GRCh38
rs776770546 Chr.5: 33546135-33546135 on Build GRCh38
rs900294957 Chr.5: 33546393-33546393 on Build GRCh38
rs776472308 Chr.5: 33546131-33546131 on Build GRCh38
rs10058544 Chr.5: 33546291-33546291 on Build GRCh38
rs761496496 Chr.5: 33546133-33546133 on Build GRCh38
rs753095566 Chr.5: 33546164-33546164 on Build GRCh38
rs769419295 Chr.5: 33546242-33546242 on Build GRCh38
rs768620994 Chr.5: 33546225-33546225 on Build GRCh38
rs775428386 Chr.5: 33546123-33546123 on Build GRCh38
rs778869978 Chr.5: 33546201-33546201 on Build GRCh38
rs749500121 Chr.5: 33546098-33546098 on Build GRCh38
rs1006106429 Chr.5: 33546396-33546396 on Build GRCh38
rs759976806 Chr.5: 33546155-33546155 on Build GRCh38
rs894031552 Chr.5: 33546383-33546383 on Build GRCh38
rs527559880 Chr.5: 33546520-33546520 on Build GRCh38
rs984639000 Chr.5: 33546150-33546150 on Build GRCh38
rs72737394 Chr.5: 33546386-33546386 on Build GRCh38
rs772166112 Chr.5: 33546211-33546211 on Build GRCh38
rs745892833 Chr.5: 33546206-33546206 on Build GRCh38
rs781598316 Chr.5: 33546072-33546072 on Build GRCh38
rs766830653 Chr.5: 33546292-33546292 on Build GRCh38
rs1018851261 Chr.5: 33546388-33546388 on Build GRCh38
rs534271617 Chr.5: 33546223-33546223 on Build GRCh38
rs760505608 Chr.5: 33546124-33546124 on Build GRCh38
rs370263637 Chr.5: 33546273-33546273 on Build GRCh38
rs763435015 Chr.5: 33546250-33546250 on Build GRCh38
rs746912265 Chr.5: 33546224-33546224 on Build GRCh38
rs542627835 Chr.5: 33546274-33546274 on Build GRCh38
rs377117812 Chr.5: 33546267-33546267 on Build GRCh38
rs377734259 Chr.5: 33546530-33546530 on Build GRCh38
rs3813472 Chr.5: 33546281-33546281 on Build GRCh38
rs893254704 Chr.5: 33546309-33546309 on Build GRCh38
rs553708888 Chr.5: 33546226-33546226 on Build GRCh38
rs749938074 Chr.5: 33546154-33546154 on Build GRCh38
rs29967 Chr.5: 33546327-33546327 on Build GRCh38
rs554453996 Chr.5: 33546159-33546159 on Build GRCh38
rs975779729 Chr.5: 33546097-33546097 on Build GRCh38
rs538002967 Chr.5: 33546114-33546114 on Build GRCh38
rs146262747 Chr.5: 33546077-33546077 on Build GRCh38
rs796258311 Chr.5: 33546376-33546376 on Build GRCh38
rs941848934 Chr.5: 33546251-33546251 on Build GRCh38
rs909015313 Chr.5: 33546167-33546167 on Build GRCh38
rs756337774 Chr.5: 33546093-33546093 on Build GRCh38
rs990658449 Chr.5: 33546460-33546460 on Build GRCh38
rs777936614 Chr.5: 33546096-33546096 on Build GRCh38
rs199556581 Chr.5: 33546173-33546173 on Build GRCh38
rs757485140 Chr.5: 33546200-33546200 on Build GRCh38
rs755506534 Chr.5: 33546068-33546068 on Build GRCh38
rs772075879 Chr.5: 33546120-33546120 on Build GRCh38
rs377536950 Chr.5: 33546194-33546194 on Build GRCh38
rs768445743 Chr.5: 33546126-33546126 on Build GRCh38
rs747820756 Chr.5: 33546229-33546229 on Build GRCh38
rs1030012078 Chr.5: 33546422-33546422 on Build GRCh38
rs74463306 Chr.5: 33546376-33546376 on Build GRCh38
rs374907311 Chr.5: 33546067-33546067 on Build GRCh38
rs950336441 Chr.5: 33546442-33546442 on Build GRCh38
rs935592068 Chr.5: 33546377-33546377 on Build GRCh38
rs950616858 Chr.5: 33546158-33546158 on Build GRCh38
rs997539843 Chr.5: 33546386-33546387 on Build GRCh38
rs949103405 Chr.5: 33546247-33546247 on Build GRCh38
rs141526328 Chr.5: 33546323-33546323 on Build GRCh38
rs950593005 Chr.5: 33546118-33546118 on Build GRCh38
rs566985924 Chr.5: 33546335-33546335 on Build GRCh38
rs770952996 Chr.5: 33546100-33546100 on Build GRCh38
rs959503648 Chr.5: 33546455-33546455 on Build GRCh38
rs764722747 Chr.5: 33546134-33546134 on Build GRCh38
rs754072330 Chr.5: 33546183-33546183 on Build GRCh38
rs557941876 Chr.5: 33546165-33546165 on Build GRCh38
rs1054011403 Chr.5: 33546378-33546378 on Build GRCh38

View Full Product Details