Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33588412-33588910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS12
Entrez Gene ID
81792
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 12
Gene Aliases
PRO4389
UniGene
Hs.12680
Gene Chromosome Location
Chr.5: 33523535-33892180 on Build GRCh38
Transcripts
NM_001324512.1 NM_030955.3 XM_017009905.1 XM_017009906.1 XM_017009907.1 XM_017009908.1 XM_017009909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6546120 c.2779C>A p.Q927K Chr.5: 33588685-33588685 on Build GRCh38
3947439 c.2764C>A p.P922T Chr.5: 33588700-33588700 on Build GRCh38
6546119 c.2805C>A p.L935L Chr.5: 33588659-33588659 on Build GRCh38
4402742 c.2684C>T p.S895L Chr.5: 33588780-33588780 on Build GRCh38
1671564 c.2717G>A p.R906Q Chr.5: 33588747-33588747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754686750 Chr.5: 33588800-33588800 on Build GRCh38
rs1020638953 Chr.5: 33588863-33588863 on Build GRCh38
rs779806882 Chr.5: 33588586-33588586 on Build GRCh38
rs138076156 Chr.5: 33588787-33588787 on Build GRCh38
rs768280943 Chr.5: 33588591-33588591 on Build GRCh38
rs746629845 Chr.5: 33588590-33588590 on Build GRCh38
rs930826780 Chr.5: 33588790-33588790 on Build GRCh38
rs566996629 Chr.5: 33588887-33588887 on Build GRCh38
rs762037150 Chr.5: 33588767-33588767 on Build GRCh38
rs1001216905 Chr.5: 33588500-33588500 on Build GRCh38
rs775660919 Chr.5: 33588747-33588747 on Build GRCh38
rs774886749 Chr.5: 33588731-33588731 on Build GRCh38
rs766184072 Chr.5: 33588859-33588859 on Build GRCh38
rs750368564 Chr.5: 33588682-33588682 on Build GRCh38
rs983994396 Chr.5: 33588643-33588643 on Build GRCh38
rs754083071 Chr.5: 33588761-33588761 on Build GRCh38
rs757206455 Chr.5: 33588570-33588570 on Build GRCh38
rs145444871 Chr.5: 33588699-33588699 on Build GRCh38
rs933531471 Chr.5: 33588619-33588619 on Build GRCh38
rs570498983 Chr.5: 33588852-33588852 on Build GRCh38
rs763383295 Chr.5: 33588781-33588782 on Build GRCh38
rs549198267 Chr.5: 33588587-33588587 on Build GRCh38
rs746389503 Chr.5: 33588840-33588840 on Build GRCh38
rs532990917 Chr.5: 33588501-33588501 on Build GRCh38
rs957999733 Chr.5: 33588498-33588498 on Build GRCh38
rs964941353 Chr.5: 33588517-33588517 on Build GRCh38
rs777699589 Chr.5: 33588549-33588549 on Build GRCh38
rs772553184 Chr.5: 33588743-33588743 on Build GRCh38
rs759945938 Chr.5: 33588736-33588736 on Build GRCh38
rs549118996 Chr.5: 33588692-33588692 on Build GRCh38
rs759814598 Chr.5: 33588850-33588851 on Build GRCh38
rs780859019 Chr.5: 33588807-33588807 on Build GRCh38
rs771297596 Chr.5: 33588814-33588814 on Build GRCh38
rs141518800 Chr.5: 33588451-33588451 on Build GRCh38
rs926051279 Chr.5: 33588637-33588637 on Build GRCh38
rs149530928 Chr.5: 33588809-33588809 on Build GRCh38
rs895469264 Chr.5: 33588832-33588832 on Build GRCh38
rs978594249 Chr.5: 33588630-33588630 on Build GRCh38
rs762967809 Chr.5: 33588858-33588858 on Build GRCh38
rs775956446 Chr.5: 33588842-33588842 on Build GRCh38
rs768103937 Chr.5: 33588570-33588570 on Build GRCh38
rs181370821 Chr.5: 33588876-33588876 on Build GRCh38
rs755678876 Chr.5: 33588823-33588823 on Build GRCh38
rs760801296 Chr.5: 33588663-33588663 on Build GRCh38
rs763462727 Chr.5: 33588598-33588598 on Build GRCh38
rs768132388 Chr.5: 33588712-33588712 on Build GRCh38
rs765414663 Chr.5: 33588855-33588855 on Build GRCh38
rs188539517 Chr.5: 33588495-33588495 on Build GRCh38
rs769263371 Chr.5: 33588724-33588724 on Build GRCh38
rs745623434 Chr.5: 33588583-33588583 on Build GRCh38
rs932033088 Chr.5: 33588713-33588713 on Build GRCh38
rs765398659 Chr.5: 33588770-33588770 on Build GRCh38
rs967595882 Chr.5: 33588625-33588625 on Build GRCh38
rs753019193 Chr.5: 33588662-33588662 on Build GRCh38
rs1027134634 Chr.5: 33588489-33588489 on Build GRCh38
rs774729144 Chr.5: 33588614-33588614 on Build GRCh38
rs757292276 Chr.5: 33588681-33588681 on Build GRCh38
rs1017859454 Chr.5: 33588511-33588511 on Build GRCh38
rs907969088 Chr.5: 33588742-33588742 on Build GRCh38
rs780699623 Chr.5: 33588719-33588719 on Build GRCh38
rs867888028 Chr.5: 33588621-33588621 on Build GRCh38
rs777259981 Chr.5: 33588830-33588830 on Build GRCh38
rs2877316 Chr.5: 33588537-33588537 on Build GRCh38
rs993961878 Chr.5: 33588473-33588473 on Build GRCh38
rs1001280669 Chr.5: 33588824-33588824 on Build GRCh38
rs751260063 Chr.5: 33588797-33588797 on Build GRCh38
rs200923737 Chr.5: 33588611-33588611 on Build GRCh38
rs773105700 Chr.5: 33588856-33588856 on Build GRCh38
rs989126243 Chr.5: 33588504-33588504 on Build GRCh38
rs775934790 Chr.5: 33588596-33588596 on Build GRCh38
rs753920986 Chr.5: 33588680-33588680 on Build GRCh38
rs759930119 Chr.5: 33588627-33588627 on Build GRCh38
rs764250440 Chr.5: 33588670-33588670 on Build GRCh38
rs753833069 Chr.5: 33588562-33588562 on Build GRCh38
rs773736825 Chr.5: 33588779-33588780 on Build GRCh38
rs764183670 Chr.5: 33588755-33588755 on Build GRCh38
rs568958735 Chr.5: 33588753-33588753 on Build GRCh38
rs767747447 Chr.5: 33588633-33588633 on Build GRCh38
rs771733709 Chr.5: 33588584-33588584 on Build GRCh38
rs747373953 Chr.5: 33588850-33588850 on Build GRCh38
rs1037009705 Chr.5: 33588737-33588737 on Build GRCh38
rs200229437 Chr.5: 33588678-33588678 on Build GRCh38
rs1046371196 Chr.5: 33588886-33588886 on Build GRCh38
rs777054940 Chr.5: 33588853-33588853 on Build GRCh38
rs776564565 Chr.5: 33588867-33588867 on Build GRCh38
rs201796524 Chr.5: 33588776-33588776 on Build GRCh38
rs56237226 Chr.5: 33588579-33588579 on Build GRCh38
rs539479028 Chr.5: 33588857-33588857 on Build GRCh38
rs774836176 Chr.5: 33588835-33588837 on Build GRCh38
rs1033984263 Chr.5: 33588514-33588514 on Build GRCh38

View Full Product Details