Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 34004441-34004938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMACR
Entrez Gene ID
23600
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
AMACRD, CBAS4, RACE, RM
UniGene
Hs.508343
Gene Chromosome Location
Chr.5: 33986986-34008115 on Build GRCh38
Transcripts
NM_001167595.1 NM_014324.5 NM_203382.2 NR_037951.1

Gene Symbol
C1QTNF3-AMACR
Entrez Gene ID
100534612
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 33986986-34124528 on Build GRCh38
Transcripts
NM_001167595.1 NM_014324.5 NM_203382.2 NR_037951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5941249 c.524G>A p.G175D Chr.5: 34004602-34004602 on Build GRCh38
3015305 c.518G>A p.R173H Chr.5: 34004608-34004608 on Build GRCh38
5978967 c.529G>A p.G177S Chr.5: 34004597-34004597 on Build GRCh38
4571506 c.476T>A p.M159K Chr.5: 34004650-34004650 on Build GRCh38
7251092 c.434C>A p.A145D Chr.5: 34004692-34004692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901447342 Chr.5: 34004854-34004854 on Build GRCh38
rs186210578 Chr.5: 34004833-34004833 on Build GRCh38
rs368427062 Chr.5: 34004697-34004697 on Build GRCh38
rs773780945 Chr.5: 34004615-34004615 on Build GRCh38
rs754192887 Chr.5: 34004557-34004557 on Build GRCh38
rs368314254 Chr.5: 34004558-34004558 on Build GRCh38
rs370924790 Chr.5: 34004531-34004531 on Build GRCh38
rs944599023 Chr.5: 34004485-34004485 on Build GRCh38
rs752030727 Chr.5: 34004862-34004862 on Build GRCh38
rs182629265 Chr.5: 34004476-34004476 on Build GRCh38
rs1000843834 Chr.5: 34004713-34004713 on Build GRCh38
rs965975823 Chr.5: 34004858-34004858 on Build GRCh38
rs768526839 Chr.5: 34004607-34004607 on Build GRCh38
rs7733150 Chr.5: 34004786-34004786 on Build GRCh38
rs147265006 Chr.5: 34004650-34004650 on Build GRCh38
rs773084615 Chr.5: 34004612-34004612 on Build GRCh38
rs527996590 Chr.5: 34004671-34004671 on Build GRCh38
rs755529761 Chr.5: 34004873-34004873 on Build GRCh38
rs748131178 Chr.5: 34004703-34004703 on Build GRCh38
rs35206983 Chr.5: 34004511-34004511 on Build GRCh38
rs766895023 Chr.5: 34004630-34004630 on Build GRCh38
rs34401823 Chr.5: 34004489-34004491 on Build GRCh38
rs762377420 Chr.5: 34004751-34004751 on Build GRCh38
rs746099405 Chr.5: 34004567-34004567 on Build GRCh38
rs756124583 Chr.5: 34004750-34004750 on Build GRCh38
rs553862435 Chr.5: 34004549-34004549 on Build GRCh38
rs757917164 Chr.5: 34004689-34004689 on Build GRCh38
rs756635815 Chr.5: 34004540-34004540 on Build GRCh38
rs10941112 Chr.5: 34004602-34004602 on Build GRCh38
rs758553398 Chr.5: 34004588-34004588 on Build GRCh38
rs1028843151 Chr.5: 34004729-34004729 on Build GRCh38
rs1029958967 Chr.5: 34004867-34004867 on Build GRCh38
rs563361766 Chr.5: 34004470-34004470 on Build GRCh38
rs762246455 Chr.5: 34004651-34004651 on Build GRCh38
rs762760236 Chr.5: 34004613-34004613 on Build GRCh38
rs749834940 Chr.5: 34004633-34004633 on Build GRCh38
rs140523570 Chr.5: 34004688-34004688 on Build GRCh38
rs201818706 Chr.5: 34004608-34004608 on Build GRCh38
rs372260537 Chr.5: 34004759-34004759 on Build GRCh38
rs769734678 Chr.5: 34004611-34004611 on Build GRCh38
rs774080915 Chr.5: 34004734-34004734 on Build GRCh38
rs1036070478 Chr.5: 34004486-34004486 on Build GRCh38
rs369179060 Chr.5: 34004497-34004499 on Build GRCh38
rs911725988 Chr.5: 34004846-34004846 on Build GRCh38
rs759053723 Chr.5: 34004620-34004620 on Build GRCh38
rs1016448674 Chr.5: 34004880-34004880 on Build GRCh38
rs747997206 Chr.5: 34004609-34004609 on Build GRCh38
rs769787979 Chr.5: 34004715-34004715 on Build GRCh38
rs745999216 Chr.5: 34004756-34004757 on Build GRCh38
rs113840552 Chr.5: 34004692-34004692 on Build GRCh38
rs779077834 Chr.5: 34004705-34004707 on Build GRCh38
rs770694101 Chr.5: 34004614-34004614 on Build GRCh38
rs138754975 Chr.5: 34004720-34004720 on Build GRCh38
rs758681312 Chr.5: 34004678-34004678 on Build GRCh38
rs201830086 Chr.5: 34004563-34004563 on Build GRCh38
rs373915688 Chr.5: 34004544-34004544 on Build GRCh38
rs557957370 Chr.5: 34004468-34004468 on Build GRCh38
rs200599829 Chr.5: 34004665-34004665 on Build GRCh38
rs911821075 Chr.5: 34004464-34004464 on Build GRCh38
rs573155402 Chr.5: 34004601-34004601 on Build GRCh38
rs777768049 Chr.5: 34004716-34004716 on Build GRCh38
rs144785688 Chr.5: 34004667-34004667 on Build GRCh38
rs998372301 Chr.5: 34004860-34004860 on Build GRCh38
rs750839179 Chr.5: 34004767-34004767 on Build GRCh38
rs770745666 Chr.5: 34004733-34004733 on Build GRCh38

View Full Product Details