Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35012861-35013408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGXT2
Entrez Gene ID
64902
Gene Name
alanine--glyoxylate aminotransferase 2
Gene Aliases
AGT2, DAIBAT
UniGene
Hs.34494
Gene Chromosome Location
Chr.5: 34998101-35048188 on Build GRCh38
Transcripts
NM_001306173.1 NM_031900.3 XM_005248337.2 XM_005248338.2 XM_017009748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5403346 c.1154_1155CC>TT p.P385L Chr.5: 35012987-35012988 on Build GRCh38
5683801 c.1129C>T p.H377Y Chr.5: 35013013-35013013 on Build GRCh38
5486955 c.1188+8C>T p.? Chr.5: 35012946-35012946 on Build GRCh38
6834914 c.1166C>T p.A389V Chr.5: 35012976-35012976 on Build GRCh38
6355052 c.1115C>T p.A372V Chr.5: 35013027-35013027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910142628 Chr.5: 35013271-35013271 on Build GRCh38
rs1020810059 Chr.5: 35013311-35013311 on Build GRCh38
rs1009866658 Chr.5: 35013106-35013106 on Build GRCh38
rs376808491 Chr.5: 35013060-35013060 on Build GRCh38
rs1019796987 Chr.5: 35013363-35013363 on Build GRCh38
rs1020633062 Chr.5: 35013145-35013145 on Build GRCh38
rs200794102 Chr.5: 35012906-35012907 on Build GRCh38
rs565446562 Chr.5: 35013205-35013205 on Build GRCh38
rs574431191 Chr.5: 35013011-35013011 on Build GRCh38
rs371039703 Chr.5: 35013126-35013126 on Build GRCh38
rs1028069979 Chr.5: 35013331-35013331 on Build GRCh38
rs764599234 Chr.5: 35013166-35013166 on Build GRCh38
rs181398397 Chr.5: 35013097-35013097 on Build GRCh38
rs895505390 Chr.5: 35013340-35013340 on Build GRCh38
rs534317229 Chr.5: 35012973-35012973 on Build GRCh38
rs530201816 Chr.5: 35013309-35013309 on Build GRCh38
rs765628996 Chr.5: 35013238-35013238 on Build GRCh38
rs765600504 Chr.5: 35012999-35012999 on Build GRCh38
rs1044480561 Chr.5: 35012987-35012987 on Build GRCh38
rs142665211 Chr.5: 35013012-35013012 on Build GRCh38
rs528148122 Chr.5: 35013079-35013079 on Build GRCh38
rs770707002 Chr.5: 35013108-35013108 on Build GRCh38
rs940297488 Chr.5: 35013243-35013243 on Build GRCh38
rs554599418 Chr.5: 35013004-35013004 on Build GRCh38
rs780144535 Chr.5: 35013033-35013033 on Build GRCh38
rs1020024232 Chr.5: 35013017-35013017 on Build GRCh38
rs545248647 Chr.5: 35013148-35013148 on Build GRCh38
rs536786734 Chr.5: 35013039-35013039 on Build GRCh38
rs762091646 Chr.5: 35012984-35012984 on Build GRCh38
rs377751540 Chr.5: 35013131-35013131 on Build GRCh38
rs750663551 Chr.5: 35013020-35013020 on Build GRCh38
rs576253909 Chr.5: 35013109-35013109 on Build GRCh38
rs931287191 Chr.5: 35013265-35013265 on Build GRCh38
rs371584296 Chr.5: 35012941-35012941 on Build GRCh38
rs751631360 Chr.5: 35013070-35013070 on Build GRCh38
rs781005296 Chr.5: 35013092-35013092 on Build GRCh38
rs755000445 Chr.5: 35013088-35013088 on Build GRCh38
rs563444197 Chr.5: 35013327-35013327 on Build GRCh38
rs758667765 Chr.5: 35013266-35013266 on Build GRCh38
rs1035386353 Chr.5: 35013044-35013044 on Build GRCh38
rs548255792 Chr.5: 35013204-35013204 on Build GRCh38
rs754400700 Chr.5: 35013211-35013211 on Build GRCh38
rs1000482238 Chr.5: 35013316-35013316 on Build GRCh38
rs751660629 Chr.5: 35013312-35013312 on Build GRCh38
rs568054343 Chr.5: 35013308-35013308 on Build GRCh38
rs750731899 Chr.5: 35013261-35013261 on Build GRCh38
rs773968211 Chr.5: 35012942-35012942 on Build GRCh38
rs775099568 Chr.5: 35013023-35013023 on Build GRCh38
rs761331893 Chr.5: 35012947-35012947 on Build GRCh38
rs905943346 Chr.5: 35013267-35013267 on Build GRCh38
rs541060681 Chr.5: 35013280-35013280 on Build GRCh38
rs953245566 Chr.5: 35013202-35013202 on Build GRCh38
rs973370071 Chr.5: 35013253-35013253 on Build GRCh38
rs566696270 Chr.5: 35013107-35013107 on Build GRCh38
rs961823637 Chr.5: 35013366-35013366 on Build GRCh38
rs938724638 Chr.5: 35013299-35013299 on Build GRCh38
rs550311947 Chr.5: 35013314-35013314 on Build GRCh38
rs150278377 Chr.5: 35013220-35013220 on Build GRCh38
rs368015775 Chr.5: 35012925-35012925 on Build GRCh38
rs759053806 Chr.5: 35012970-35012970 on Build GRCh38
rs965400889 Chr.5: 35013229-35013229 on Build GRCh38
rs114286107 Chr.5: 35012953-35012953 on Build GRCh38
rs754952540 Chr.5: 35013362-35013362 on Build GRCh38
rs912806383 Chr.5: 35012923-35012923 on Build GRCh38
rs1000010591 Chr.5: 35013125-35013125 on Build GRCh38
rs967666876 Chr.5: 35013176-35013176 on Build GRCh38
rs1041880396 Chr.5: 35013383-35013383 on Build GRCh38
rs918654364 Chr.5: 35013248-35013248 on Build GRCh38
rs1056307077 Chr.5: 35013306-35013306 on Build GRCh38

View Full Product Details