Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35920794-35921341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPSL
Entrez Gene ID
133690
Gene Name
calcyphosine like
Gene Aliases
-
UniGene
Hs.55150
Gene Chromosome Location
Chr.5: 35904295-35938779 on Build GRCh38
Transcripts
NM_001042625.1 NM_144647.3 XM_006714444.3 XM_006714445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4523125 c.119G>A p.G40E Chr.5: 35921002-35921002 on Build GRCh38
482755 c.74C>G p.P25R Chr.5: 35921047-35921047 on Build GRCh38
1311120 c.48G>C p.K16N Chr.5: 35921073-35921073 on Build GRCh38
6546150 c.137G>A p.R46K Chr.5: 35920984-35920984 on Build GRCh38
1247333 c.131T>G p.L44R Chr.5: 35920990-35920990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375667323 Chr.5: 35921052-35921052 on Build GRCh38
rs746060094 Chr.5: 35921143-35921143 on Build GRCh38
rs887211381 Chr.5: 35921300-35921300 on Build GRCh38
rs769926042 Chr.5: 35921144-35921144 on Build GRCh38
rs867290050 Chr.5: 35920998-35920998 on Build GRCh38
rs915369416 Chr.5: 35921248-35921248 on Build GRCh38
rs748089997 Chr.5: 35921047-35921047 on Build GRCh38
rs755484518 Chr.5: 35920944-35920944 on Build GRCh38
rs553073975 Chr.5: 35921017-35921017 on Build GRCh38
rs1041693808 Chr.5: 35920895-35920895 on Build GRCh38
rs942992712 Chr.5: 35921251-35921251 on Build GRCh38
rs772171488 Chr.5: 35921008-35921008 on Build GRCh38
rs1000386390 Chr.5: 35920928-35920928 on Build GRCh38
rs200069514 Chr.5: 35920958-35920958 on Build GRCh38
rs889364257 Chr.5: 35921241-35921241 on Build GRCh38
rs776979043 Chr.5: 35921079-35921079 on Build GRCh38
rs575446116 Chr.5: 35921096-35921096 on Build GRCh38
rs935722472 Chr.5: 35921218-35921218 on Build GRCh38
rs976823033 Chr.5: 35920910-35920910 on Build GRCh38
rs370687666 Chr.5: 35921106-35921106 on Build GRCh38
rs1016743888 Chr.5: 35921145-35921145 on Build GRCh38
rs937815776 Chr.5: 35920856-35920856 on Build GRCh38
rs951313134 Chr.5: 35921116-35921116 on Build GRCh38
rs778624516 Chr.5: 35921044-35921044 on Build GRCh38
rs1007061080 Chr.5: 35921077-35921077 on Build GRCh38
rs371821824 Chr.5: 35921160-35921160 on Build GRCh38
rs765845508 Chr.5: 35921172-35921172 on Build GRCh38
rs771076658 Chr.5: 35921084-35921085 on Build GRCh38
rs777951969 Chr.5: 35921058-35921058 on Build GRCh38
rs903222122 Chr.5: 35920918-35920918 on Build GRCh38
rs924404460 Chr.5: 35921190-35921190 on Build GRCh38
rs3734107 Chr.5: 35921164-35921164 on Build GRCh38
rs537798616 Chr.5: 35920909-35920909 on Build GRCh38
rs780613084 Chr.5: 35920979-35920979 on Build GRCh38
rs779689546 Chr.5: 35921032-35921032 on Build GRCh38
rs926580505 Chr.5: 35920831-35920831 on Build GRCh38
rs781458664 Chr.5: 35921142-35921142 on Build GRCh38
rs139830420 Chr.5: 35921021-35921021 on Build GRCh38
rs749099220 Chr.5: 35920990-35920990 on Build GRCh38
rs773467327 Chr.5: 35921011-35921011 on Build GRCh38
rs1025938340 Chr.5: 35920913-35920913 on Build GRCh38
rs777068806 Chr.5: 35921015-35921015 on Build GRCh38
rs758288729 Chr.5: 35921121-35921121 on Build GRCh38
rs781257371 Chr.5: 35920823-35920823 on Build GRCh38
rs916881613 Chr.5: 35921001-35921001 on Build GRCh38
rs917285028 Chr.5: 35920865-35920865 on Build GRCh38
rs141223346 Chr.5: 35921256-35921256 on Build GRCh38
rs369193922 Chr.5: 35920963-35920963 on Build GRCh38
rs748095038 Chr.5: 35921004-35921004 on Build GRCh38
rs529629419 Chr.5: 35921313-35921313 on Build GRCh38
rs548690696 Chr.5: 35921269-35921269 on Build GRCh38
rs774829197 Chr.5: 35921163-35921163 on Build GRCh38
rs369384546 Chr.5: 35920986-35920986 on Build GRCh38
rs149827266 Chr.5: 35921031-35921031 on Build GRCh38
rs991162863 Chr.5: 35920997-35920997 on Build GRCh38
rs965441078 Chr.5: 35920899-35920899 on Build GRCh38
rs1041461860 Chr.5: 35921018-35921018 on Build GRCh38
rs373881430 Chr.5: 35921085-35921085 on Build GRCh38
rs375106222 Chr.5: 35921107-35921107 on Build GRCh38
rs377089913 Chr.5: 35921033-35921033 on Build GRCh38
rs777848051 Chr.5: 35921128-35921128 on Build GRCh38
rs6859892 Chr.5: 35920967-35920967 on Build GRCh38
rs148224785 Chr.5: 35920937-35920937 on Build GRCh38
rs746998483 Chr.5: 35921136-35921136 on Build GRCh38
rs770162293 Chr.5: 35921086-35921086 on Build GRCh38
rs749872923 Chr.5: 35921095-35921095 on Build GRCh38
rs202096708 Chr.5: 35921023-35921023 on Build GRCh38
rs774343844 Chr.5: 35920999-35920999 on Build GRCh38
rs775862681 Chr.5: 35921157-35921157 on Build GRCh38
rs151068422 Chr.5: 35921012-35921012 on Build GRCh38
rs894076410 Chr.5: 35921049-35921049 on Build GRCh38
rs17851540 Chr.5: 35921051-35921051 on Build GRCh38
rs200635899 Chr.5: 35921059-35921059 on Build GRCh38
rs371171403 Chr.5: 35920983-35920983 on Build GRCh38
rs187877022 Chr.5: 35921022-35921022 on Build GRCh38
rs771201246 Chr.5: 35921062-35921062 on Build GRCh38
rs1018200356 Chr.5: 35920830-35920830 on Build GRCh38
rs761720452 Chr.5: 35920822-35920822 on Build GRCh38
rs754791143 Chr.5: 35921034-35921034 on Build GRCh38
rs867185451 Chr.5: 35921125-35921125 on Build GRCh38
rs771111318 Chr.5: 35921141-35921141 on Build GRCh38
rs950176811 Chr.5: 35920883-35920883 on Build GRCh38
rs368551299 Chr.5: 35921104-35921104 on Build GRCh38
rs754628339 Chr.5: 35921105-35921105 on Build GRCh38
rs768414245 Chr.5: 35920994-35920994 on Build GRCh38
rs763465521 Chr.5: 35921093-35921093 on Build GRCh38
rs767009111 Chr.5: 35921025-35921025 on Build GRCh38
rs1005401812 Chr.5: 35921138-35921138 on Build GRCh38
rs1039922645 Chr.5: 35921254-35921254 on Build GRCh38
rs540301117 Chr.5: 35921179-35921179 on Build GRCh38
rs971076465 Chr.5: 35921176-35921176 on Build GRCh38
rs984727413 Chr.5: 35920825-35920825 on Build GRCh38
rs1047481086 Chr.5: 35921296-35921296 on Build GRCh38
rs749954681 Chr.5: 35921030-35921030 on Build GRCh38
rs368843089 Chr.5: 35921156-35921156 on Build GRCh38
rs76940881 Chr.5: 35921174-35921174 on Build GRCh38
rs560266011 Chr.5: 35921243-35921243 on Build GRCh38
rs570721664 Chr.5: 35920949-35920949 on Build GRCh38
rs1056310321 Chr.5: 35920861-35920861 on Build GRCh38
rs878903625 Chr.5: 35921019-35921019 on Build GRCh38

View Full Product Details