Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 40909407-40909908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7
Entrez Gene ID
730
Gene Name
complement component 7
Gene Aliases
-
UniGene
Hs.78065
Gene Chromosome Location
Chr.5: 40909497-40982939 on Build GRCh38
Transcripts
NM_000587.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4556708 c.6G>A p.K2K Chr.5: 40909616-40909616 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780368424 Chr.5: 40909740-40909740 on Build GRCh38
rs564759065 Chr.5: 40909769-40909769 on Build GRCh38
rs984233545 Chr.5: 40909433-40909433 on Build GRCh38
rs919206651 Chr.5: 40909522-40909522 on Build GRCh38
rs879056067 Chr.5: 40909430-40909430 on Build GRCh38
rs138172295 Chr.5: 40909695-40909695 on Build GRCh38
rs906801011 Chr.5: 40909651-40909651 on Build GRCh38
rs539088570 Chr.5: 40909459-40909459 on Build GRCh38
rs780687790 Chr.5: 40909573-40909575 on Build GRCh38
rs987415437 Chr.5: 40909801-40909801 on Build GRCh38
rs117513464 Chr.5: 40909717-40909717 on Build GRCh38
rs764328445 Chr.5: 40909583-40909583 on Build GRCh38
rs573909175 Chr.5: 40909648-40909648 on Build GRCh38
rs1006050018 Chr.5: 40909711-40909711 on Build GRCh38
rs910338278 Chr.5: 40909815-40909815 on Build GRCh38
rs775574494 Chr.5: 40909571-40909571 on Build GRCh38
rs571693986 Chr.5: 40909437-40909437 on Build GRCh38
rs971969021 Chr.5: 40909485-40909485 on Build GRCh38
rs560608668 Chr.5: 40909704-40909704 on Build GRCh38
rs115456770 Chr.5: 40909592-40909592 on Build GRCh38
rs546418997 Chr.5: 40909823-40909824 on Build GRCh38
rs989181669 Chr.5: 40909859-40909859 on Build GRCh38
rs377586073 Chr.5: 40909870-40909870 on Build GRCh38
rs754308110 Chr.5: 40909431-40909431 on Build GRCh38
rs763097788 Chr.5: 40909581-40909581 on Build GRCh38
rs778287275 Chr.5: 40909618-40909618 on Build GRCh38
rs531925674 Chr.5: 40909772-40909772 on Build GRCh38
rs374810598 Chr.5: 40909621-40909621 on Build GRCh38
rs926063221 Chr.5: 40909535-40909535 on Build GRCh38
rs998449805 Chr.5: 40909767-40909767 on Build GRCh38
rs576261033 Chr.5: 40909532-40909532 on Build GRCh38
rs954759660 Chr.5: 40909746-40909746 on Build GRCh38
rs752191312 Chr.5: 40909629-40909629 on Build GRCh38
rs539333412 Chr.5: 40909669-40909669 on Build GRCh38
rs961411397 Chr.5: 40909469-40909469 on Build GRCh38
rs779041347 Chr.5: 40909555-40909555 on Build GRCh38
rs1029030963 Chr.5: 40909817-40909817 on Build GRCh38
rs961357399 Chr.5: 40909726-40909726 on Build GRCh38
rs558030823 Chr.5: 40909531-40909531 on Build GRCh38
rs189419523 Chr.5: 40909728-40909728 on Build GRCh38
rs750983966 Chr.5: 40909608-40909608 on Build GRCh38
rs367577475 Chr.5: 40909479-40909479 on Build GRCh38
rs769540731 Chr.5: 40909774-40909774 on Build GRCh38
rs756607429 Chr.5: 40909617-40909617 on Build GRCh38
rs758008310 Chr.5: 40909632-40909632 on Build GRCh38
rs540446790 Chr.5: 40909470-40909470 on Build GRCh38
rs767848773 Chr.5: 40909599-40909599 on Build GRCh38
rs867510067 Chr.5: 40909758-40909758 on Build GRCh38
rs550323193 Chr.5: 40909831-40909831 on Build GRCh38
rs770066898 Chr.5: 40909565-40909565 on Build GRCh38
rs757493190 Chr.5: 40909435-40909435 on Build GRCh38
rs888221846 Chr.5: 40909686-40909686 on Build GRCh38
rs910399830 Chr.5: 40909536-40909536 on Build GRCh38
rs1048131405 Chr.5: 40909674-40909674 on Build GRCh38
rs561756330 Chr.5: 40909647-40909647 on Build GRCh38
rs751904932 Chr.5: 40909584-40909584 on Build GRCh38
rs759522057 Chr.5: 40909564-40909564 on Build GRCh38

View Full Product Details