Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 50402201-50402715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMB
Entrez Gene ID
133418
Gene Name
embigin
Gene Aliases
GP70
UniGene
Hs.561411
Gene Chromosome Location
Chr.5: 50396192-50443299 on Build GRCh38
Transcripts
NM_198449.2 XM_011543146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1068533 c.889G>T p.E297* Chr.5: 50402308-50402308 on Build GRCh38
3616826 c.883G>A p.G295R Chr.5: 50402314-50402314 on Build GRCh38
4680851 c.894T>C p.F298F Chr.5: 50402303-50402303 on Build GRCh38
5169300 c.897G>T p.E299D Chr.5: 50402300-50402300 on Build GRCh38
2156580 c.878-1G>A p.? Chr.5: 50402320-50402320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766835596 Chr.5: 50402265-50402265 on Build GRCh38
rs763758075 Chr.5: 50402332-50402332 on Build GRCh38
rs575621641 Chr.5: 50402630-50402630 on Build GRCh38
rs181260350 Chr.5: 50402378-50402378 on Build GRCh38
rs954106075 Chr.5: 50402426-50402426 on Build GRCh38
rs759837472 Chr.5: 50402316-50402316 on Build GRCh38
rs965924170 Chr.5: 50402456-50402456 on Build GRCh38
rs796675413 Chr.5: 50402401-50402404 on Build GRCh38
rs911835313 Chr.5: 50402442-50402442 on Build GRCh38
rs1028751501 Chr.5: 50402420-50402420 on Build GRCh38
rs537675754 Chr.5: 50402353-50402353 on Build GRCh38
rs770405192 Chr.5: 50402593-50402594 on Build GRCh38
rs567832431 Chr.5: 50402616-50402616 on Build GRCh38
rs761258046 Chr.5: 50402264-50402264 on Build GRCh38
rs866356280 Chr.5: 50402519-50402519 on Build GRCh38
rs375934195 Chr.5: 50402377-50402377 on Build GRCh38
rs756695537 Chr.5: 50402343-50402343 on Build GRCh38
rs13172025 Chr.5: 50402346-50402346 on Build GRCh38
rs946464875 Chr.5: 50402597-50402597 on Build GRCh38
rs926360479 Chr.5: 50402654-50402654 on Build GRCh38
rs776922184 Chr.5: 50402279-50402282 on Build GRCh38
rs756018696 Chr.5: 50402354-50402354 on Build GRCh38
rs779909557 Chr.5: 50402363-50402363 on Build GRCh38
rs1052204697 Chr.5: 50402234-50402234 on Build GRCh38
rs754207099 Chr.5: 50402294-50402294 on Build GRCh38
rs564051994 Chr.5: 50402467-50402468 on Build GRCh38
rs986761873 Chr.5: 50402432-50402432 on Build GRCh38
rs545861380 Chr.5: 50402471-50402471 on Build GRCh38
rs751174022 Chr.5: 50402340-50402340 on Build GRCh38
rs375129797 Chr.5: 50402381-50402381 on Build GRCh38
rs369486944 Chr.5: 50402247-50402247 on Build GRCh38
rs112624623 Chr.5: 50402617-50402617 on Build GRCh38
rs1004443456 Chr.5: 50402683-50402683 on Build GRCh38
rs556031370 Chr.5: 50402556-50402556 on Build GRCh38
rs1015740267 Chr.5: 50402399-50402399 on Build GRCh38
rs561347491 Chr.5: 50402678-50402678 on Build GRCh38
rs937713857 Chr.5: 50402671-50402671 on Build GRCh38
rs568724287 Chr.5: 50402308-50402308 on Build GRCh38
rs995566255 Chr.5: 50402415-50402415 on Build GRCh38
rs749063059 Chr.5: 50402367-50402367 on Build GRCh38
rs913716522 Chr.5: 50402592-50402592 on Build GRCh38
rs185848056 Chr.5: 50402392-50402392 on Build GRCh38
rs750005812 Chr.5: 50402347-50402347 on Build GRCh38
rs767529749 Chr.5: 50402646-50402646 on Build GRCh38
rs896720851 Chr.5: 50402380-50402380 on Build GRCh38
rs372670860 Chr.5: 50402421-50402421 on Build GRCh38
rs58664042 Chr.5: 50402642-50402642 on Build GRCh38
rs541382174 Chr.5: 50402677-50402677 on Build GRCh38

View Full Product Details