Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 52849223-52849722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA1
Entrez Gene ID
3672
Gene Name
integrin subunit alpha 1
Gene Aliases
CD49a, VLA1
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52788302-52953655 on Build GRCh38
Transcripts
NM_181501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2156419 c.70C>T p.R24C Chr.5: 52849373-52849373 on Build GRCh38
3855482 c.126G>A p.P42P Chr.5: 52849429-52849429 on Build GRCh38
592750 c.112A>T p.T38S Chr.5: 52849415-52849415 on Build GRCh38
5007025 c.92T>A p.V31D Chr.5: 52849395-52849395 on Build GRCh38
1137351 c.182+1G>T p.? Chr.5: 52849486-52849486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923367974 Chr.5: 52849596-52849596 on Build GRCh38
rs567222496 Chr.5: 52849466-52849466 on Build GRCh38
rs923369259 Chr.5: 52849328-52849328 on Build GRCh38
rs759463731 Chr.5: 52849374-52849374 on Build GRCh38
rs774902695 Chr.5: 52849337-52849337 on Build GRCh38
rs371540422 Chr.5: 52849491-52849491 on Build GRCh38
rs141004541 Chr.5: 52849509-52849509 on Build GRCh38
rs201300374 Chr.5: 52849646-52849647 on Build GRCh38
rs768243715 Chr.5: 52849535-52849535 on Build GRCh38
rs181851084 Chr.5: 52849293-52849293 on Build GRCh38
rs376322589 Chr.5: 52849347-52849347 on Build GRCh38
rs201009495 Chr.5: 52849429-52849429 on Build GRCh38
rs754071710 Chr.5: 52849496-52849496 on Build GRCh38
rs776279363 Chr.5: 52849356-52849356 on Build GRCh38
rs752812338 Chr.5: 52849549-52849549 on Build GRCh38
rs769116421 Chr.5: 52849331-52849331 on Build GRCh38
rs780856119 Chr.5: 52849382-52849382 on Build GRCh38
rs547998897 Chr.5: 52849256-52849256 on Build GRCh38
rs78526623 Chr.5: 52849658-52849658 on Build GRCh38
rs775452162 Chr.5: 52849319-52849319 on Build GRCh38
rs772439591 Chr.5: 52849428-52849428 on Build GRCh38
rs987209585 Chr.5: 52849576-52849576 on Build GRCh38
rs374090897 Chr.5: 52849424-52849424 on Build GRCh38
rs13360413 Chr.5: 52849559-52849559 on Build GRCh38
rs538432113 Chr.5: 52849557-52849557 on Build GRCh38
rs189872081 Chr.5: 52849655-52849655 on Build GRCh38
rs562010752 Chr.5: 52849645-52849646 on Build GRCh38
rs762703058 Chr.5: 52849527-52849527 on Build GRCh38
rs764411106 Chr.5: 52849683-52849683 on Build GRCh38
rs184979149 Chr.5: 52849607-52849607 on Build GRCh38
rs776860557 Chr.5: 52849329-52849329 on Build GRCh38
rs764756563 Chr.5: 52849362-52849362 on Build GRCh38
rs758022183 Chr.5: 52849372-52849372 on Build GRCh38
rs16880359 Chr.5: 52849300-52849300 on Build GRCh38
rs542954703 Chr.5: 52849640-52849640 on Build GRCh38
rs768778866 Chr.5: 52849423-52849423 on Build GRCh38
rs911019497 Chr.5: 52849267-52849267 on Build GRCh38
rs936668438 Chr.5: 52849685-52849685 on Build GRCh38
rs780361762 Chr.5: 52849392-52849392 on Build GRCh38
rs752127821 Chr.5: 52849364-52849364 on Build GRCh38
rs759141937 Chr.5: 52849361-52849361 on Build GRCh38
rs1051955294 Chr.5: 52849274-52849274 on Build GRCh38
rs775229394 Chr.5: 52849517-52849517 on Build GRCh38
rs368205248 Chr.5: 52849510-52849510 on Build GRCh38
rs778993970 Chr.5: 52849506-52849506 on Build GRCh38
rs775350374 Chr.5: 52849449-52849449 on Build GRCh38
rs370114923 Chr.5: 52849367-52849367 on Build GRCh38
rs1047752658 Chr.5: 52849385-52849385 on Build GRCh38
rs911649566 Chr.5: 52849652-52849652 on Build GRCh38
rs763714314 Chr.5: 52849463-52849463 on Build GRCh38
rs764767736 Chr.5: 52849433-52849433 on Build GRCh38
rs567216405 Chr.5: 52849472-52849472 on Build GRCh38
rs762150639 Chr.5: 52849344-52849344 on Build GRCh38
rs750151289 Chr.5: 52849477-52849477 on Build GRCh38
rs560339233 Chr.5: 52849375-52849375 on Build GRCh38
rs764123493 Chr.5: 52849373-52849373 on Build GRCh38
rs746066086 Chr.5: 52849384-52849384 on Build GRCh38
rs754984515 Chr.5: 52849498-52849498 on Build GRCh38
rs762539823 Chr.5: 52849460-52849460 on Build GRCh38
rs749360848 Chr.5: 52849407-52849407 on Build GRCh38
rs757065276 Chr.5: 52849381-52849381 on Build GRCh38
rs771386570 Chr.5: 52849516-52849516 on Build GRCh38
rs1024816581 Chr.5: 52849451-52849451 on Build GRCh38
rs978174195 Chr.5: 52849613-52849613 on Build GRCh38
rs966788233 Chr.5: 52849565-52849565 on Build GRCh38
rs372555630 Chr.5: 52849310-52849310 on Build GRCh38
rs755999231 Chr.5: 52849479-52849479 on Build GRCh38
rs78958139 Chr.5: 52849546-52849546 on Build GRCh38
rs985260489 Chr.5: 52849263-52849263 on Build GRCh38
rs945482232 Chr.5: 52849601-52849601 on Build GRCh38
rs747330970 Chr.5: 52849513-52849513 on Build GRCh38
rs200460098 Chr.5: 52849249-52849250 on Build GRCh38
rs977014874 Chr.5: 52849564-52849564 on Build GRCh38
rs367961574 Chr.5: 52849473-52849473 on Build GRCh38

View Full Product Details