Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55108004-55108497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMA
Entrez Gene ID
3001
Gene Name
granzyme A
Gene Aliases
CTLA3, HFSP
UniGene
Hs.90708
Gene Chromosome Location
Chr.5: 55102646-55110252 on Build GRCh38
Transcripts
NM_006144.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855591 c.542A>G p.D181G Chr.5: 55108309-55108309 on Build GRCh38
6103966 c.592G>T p.A198S Chr.5: 55108359-55108359 on Build GRCh38
3855590 c.489C>T p.S163S Chr.5: 55108256-55108256 on Build GRCh38
4141997 c.372A>C p.A124A Chr.5: 55108139-55108139 on Build GRCh38
6189119 c.539A>G p.N180S Chr.5: 55108306-55108306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541308949 Chr.5: 55108395-55108395 on Build GRCh38
rs754031523 Chr.5: 55108285-55108285 on Build GRCh38
rs368948899 Chr.5: 55108311-55108311 on Build GRCh38
rs752730777 Chr.5: 55108425-55108426 on Build GRCh38
rs904744139 Chr.5: 55108043-55108043 on Build GRCh38
rs151159567 Chr.5: 55108314-55108314 on Build GRCh38
rs530263188 Chr.5: 55108094-55108094 on Build GRCh38
rs778251293 Chr.5: 55108319-55108319 on Build GRCh38
rs779513892 Chr.5: 55108231-55108231 on Build GRCh38
rs139102453 Chr.5: 55108316-55108316 on Build GRCh38
rs767710019 Chr.5: 55108324-55108325 on Build GRCh38
rs1030106381 Chr.5: 55108228-55108228 on Build GRCh38
rs2270627 Chr.5: 55108225-55108225 on Build GRCh38
rs374377355 Chr.5: 55108322-55108322 on Build GRCh38
rs751955576 Chr.5: 55108250-55108250 on Build GRCh38
rs145934520 Chr.5: 55108261-55108261 on Build GRCh38
rs771220491 Chr.5: 55108327-55108327 on Build GRCh38
rs766675685 Chr.5: 55108284-55108284 on Build GRCh38
rs767290041 Chr.5: 55108266-55108266 on Build GRCh38
rs772507747 Chr.5: 55108238-55108238 on Build GRCh38
rs915966919 Chr.5: 55108341-55108341 on Build GRCh38
rs760470481 Chr.5: 55108372-55108372 on Build GRCh38
rs61730831 Chr.5: 55108350-55108350 on Build GRCh38
rs368307665 Chr.5: 55108374-55108374 on Build GRCh38
rs778366524 Chr.5: 55108445-55108445 on Build GRCh38
rs554646660 Chr.5: 55108264-55108264 on Build GRCh38
rs556099504 Chr.5: 55108190-55108190 on Build GRCh38
rs149445760 Chr.5: 55108257-55108257 on Build GRCh38
rs61730832 Chr.5: 55108282-55108282 on Build GRCh38
rs576458803 Chr.5: 55108371-55108371 on Build GRCh38
rs756084513 Chr.5: 55108079-55108079 on Build GRCh38
rs759740976 Chr.5: 55108276-55108277 on Build GRCh38
rs754612547 Chr.5: 55108256-55108256 on Build GRCh38
rs1875506 Chr.5: 55108187-55108187 on Build GRCh38
rs751516645 Chr.5: 55108168-55108168 on Build GRCh38
rs565866692 Chr.5: 55108244-55108244 on Build GRCh38
rs907128511 Chr.5: 55108281-55108281 on Build GRCh38
rs142272396 Chr.5: 55108248-55108248 on Build GRCh38
rs766895884 Chr.5: 55108236-55108237 on Build GRCh38
rs748961001 Chr.5: 55108436-55108436 on Build GRCh38
rs775130512 Chr.5: 55108156-55108156 on Build GRCh38
rs756169021 Chr.5: 55108433-55108433 on Build GRCh38
rs774550337 Chr.5: 55108106-55108106 on Build GRCh38
rs752035472 Chr.5: 55108415-55108415 on Build GRCh38
rs149924275 Chr.5: 55108345-55108345 on Build GRCh38
rs142259151 Chr.5: 55108363-55108363 on Build GRCh38
rs563675975 Chr.5: 55108119-55108119 on Build GRCh38
rs558289848 Chr.5: 55108351-55108351 on Build GRCh38
rs753992275 Chr.5: 55108226-55108226 on Build GRCh38
rs559498444 Chr.5: 55108082-55108082 on Build GRCh38
rs369222212 Chr.5: 55108162-55108162 on Build GRCh38
rs372514056 Chr.5: 55108142-55108142 on Build GRCh38
rs764180233 Chr.5: 55108381-55108381 on Build GRCh38
rs762765865 Chr.5: 55108158-55108158 on Build GRCh38
rs552295796 Chr.5: 55108130-55108130 on Build GRCh38
rs1051846 Chr.5: 55108229-55108229 on Build GRCh38
rs750288604 Chr.5: 55108272-55108272 on Build GRCh38
rs778742153 Chr.5: 55108342-55108342 on Build GRCh38
rs977069707 Chr.5: 55108369-55108369 on Build GRCh38
rs371890271 Chr.5: 55108444-55108444 on Build GRCh38
rs374209345 Chr.5: 55108416-55108416 on Build GRCh38
rs769525448 Chr.5: 55108252-55108252 on Build GRCh38
rs142064253 Chr.5: 55108312-55108312 on Build GRCh38
rs758950311 Chr.5: 55108396-55108396 on Build GRCh38
rs887102692 Chr.5: 55108242-55108242 on Build GRCh38
rs200886364 Chr.5: 55108421-55108421 on Build GRCh38
rs189633369 Chr.5: 55108100-55108100 on Build GRCh38
rs997168528 Chr.5: 55108380-55108381 on Build GRCh38
rs776846664 Chr.5: 55108375-55108375 on Build GRCh38
rs774306246 Chr.5: 55108388-55108388 on Build GRCh38
rs764644623 Chr.5: 55108410-55108410 on Build GRCh38
rs748574049 Chr.5: 55108108-55108108 on Build GRCh38
rs776180292 Chr.5: 55108246-55108246 on Build GRCh38
rs779138746 Chr.5: 55108300-55108300 on Build GRCh38
rs772039884 Chr.5: 55108367-55108367 on Build GRCh38
rs755045725 Chr.5: 55108298-55108298 on Build GRCh38
rs756420443 Chr.5: 55108201-55108201 on Build GRCh38
rs750590418 Chr.5: 55108200-55108200 on Build GRCh38
rs3104233 Chr.5: 55108129-55108129 on Build GRCh38
rs764138358 Chr.5: 55108164-55108164 on Build GRCh38
rs921433403 Chr.5: 55108455-55108455 on Build GRCh38
rs3104233 Chr.5: 55108129-55108129 on Build GRCh38

View Full Product Details