Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 58493533-58494029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAPT
Entrez Gene ID
202309
Gene Name
GRB2-binding adaptor protein, transmembrane
Gene Aliases
C5orf29
UniGene
Hs.547697
Gene Chromosome Location
Chr.5: 58491435-58496358 on Build GRCh38
Transcripts
NM_001304428.1 NM_001304429.1 NM_001304431.1 NM_152687.3

SNP Detail

SNP ID Chromosome Location
rs1021187527 Chr.5: 58493621-58493621 on Build GRCh38
rs546997771 Chr.5: 58493943-58493943 on Build GRCh38
rs890674897 Chr.5: 58493858-58493858 on Build GRCh38
rs1024390027 Chr.5: 58493832-58493832 on Build GRCh38
rs185683957 Chr.5: 58493973-58493973 on Build GRCh38
rs898787225 Chr.5: 58493598-58493598 on Build GRCh38
rs1053465004 Chr.5: 58493799-58493799 on Build GRCh38
rs552784021 Chr.5: 58493848-58493848 on Build GRCh38
rs545985821 Chr.5: 58493610-58493610 on Build GRCh38
rs986858494 Chr.5: 58493983-58493983 on Build GRCh38
rs746732405 Chr.5: 58493573-58493573 on Build GRCh38
rs983135444 Chr.5: 58493719-58493719 on Build GRCh38
rs75600256 Chr.5: 58493808-58493808 on Build GRCh38
rs143954927 Chr.5: 58493643-58493643 on Build GRCh38
rs561380117 Chr.5: 58493659-58493659 on Build GRCh38
rs940096019 Chr.5: 58493740-58493740 on Build GRCh38
rs142688260 Chr.5: 58493794-58493794 on Build GRCh38
rs932675086 Chr.5: 58493763-58493763 on Build GRCh38
rs1043051626 Chr.5: 58493857-58493857 on Build GRCh38
rs545357142 Chr.5: 58493642-58493642 on Build GRCh38
rs3215050 Chr.5: 58493844-58493844 on Build GRCh38
rs397802869 Chr.5: 58493846-58493846 on Build GRCh38
rs1046188233 Chr.5: 58493781-58493781 on Build GRCh38
rs140157334 Chr.5: 58493969-58493969 on Build GRCh38
rs994947552 Chr.5: 58493923-58493923 on Build GRCh38
rs1013957310 Chr.5: 58493825-58493825 on Build GRCh38
rs1038556118 Chr.5: 58493761-58493761 on Build GRCh38
rs1009174294 Chr.5: 58493831-58493831 on Build GRCh38
rs552887865 Chr.5: 58493718-58493718 on Build GRCh38
rs998998528 Chr.5: 58493862-58493862 on Build GRCh38
rs748110503 Chr.5: 58493737-58493737 on Build GRCh38
rs147359272 Chr.5: 58493797-58493797 on Build GRCh38
rs149474120 Chr.5: 58493638-58493638 on Build GRCh38
rs766020634 Chr.5: 58493626-58493627 on Build GRCh38
rs542117583 Chr.5: 58493627-58493627 on Build GRCh38
rs964874787 Chr.5: 58493896-58493896 on Build GRCh38
rs977322277 Chr.5: 58493652-58493652 on Build GRCh38
rs148233134 Chr.5: 58493714-58493714 on Build GRCh38
rs958370848 Chr.5: 58493625-58493625 on Build GRCh38
rs575314168 Chr.5: 58493570-58493570 on Build GRCh38
rs76703931 Chr.5: 58493881-58493881 on Build GRCh38
rs957313641 Chr.5: 58493664-58493664 on Build GRCh38
rs375376455 Chr.5: 58493812-58493812 on Build GRCh38
rs973476997 Chr.5: 58493665-58493665 on Build GRCh38
rs955072507 Chr.5: 58493624-58493624 on Build GRCh38
rs535161043 Chr.5: 58493911-58493911 on Build GRCh38
rs1027728831 Chr.5: 58493951-58493951 on Build GRCh38
rs1009488811 Chr.5: 58493875-58493875 on Build GRCh38
rs183276019 Chr.5: 58493716-58493716 on Build GRCh38

View Full Product Details