Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62348053-62348598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2A
Entrez Gene ID
3796
Gene Name
kinesin family member 2A
Gene Aliases
CDCBM3, HK2, KIF2
UniGene
Hs.558351
Gene Chromosome Location
Chr.5: 62306162-62387184 on Build GRCh38
Transcripts
NM_001098511.2 NM_001243952.1 NM_001243953.1 NM_004520.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855798 c.188A>C p.K63T Chr.5: 62348157-62348157 on Build GRCh38
1069238 c.189A>C p.K63N Chr.5: 62348158-62348158 on Build GRCh38
449950 c.127G>A p.E43K Chr.5: 62348096-62348096 on Build GRCh38
1069236 c.140C>T p.P47L Chr.5: 62348109-62348109 on Build GRCh38
3373930 c.255A>T p.S85S Chr.5: 62348143-62348143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780248688 Chr.5: 62348090-62348090 on Build GRCh38
rs146893123 Chr.5: 62348350-62348350 on Build GRCh38
rs768662373 Chr.5: 62348191-62348192 on Build GRCh38
rs1008585723 Chr.5: 62348563-62348563 on Build GRCh38
rs756381140 Chr.5: 62348209-62348209 on Build GRCh38
rs3194481 Chr.5: 62348135-62348135 on Build GRCh38
rs763972861 Chr.5: 62348161-62348161 on Build GRCh38
rs769337181 Chr.5: 62348141-62348141 on Build GRCh38
rs758544103 Chr.5: 62348567-62348570 on Build GRCh38
rs748463159 Chr.5: 62348114-62348114 on Build GRCh38
rs772419116 Chr.5: 62348122-62348122 on Build GRCh38
rs923750973 Chr.5: 62348367-62348367 on Build GRCh38
rs755018696 Chr.5: 62348217-62348217 on Build GRCh38
rs73102041 Chr.5: 62348083-62348083 on Build GRCh38
rs779425808 Chr.5: 62348240-62348240 on Build GRCh38
rs767409036 Chr.5: 62348188-62348188 on Build GRCh38
rs11545222 Chr.5: 62348144-62348144 on Build GRCh38
rs950127001 Chr.5: 62348245-62348245 on Build GRCh38
rs1055026222 Chr.5: 62348227-62348227 on Build GRCh38
rs369749378 Chr.5: 62348202-62348202 on Build GRCh38
rs955223691 Chr.5: 62348372-62348372 on Build GRCh38
rs571500254 Chr.5: 62348127-62348127 on Build GRCh38
rs749857734 Chr.5: 62348094-62348094 on Build GRCh38
rs931626300 Chr.5: 62348453-62348453 on Build GRCh38
rs138676317 Chr.5: 62348078-62348078 on Build GRCh38
rs774962082 Chr.5: 62348151-62348151 on Build GRCh38
rs115287860 Chr.5: 62348524-62348524 on Build GRCh38
rs919775850 Chr.5: 62348497-62348497 on Build GRCh38
rs536209140 Chr.5: 62348474-62348474 on Build GRCh38
rs986443501 Chr.5: 62348430-62348430 on Build GRCh38
rs1028358408 Chr.5: 62348190-62348190 on Build GRCh38
rs373786687 Chr.5: 62348387-62348387 on Build GRCh38
rs903266846 Chr.5: 62348288-62348288 on Build GRCh38
rs1053700342 Chr.5: 62348431-62348431 on Build GRCh38
rs755463219 Chr.5: 62348104-62348104 on Build GRCh38
rs937891761 Chr.5: 62348222-62348222 on Build GRCh38
rs1014219179 Chr.5: 62348237-62348237 on Build GRCh38
rs750742767 Chr.5: 62348197-62348197 on Build GRCh38
rs376138524 Chr.5: 62348182-62348182 on Build GRCh38
rs759034267 Chr.5: 62348530-62348530 on Build GRCh38
rs766657872 Chr.5: 62348210-62348210 on Build GRCh38
rs555636164 Chr.5: 62348517-62348517 on Build GRCh38
rs894478216 Chr.5: 62348228-62348228 on Build GRCh38
rs553818858 Chr.5: 62348402-62348402 on Build GRCh38
rs1033129240 Chr.5: 62348316-62348316 on Build GRCh38
rs756321886 Chr.5: 62348084-62348084 on Build GRCh38
rs761527371 Chr.5: 62348177-62348177 on Build GRCh38
rs573710637 Chr.5: 62348473-62348473 on Build GRCh38
rs1055457835 Chr.5: 62348542-62348542 on Build GRCh38
rs762480935 Chr.5: 62348155-62348155 on Build GRCh38
rs559315358 Chr.5: 62348321-62348321 on Build GRCh38
rs1028546355 Chr.5: 62348361-62348361 on Build GRCh38
rs368489768 Chr.5: 62348444-62348444 on Build GRCh38
rs961102958 Chr.5: 62348458-62348458 on Build GRCh38
rs999015253 Chr.5: 62348315-62348315 on Build GRCh38
rs375423887 Chr.5: 62348216-62348216 on Build GRCh38
rs893752236 Chr.5: 62348433-62348433 on Build GRCh38
rs779434615 Chr.5: 62348108-62348108 on Build GRCh38
rs780422626 Chr.5: 62348551-62348551 on Build GRCh38
rs774278609 Chr.5: 62348162-62348162 on Build GRCh38
rs772745432 Chr.5: 62348376-62348376 on Build GRCh38

View Full Product Details