Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65149834-65150327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS6
Entrez Gene ID
11174
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 6
Gene Aliases
ADAM-TS 6, ADAM-TS6, ADAMTS-6
UniGene
Hs.482291
Gene Chromosome Location
Chr.5: 65148736-65482014 on Build GRCh38
Transcripts
NM_197941.3 NR_135689.1 XM_011543113.2 XM_011543114.2 XM_011543115.2 XM_011543116.2 XM_011543117.1 XM_011543121.2 XM_011543123.2 XM_011543124.2 XM_011543125.1

SNP Detail

SNP ID Chromosome Location
rs1051662265 Chr.5: 65149950-65149950 on Build GRCh38
rs892916576 Chr.5: 65150155-65150155 on Build GRCh38
rs572947959 Chr.5: 65150060-65150060 on Build GRCh38
rs143566104 Chr.5: 65150255-65150255 on Build GRCh38
rs1030099336 Chr.5: 65150200-65150200 on Build GRCh38
rs139541144 Chr.5: 65150247-65150247 on Build GRCh38
rs974153157 Chr.5: 65149898-65149898 on Build GRCh38
rs543755544 Chr.5: 65150234-65150234 on Build GRCh38
rs937111949 Chr.5: 65150301-65150301 on Build GRCh38
rs1055898258 Chr.5: 65150052-65150052 on Build GRCh38
rs898910518 Chr.5: 65150038-65150038 on Build GRCh38
rs1000774466 Chr.5: 65150098-65150098 on Build GRCh38
rs763910438 Chr.5: 65150128-65150128 on Build GRCh38
rs906869837 Chr.5: 65150216-65150216 on Build GRCh38
rs370103510 Chr.5: 65150059-65150059 on Build GRCh38
rs575248173 Chr.5: 65150161-65150161 on Build GRCh38
rs920033875 Chr.5: 65149984-65149984 on Build GRCh38
rs1009998731 Chr.5: 65150164-65150164 on Build GRCh38
rs1037504471 Chr.5: 65150020-65150020 on Build GRCh38
rs1001026451 Chr.5: 65150145-65150145 on Build GRCh38
rs903411865 Chr.5: 65150075-65150075 on Build GRCh38
rs189103303 Chr.5: 65150240-65150240 on Build GRCh38
rs372952838 Chr.5: 65150300-65150300 on Build GRCh38
rs964122388 Chr.5: 65149886-65149886 on Build GRCh38
rs1054114307 Chr.5: 65150148-65150148 on Build GRCh38
rs539558059 Chr.5: 65150044-65150044 on Build GRCh38
rs775212244 Chr.5: 65150009-65150009 on Build GRCh38
rs973681550 Chr.5: 65150282-65150282 on Build GRCh38
rs760259084 Chr.5: 65150072-65150072 on Build GRCh38
rs944249408 Chr.5: 65150079-65150079 on Build GRCh38
rs933252615 Chr.5: 65149949-65149949 on Build GRCh38
rs17812189 Chr.5: 65149881-65149881 on Build GRCh38
rs964510900 Chr.5: 65150121-65150121 on Build GRCh38
rs555161291 Chr.5: 65150158-65150158 on Build GRCh38
rs1020509113 Chr.5: 65150069-65150069 on Build GRCh38
rs541646919 Chr.5: 65150116-65150116 on Build GRCh38
rs111792024 Chr.5: 65150256-65150256 on Build GRCh38
rs975860150 Chr.5: 65149888-65149888 on Build GRCh38
rs929950695 Chr.5: 65150019-65150019 on Build GRCh38
rs907770124 Chr.5: 65149961-65149961 on Build GRCh38
rs922619342 Chr.5: 65149889-65149889 on Build GRCh38
rs955376268 Chr.5: 65150214-65150214 on Build GRCh38
rs1039964470 Chr.5: 65150122-65150122 on Build GRCh38
rs540501368 Chr.5: 65149980-65149993 on Build GRCh38
rs556966773 Chr.5: 65149887-65149887 on Build GRCh38
rs576743355 Chr.5: 65149919-65149919 on Build GRCh38
rs536904859 Chr.5: 65149885-65149885 on Build GRCh38
rs761401335 Chr.5: 65150271-65150271 on Build GRCh38
rs912583622 Chr.5: 65150076-65150076 on Build GRCh38

View Full Product Details