Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65172746-65173254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS6
Entrez Gene ID
11174
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 6
Gene Aliases
ADAM-TS 6, ADAM-TS6, ADAMTS-6
UniGene
Hs.482291
Gene Chromosome Location
Chr.5: 65148736-65482014 on Build GRCh38
Transcripts
NM_197941.3 NR_135689.1 XM_011543113.2 XM_011543114.2 XM_011543115.2 XM_011543116.2 XM_011543117.1 XM_011543121.2 XM_011543123.2 XM_011543124.2 XM_011543125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6594288 c.3026G>A p.R1009H Chr.5: 65172893-65172893 on Build GRCh38
461965 c.2979G>C p.K993N Chr.5: 65172940-65172940 on Build GRCh38
1695826 c.3007G>A p.E1003K Chr.5: 65172912-65172912 on Build GRCh38
1283276 c.3032G>A p.R1011H Chr.5: 65172887-65172887 on Build GRCh38
4659638 c.3006G>T p.E1002D Chr.5: 65172913-65172913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046055403 Chr.5: 65172960-65172960 on Build GRCh38
rs943612435 Chr.5: 65172883-65172883 on Build GRCh38
rs372254854 Chr.5: 65173033-65173033 on Build GRCh38
rs892231228 Chr.5: 65172926-65172926 on Build GRCh38
rs928024958 Chr.5: 65173152-65173152 on Build GRCh38
rs780680737 Chr.5: 65172837-65172837 on Build GRCh38
rs768347461 Chr.5: 65172958-65172958 on Build GRCh38
rs376192625 Chr.5: 65173039-65173039 on Build GRCh38
rs544690009 Chr.5: 65173038-65173038 on Build GRCh38
rs575761382 Chr.5: 65172895-65172895 on Build GRCh38
rs758828270 Chr.5: 65172975-65172975 on Build GRCh38
rs200963830 Chr.5: 65172887-65172887 on Build GRCh38
rs756174305 Chr.5: 65172835-65172835 on Build GRCh38
rs763809434 Chr.5: 65172817-65172817 on Build GRCh38
rs372847337 Chr.5: 65173098-65173100 on Build GRCh38
rs750553528 Chr.5: 65172827-65172827 on Build GRCh38
rs775007475 Chr.5: 65172893-65172893 on Build GRCh38
rs779461384 Chr.5: 65173017-65173017 on Build GRCh38
rs555737820 Chr.5: 65172894-65172894 on Build GRCh38
rs144337843 Chr.5: 65173096-65173096 on Build GRCh38
rs775284460 Chr.5: 65172815-65172815 on Build GRCh38
rs780850470 Chr.5: 65172974-65172974 on Build GRCh38
rs770751710 Chr.5: 65172872-65172872 on Build GRCh38
rs976380836 Chr.5: 65172976-65172976 on Build GRCh38
rs1018992360 Chr.5: 65173120-65173120 on Build GRCh38
rs939360892 Chr.5: 65173158-65173158 on Build GRCh38
rs767786776 Chr.5: 65172916-65172916 on Build GRCh38
rs372898244 Chr.5: 65173090-65173090 on Build GRCh38
rs755742862 Chr.5: 65173015-65173015 on Build GRCh38
rs980856841 Chr.5: 65173145-65173145 on Build GRCh38
rs906451494 Chr.5: 65173018-65173018 on Build GRCh38
rs370452008 Chr.5: 65172863-65172863 on Build GRCh38
rs764347208 Chr.5: 65173020-65173021 on Build GRCh38
rs376868130 Chr.5: 65172841-65172841 on Build GRCh38
rs753909590 Chr.5: 65172937-65172937 on Build GRCh38
rs750861930 Chr.5: 65172802-65172802 on Build GRCh38
rs768524726 Chr.5: 65173026-65173026 on Build GRCh38
rs769190743 Chr.5: 65172888-65172888 on Build GRCh38
rs780976067 Chr.5: 65172873-65172873 on Build GRCh38
rs754133442 Chr.5: 65173053-65173054 on Build GRCh38
rs755458554 Chr.5: 65172847-65172847 on Build GRCh38
rs892436653 Chr.5: 65173223-65173223 on Build GRCh38
rs867479345 Chr.5: 65172912-65172912 on Build GRCh38
rs74464814 Chr.5: 65172857-65172857 on Build GRCh38
rs762494963 Chr.5: 65172790-65172790 on Build GRCh38
rs759091794 Chr.5: 65172798-65172798 on Build GRCh38
rs766386810 Chr.5: 65172936-65172936 on Build GRCh38
rs936141710 Chr.5: 65173215-65173215 on Build GRCh38
rs778214796 Chr.5: 65172980-65172980 on Build GRCh38
rs776317669 Chr.5: 65172795-65172795 on Build GRCh38
rs768143558 Chr.5: 65172820-65172821 on Build GRCh38
rs753253554 Chr.5: 65172858-65172860 on Build GRCh38
rs370621133 Chr.5: 65172923-65172923 on Build GRCh38
rs909982674 Chr.5: 65172813-65172813 on Build GRCh38
rs748801493 Chr.5: 65173025-65173025 on Build GRCh38
rs113712080 Chr.5: 65172812-65172812 on Build GRCh38
rs146459028 Chr.5: 65173006-65173006 on Build GRCh38
rs964958195 Chr.5: 65173151-65173151 on Build GRCh38
rs199992016 Chr.5: 65172824-65172824 on Build GRCh38
rs773421984 Chr.5: 65172925-65172925 on Build GRCh38
rs767696600 Chr.5: 65172825-65172825 on Build GRCh38
rs796324928 Chr.5: 65173052-65173052 on Build GRCh38
rs376199311 Chr.5: 65172821-65172821 on Build GRCh38
rs373608716 Chr.5: 65172886-65172886 on Build GRCh38
rs368288001 Chr.5: 65172930-65172930 on Build GRCh38
rs761575652 Chr.5: 65172901-65172901 on Build GRCh38
rs769825050 Chr.5: 65172811-65172811 on Build GRCh38
rs553801975 Chr.5: 65172848-65172848 on Build GRCh38
rs540776105 Chr.5: 65173198-65173198 on Build GRCh38
rs899023542 Chr.5: 65173065-65173065 on Build GRCh38
rs761162917 Chr.5: 65173017-65173021 on Build GRCh38
rs142540751 Chr.5: 65172997-65172997 on Build GRCh38
rs765748431 Chr.5: 65172966-65172966 on Build GRCh38

View Full Product Details