Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65214661-65215165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS6
Entrez Gene ID
11174
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 6
Gene Aliases
ADAM-TS 6, ADAM-TS6, ADAMTS-6
UniGene
Hs.482291
Gene Chromosome Location
Chr.5: 65148736-65482014 on Build GRCh38
Transcripts
NM_197941.3 NR_135689.1 XM_011543113.2 XM_011543114.2 XM_011543115.2 XM_011543116.2 XM_011543117.1 XM_011543119.2 XM_011543120.2 XM_011543121.2 XM_011543122.2 XM_011543123.2 XM_011543124.2 XM_011543125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596222 c.2462G>A p.G821E Chr.5: 65214907-65214907 on Build GRCh38
5759204 c.2558C>T p.S853L Chr.5: 65214811-65214811 on Build GRCh38
1181941 c.2512A>G p.N838D Chr.5: 65214857-65214857 on Build GRCh38
73578 c.2509G>C p.D837H Chr.5: 65214860-65214860 on Build GRCh38
5403703 c.2454G>A p.Q818Q Chr.5: 65214915-65214915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866435522 Chr.5: 65214744-65214744 on Build GRCh38
rs140414926 Chr.5: 65214857-65214857 on Build GRCh38
rs903484330 Chr.5: 65214851-65214851 on Build GRCh38
rs894559900 Chr.5: 65215032-65215032 on Build GRCh38
rs942014416 Chr.5: 65215070-65215070 on Build GRCh38
rs779929237 Chr.5: 65214711-65214711 on Build GRCh38
rs761214925 Chr.5: 65214840-65214840 on Build GRCh38
rs146678197 Chr.5: 65214992-65214992 on Build GRCh38
rs760985020 Chr.5: 65215100-65215100 on Build GRCh38
rs995492397 Chr.5: 65215055-65215055 on Build GRCh38
rs150401921 Chr.5: 65214879-65214879 on Build GRCh38
rs145149187 Chr.5: 65214911-65214911 on Build GRCh38
rs753648478 Chr.5: 65214874-65214874 on Build GRCh38
rs1019734145 Chr.5: 65215140-65215140 on Build GRCh38
rs755272120 Chr.5: 65214875-65214875 on Build GRCh38
rs945964006 Chr.5: 65214902-65214902 on Build GRCh38
rs746113036 Chr.5: 65214919-65214919 on Build GRCh38
rs762623553 Chr.5: 65214781-65214781 on Build GRCh38
rs768413501 Chr.5: 65214929-65214929 on Build GRCh38
rs937868512 Chr.5: 65215120-65215120 on Build GRCh38
rs544237501 Chr.5: 65214783-65214783 on Build GRCh38
rs140289344 Chr.5: 65215102-65215102 on Build GRCh38
rs564381574 Chr.5: 65215014-65215014 on Build GRCh38
rs746208213 Chr.5: 65214927-65214927 on Build GRCh38
rs771295795 Chr.5: 65214777-65214777 on Build GRCh38
rs777768727 Chr.5: 65214898-65214898 on Build GRCh38
rs776929233 Chr.5: 65214779-65214779 on Build GRCh38
rs747534573 Chr.5: 65214900-65214900 on Build GRCh38
rs1055845300 Chr.5: 65215017-65215017 on Build GRCh38
rs201944968 Chr.5: 65214972-65214972 on Build GRCh38
rs781574260 Chr.5: 65214915-65214915 on Build GRCh38
rs767558611 Chr.5: 65214849-65214849 on Build GRCh38
rs750391383 Chr.5: 65214850-65214850 on Build GRCh38
rs926527721 Chr.5: 65215112-65215112 on Build GRCh38
rs374748786 Chr.5: 65214998-65214998 on Build GRCh38
rs1056695752 Chr.5: 65215122-65215122 on Build GRCh38
rs1009271443 Chr.5: 65214721-65214721 on Build GRCh38
rs771496788 Chr.5: 65214974-65214974 on Build GRCh38
rs896268374 Chr.5: 65215142-65215142 on Build GRCh38
rs149663058 Chr.5: 65214750-65214750 on Build GRCh38
rs369825879 Chr.5: 65214748-65214748 on Build GRCh38
rs773870792 Chr.5: 65214931-65214931 on Build GRCh38
rs755969509 Chr.5: 65214708-65214708 on Build GRCh38
rs973976200 Chr.5: 65215094-65215094 on Build GRCh38
rs983523935 Chr.5: 65215004-65215004 on Build GRCh38
rs1048895515 Chr.5: 65215056-65215056 on Build GRCh38
rs909210051 Chr.5: 65215021-65215021 on Build GRCh38
rs555294248 Chr.5: 65214894-65214894 on Build GRCh38
rs756059375 Chr.5: 65214855-65214855 on Build GRCh38
rs758409206 Chr.5: 65214897-65214897 on Build GRCh38
rs373191563 Chr.5: 65214765-65214765 on Build GRCh38
rs749536053 Chr.5: 65214736-65214736 on Build GRCh38
rs772312553 Chr.5: 65214939-65214939 on Build GRCh38
rs760728280 Chr.5: 65214982-65214982 on Build GRCh38
rs773584382 Chr.5: 65214755-65214755 on Build GRCh38
rs1021031734 Chr.5: 65214846-65214846 on Build GRCh38
rs553546684 Chr.5: 65214749-65214749 on Build GRCh38
rs773959312 Chr.5: 65214800-65214800 on Build GRCh38
rs772802081 Chr.5: 65214980-65214980 on Build GRCh38
rs763622443 Chr.5: 65214797-65214797 on Build GRCh38
rs755109707 Chr.5: 65214739-65214739 on Build GRCh38
rs750512680 Chr.5: 65214767-65214767 on Build GRCh38
rs1004451467 Chr.5: 65215128-65215128 on Build GRCh38
rs928481113 Chr.5: 65214976-65214976 on Build GRCh38

View Full Product Details