Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73499049-73499543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTF3
Entrez Gene ID
689
Gene Name
basic transcription factor 3
Gene Aliases
BETA-NAC, BTF3a, BTF3b, NACB
UniGene
Hs.591768
Gene Chromosome Location
Chr.5: 73498425-73505623 on Build GRCh38
Transcripts
NM_001037637.1 NM_001207.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5432208 c.179C>A p.A60E Chr.5: 73499180-73499180 on Build GRCh38
6370670 c.141A>T p.E47D Chr.5: 73499142-73499142 on Build GRCh38
3617792 c.139G>C p.E47Q Chr.5: 73499140-73499140 on Build GRCh38
6865417 c.154C>G p.Q52E Chr.5: 73499155-73499155 on Build GRCh38
6339376 c.165C>T p.L55L Chr.5: 73499166-73499166 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752042689 Chr.5: 73499157-73499157 on Build GRCh38
rs577889923 Chr.5: 73499095-73499095 on Build GRCh38
rs770943776 Chr.5: 73499257-73499257 on Build GRCh38
rs376932769 Chr.5: 73499406-73499406 on Build GRCh38
rs746744604 Chr.5: 73499305-73499305 on Build GRCh38
rs565095680 Chr.5: 73499335-73499335 on Build GRCh38
rs368525613 Chr.5: 73499382-73499382 on Build GRCh38
rs73103660 Chr.5: 73499264-73499264 on Build GRCh38
rs764646227 Chr.5: 73499154-73499154 on Build GRCh38
rs893703782 Chr.5: 73499486-73499486 on Build GRCh38
rs760847457 Chr.5: 73499348-73499348 on Build GRCh38
rs531902391 Chr.5: 73499436-73499437 on Build GRCh38
rs775965358 Chr.5: 73499102-73499102 on Build GRCh38
rs773267383 Chr.5: 73499087-73499087 on Build GRCh38
rs755265592 Chr.5: 73499367-73499367 on Build GRCh38
rs760309169 Chr.5: 73499088-73499088 on Build GRCh38
rs751870387 Chr.5: 73499276-73499276 on Build GRCh38
rs763359106 Chr.5: 73499269-73499269 on Build GRCh38
rs528584760 Chr.5: 73499291-73499291 on Build GRCh38
rs988571794 Chr.5: 73499108-73499108 on Build GRCh38
rs761296238 Chr.5: 73499333-73499337 on Build GRCh38
rs758908483 Chr.5: 73499259-73499259 on Build GRCh38
rs989586721 Chr.5: 73499295-73499295 on Build GRCh38
rs765395512 Chr.5: 73499302-73499302 on Build GRCh38
rs754080973 Chr.5: 73499290-73499290 on Build GRCh38
rs957843817 Chr.5: 73499282-73499282 on Build GRCh38
rs769641874 Chr.5: 73499231-73499231 on Build GRCh38
rs397738621 Chr.5: 73499240-73499241 on Build GRCh38
rs560519985 Chr.5: 73499227-73499228 on Build GRCh38
rs780991179 Chr.5: 73499311-73499311 on Build GRCh38
rs368199965 Chr.5: 73499450-73499451 on Build GRCh38
rs183222687 Chr.5: 73499519-73499519 on Build GRCh38
rs750408507 Chr.5: 73499180-73499180 on Build GRCh38
rs150691010 Chr.5: 73499460-73499460 on Build GRCh38
rs750871127 Chr.5: 73499340-73499340 on Build GRCh38
rs754154679 Chr.5: 73499335-73499336 on Build GRCh38
rs926939545 Chr.5: 73499194-73499194 on Build GRCh38
rs765489412 Chr.5: 73499370-73499370 on Build GRCh38
rs770634932 Chr.5: 73499264-73499265 on Build GRCh38
rs980252379 Chr.5: 73499503-73499503 on Build GRCh38
rs568759234 Chr.5: 73499378-73499378 on Build GRCh38
rs771967925 Chr.5: 73499225-73499225 on Build GRCh38
rs758761066 Chr.5: 73499136-73499136 on Build GRCh38
rs1050526953 Chr.5: 73499404-73499404 on Build GRCh38
rs764739115 Chr.5: 73499105-73499105 on Build GRCh38
rs375912895 Chr.5: 73499124-73499124 on Build GRCh38
rs999473685 Chr.5: 73499473-73499476 on Build GRCh38
rs745445777 Chr.5: 73499324-73499324 on Build GRCh38
rs942404854 Chr.5: 73499455-73499455 on Build GRCh38
rs756363331 Chr.5: 73499202-73499202 on Build GRCh38
rs930833084 Chr.5: 73499399-73499399 on Build GRCh38
rs762160587 Chr.5: 73499334-73499334 on Build GRCh38
rs2115439 Chr.5: 73499351-73499351 on Build GRCh38
rs569102851 Chr.5: 73499391-73499391 on Build GRCh38
rs753113625 Chr.5: 73499121-73499121 on Build GRCh38
rs3057009 Chr.5: 73499336-73499340 on Build GRCh38
rs750582934 Chr.5: 73499280-73499280 on Build GRCh38
rs3057007 Chr.5: 73499447-73499448 on Build GRCh38
rs561385707 Chr.5: 73499270-73499270 on Build GRCh38
rs773705326 Chr.5: 73499278-73499280 on Build GRCh38
rs529631804 Chr.5: 73499104-73499104 on Build GRCh38
rs191359554 Chr.5: 73499372-73499372 on Build GRCh38
rs770906375 Chr.5: 73499308-73499308 on Build GRCh38
rs774698723 Chr.5: 73499331-73499331 on Build GRCh38
rs889553507 Chr.5: 73499441-73499441 on Build GRCh38
rs931097057 Chr.5: 73499129-73499129 on Build GRCh38
rs780371955 Chr.5: 73499288-73499288 on Build GRCh38
rs112073100 Chr.5: 73499325-73499325 on Build GRCh38
rs772202683 Chr.5: 73499086-73499086 on Build GRCh38
rs900827490 Chr.5: 73499465-73499466 on Build GRCh38
rs756993051 Chr.5: 73499385-73499385 on Build GRCh38
rs757719063 Chr.5: 73499170-73499170 on Build GRCh38
rs756133112 Chr.5: 73499284-73499284 on Build GRCh38
rs972280045 Chr.5: 73499368-73499369 on Build GRCh38
rs76097803 Chr.5: 73499360-73499360 on Build GRCh38
rs1030903497 Chr.5: 73499484-73499484 on Build GRCh38
rs538022249 Chr.5: 73499436-73499436 on Build GRCh38
rs768237035 Chr.5: 73499329-73499329 on Build GRCh38
rs746975751 Chr.5: 73499241-73499241 on Build GRCh38
rs753922771 Chr.5: 73499356-73499356 on Build GRCh38
rs774995808 Chr.5: 73499267-73499267 on Build GRCh38
rs949816934 Chr.5: 73499167-73499167 on Build GRCh38
rs2115440 Chr.5: 73499224-73499224 on Build GRCh38
rs764470553 Chr.5: 73499271-73499271 on Build GRCh38
rs777899021 Chr.5: 73499231-73499231 on Build GRCh38
rs941070360 Chr.5: 73499219-73499219 on Build GRCh38
rs955961174 Chr.5: 73499072-73499072 on Build GRCh38
rs35216563 Chr.5: 73499230-73499231 on Build GRCh38
rs56029107 Chr.5: 73499178-73499178 on Build GRCh38
rs13158968 Chr.5: 73499490-73499490 on Build GRCh38
rs556919645 Chr.5: 73499211-73499211 on Build GRCh38
rs368111625 Chr.5: 73499083-73499083 on Build GRCh38

View Full Product Details