Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73502785-73503288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTF3
Entrez Gene ID
689
Gene Name
basic transcription factor 3
Gene Aliases
BETA-NAC, BTF3a, BTF3b, NACB
UniGene
Hs.591768
Gene Chromosome Location
Chr.5: 73498425-73505623 on Build GRCh38
Transcripts
NM_001037637.1 NM_001207.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069942 c.400A>G p.I134V Chr.5: 73503000-73503000 on Build GRCh38
3828392 c.473G>C p.S158T Chr.5: 73503073-73503073 on Build GRCh38
318960 c.508C>T p.P170S Chr.5: 73503108-73503108 on Build GRCh38
3828391 c.415G>C p.E139Q Chr.5: 73503015-73503015 on Build GRCh38
5447649 c.436A>G p.M146V Chr.5: 73503036-73503036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6862039 Chr.5: 73503170-73503170 on Build GRCh38
rs778415639 Chr.5: 73502931-73502931 on Build GRCh38
rs186931485 Chr.5: 73503099-73503099 on Build GRCh38
rs746411488 Chr.5: 73502997-73502997 on Build GRCh38
rs568426738 Chr.5: 73503148-73503148 on Build GRCh38
rs140372053 Chr.5: 73503179-73503179 on Build GRCh38
rs923457323 Chr.5: 73503184-73503184 on Build GRCh38
rs951669972 Chr.5: 73502993-73502993 on Build GRCh38
rs768814807 Chr.5: 73503032-73503032 on Build GRCh38
rs111757484 Chr.5: 73502954-73502954 on Build GRCh38
rs763094367 Chr.5: 73502886-73502886 on Build GRCh38
rs765378612 Chr.5: 73502866-73502866 on Build GRCh38
rs151143923 Chr.5: 73502951-73502951 on Build GRCh38
rs752195686 Chr.5: 73502936-73502936 on Build GRCh38
rs1050429962 Chr.5: 73502937-73502937 on Build GRCh38
rs372460087 Chr.5: 73503067-73503067 on Build GRCh38
rs747392162 Chr.5: 73502944-73502944 on Build GRCh38
rs878981435 Chr.5: 73502962-73502962 on Build GRCh38
rs766351583 Chr.5: 73502908-73502908 on Build GRCh38
rs757993865 Chr.5: 73502940-73502940 on Build GRCh38
rs557675678 Chr.5: 73503261-73503261 on Build GRCh38
rs1055598002 Chr.5: 73503249-73503249 on Build GRCh38
rs182373192 Chr.5: 73502809-73502809 on Build GRCh38
rs575670734 Chr.5: 73502842-73502842 on Build GRCh38
rs758587066 Chr.5: 73503116-73503117 on Build GRCh38
rs753579016 Chr.5: 73503098-73503098 on Build GRCh38
rs201362870 Chr.5: 73503008-73503008 on Build GRCh38
rs572647489 Chr.5: 73503230-73503230 on Build GRCh38
rs116176540 Chr.5: 73502884-73502884 on Build GRCh38
rs347233 Chr.5: 73502899-73502899 on Build GRCh38
rs143150590 Chr.5: 73503048-73503048 on Build GRCh38
rs966602011 Chr.5: 73502965-73502965 on Build GRCh38
rs754868948 Chr.5: 73502920-73502920 on Build GRCh38
rs780992997 Chr.5: 73503131-73503133 on Build GRCh38
rs372028278 Chr.5: 73502862-73502862 on Build GRCh38
rs904414272 Chr.5: 73503234-73503234 on Build GRCh38
rs150380551 Chr.5: 73503248-73503248 on Build GRCh38
rs368135071 Chr.5: 73503157-73503157 on Build GRCh38
rs550156544 Chr.5: 73503110-73503110 on Build GRCh38
rs1010156541 Chr.5: 73502821-73502821 on Build GRCh38
rs760502541 Chr.5: 73503077-73503077 on Build GRCh38
rs764307835 Chr.5: 73502898-73502898 on Build GRCh38
rs770379410 Chr.5: 73503002-73503002 on Build GRCh38
rs543410706 Chr.5: 73502855-73502855 on Build GRCh38
rs41271745 Chr.5: 73503018-73503018 on Build GRCh38
rs755041505 Chr.5: 73503124-73503136 on Build GRCh38
rs547165276 Chr.5: 73503171-73503171 on Build GRCh38
rs940827826 Chr.5: 73502880-73502880 on Build GRCh38
rs751197960 Chr.5: 73503169-73503169 on Build GRCh38
rs777342643 Chr.5: 73503165-73503165 on Build GRCh38
rs958788578 Chr.5: 73502911-73502911 on Build GRCh38
rs996682498 Chr.5: 73502922-73502922 on Build GRCh38
rs72768738 Chr.5: 73503068-73503068 on Build GRCh38
rs753743654 Chr.5: 73502912-73502912 on Build GRCh38
rs1050504145 Chr.5: 73503194-73503194 on Build GRCh38
rs774048914 Chr.5: 73503193-73503193 on Build GRCh38
rs766528717 Chr.5: 73503120-73503121 on Build GRCh38
rs137911027 Chr.5: 73502987-73502987 on Build GRCh38
rs14010 Chr.5: 73503020-73503020 on Build GRCh38
rs201235182 Chr.5: 73503049-73503049 on Build GRCh38
rs561590492 Chr.5: 73502860-73502860 on Build GRCh38
rs931683221 Chr.5: 73503144-73503144 on Build GRCh38
rs766190108 Chr.5: 73503081-73503081 on Build GRCh38

View Full Product Details