Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73504210-73504755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTF3
Entrez Gene ID
689
Gene Name
basic transcription factor 3
Gene Aliases
BETA-NAC, BTF3a, BTF3b, NACB
UniGene
Hs.591768
Gene Chromosome Location
Chr.5: 73498425-73505623 on Build GRCh38
Transcripts
NM_001037637.1 NM_001207.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6323887 c.548G>A p.G183E Chr.5: 73504377-73504377 on Build GRCh38
1069943 c.557A>G p.D186G Chr.5: 73504386-73504386 on Build GRCh38
3617794 c.572C>T p.P191L Chr.5: 73504401-73504401 on Build GRCh38
3737378 c.574+8C>T p.? Chr.5: 73504411-73504411 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756028701 Chr.5: 73504381-73504382 on Build GRCh38
rs763692757 Chr.5: 73504403-73504403 on Build GRCh38
rs530706564 Chr.5: 73504707-73504707 on Build GRCh38
rs756986460 Chr.5: 73504412-73504412 on Build GRCh38
rs762043512 Chr.5: 73504338-73504338 on Build GRCh38
rs759324851 Chr.5: 73504364-73504364 on Build GRCh38
rs56381033 Chr.5: 73504585-73504585 on Build GRCh38
rs1033120011 Chr.5: 73504332-73504332 on Build GRCh38
rs879171657 Chr.5: 73504455-73504455 on Build GRCh38
rs1012587235 Chr.5: 73504681-73504681 on Build GRCh38
rs564526972 Chr.5: 73504268-73504278 on Build GRCh38
rs748991364 Chr.5: 73504449-73504454 on Build GRCh38
rs767270255 Chr.5: 73504417-73504417 on Build GRCh38
rs761837491 Chr.5: 73504267-73504268 on Build GRCh38
rs1022585610 Chr.5: 73504696-73504696 on Build GRCh38
rs750871538 Chr.5: 73504420-73504420 on Build GRCh38
rs751971137 Chr.5: 73504687-73504687 on Build GRCh38
rs919121882 Chr.5: 73504618-73504618 on Build GRCh38
rs992954803 Chr.5: 73504494-73504494 on Build GRCh38
rs55910008 Chr.5: 73504472-73504472 on Build GRCh38
rs368717420 Chr.5: 73504447-73504447 on Build GRCh38
rs375717813 Chr.5: 73504331-73504331 on Build GRCh38
rs746165710 Chr.5: 73504307-73504307 on Build GRCh38
rs529333109 Chr.5: 73504540-73504540 on Build GRCh38
rs933054913 Chr.5: 73504626-73504626 on Build GRCh38
rs780592999 Chr.5: 73504311-73504311 on Build GRCh38
rs777708889 Chr.5: 73504405-73504406 on Build GRCh38
rs10068771 Chr.5: 73504295-73504295 on Build GRCh38
rs1042714638 Chr.5: 73504721-73504721 on Build GRCh38
rs559388716 Chr.5: 73504587-73504587 on Build GRCh38
rs762412758 Chr.5: 73504396-73504396 on Build GRCh38
rs776929230 Chr.5: 73504597-73504597 on Build GRCh38
rs202126534 Chr.5: 73504453-73504453 on Build GRCh38
rs527607966 Chr.5: 73504557-73504557 on Build GRCh38
rs1036581207 Chr.5: 73504613-73504613 on Build GRCh38
rs369443632 Chr.5: 73504657-73504657 on Build GRCh38
rs748057618 Chr.5: 73504382-73504384 on Build GRCh38
rs368386404 Chr.5: 73504437-73504437 on Build GRCh38
rs369157570 Chr.5: 73504247-73504247 on Build GRCh38
rs569869806 Chr.5: 73504694-73504694 on Build GRCh38
rs760319904 Chr.5: 73504384-73504384 on Build GRCh38
rs773022738 Chr.5: 73504367-73504367 on Build GRCh38
rs750989476 Chr.5: 73504411-73504411 on Build GRCh38
rs781709809 Chr.5: 73504304-73504304 on Build GRCh38
rs201496691 Chr.5: 73504267-73504267 on Build GRCh38
rs12332640 Chr.5: 73504361-73504361 on Build GRCh38
rs12656416 Chr.5: 73504470-73504470 on Build GRCh38
rs703882 Chr.5: 73504583-73504583 on Build GRCh38
rs997294832 Chr.5: 73504281-73504281 on Build GRCh38
rs957314942 Chr.5: 73504664-73504664 on Build GRCh38
rs768749052 Chr.5: 73504329-73504329 on Build GRCh38
rs2703643 Chr.5: 73504350-73504350 on Build GRCh38
rs200903571 Chr.5: 73504363-73504363 on Build GRCh38
rs932680485 Chr.5: 73504480-73504480 on Build GRCh38
rs778077997 Chr.5: 73504614-73504614 on Build GRCh38
rs765445157 Chr.5: 73504546-73504546 on Build GRCh38
rs879107660 Chr.5: 73504268-73504270 on Build GRCh38
rs58721358 Chr.5: 73504292-73504292 on Build GRCh38
rs756933034 Chr.5: 73504300-73504300 on Build GRCh38
rs1051558023 Chr.5: 73504665-73504665 on Build GRCh38
rs908376875 Chr.5: 73504441-73504441 on Build GRCh38
rs749373640 Chr.5: 73504318-73504318 on Build GRCh38
rs1008480471 Chr.5: 73504549-73504549 on Build GRCh38
rs115669743 Chr.5: 73504432-73504432 on Build GRCh38
rs991534070 Chr.5: 73504386-73504386 on Build GRCh38
rs998606801 Chr.5: 73504247-73504249 on Build GRCh38
rs869200126 Chr.5: 73504292-73504293 on Build GRCh38
rs34051700 Chr.5: 73504268-73504268 on Build GRCh38
rs765146287 Chr.5: 73504373-73504373 on Build GRCh38
rs943001059 Chr.5: 73504713-73504713 on Build GRCh38
rs779062571 Chr.5: 73504449-73504449 on Build GRCh38
rs915890331 Chr.5: 73504523-73504523 on Build GRCh38
rs148302315 Chr.5: 73504492-73504492 on Build GRCh38
rs879233683 Chr.5: 73504268-73504279 on Build GRCh38
rs533274546 Chr.5: 73504612-73504612 on Build GRCh38
rs551141724 Chr.5: 73504653-73504653 on Build GRCh38
rs772223535 Chr.5: 73504349-73504349 on Build GRCh38
rs763308410 Chr.5: 73504275-73504276 on Build GRCh38

View Full Product Details