Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74738375-74738908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM2
Entrez Gene ID
84340
Gene Name
G elongation factor mitochondrial 2
Gene Aliases
EF-G2mt, EFG2, MRRF2, MST027, MSTP027, RRF2, RRF2mt, hEFG2, mEF-G 2
UniGene
Hs.277154
Gene Chromosome Location
Chr.5: 74721204-74767371 on Build GRCh38
Transcripts
NM_001281302.1 NM_032380.4 NM_170681.2 NM_170691.2 NR_104006.1 XM_011543691.2 XM_017009986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3856278 c.1159C>T p.R387C Chr.5: 74738563-74738563 on Build GRCh38
5174522 c.1155delT p.F385fs*9 Chr.5: 74738567-74738567 on Build GRCh38
6353673 c.1196T>C p.I399T Chr.5: 74738526-74738526 on Build GRCh38
3073216 c.1127A>G p.H376R Chr.5: 74738595-74738595 on Build GRCh38
5652030 c.1213A>C p.N405H Chr.5: 74738509-74738509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020096131 Chr.5: 74738735-74738735 on Build GRCh38
rs778605094 Chr.5: 74738683-74738683 on Build GRCh38
rs78674702 Chr.5: 74738462-74738462 on Build GRCh38
rs191007875 Chr.5: 74738602-74738602 on Build GRCh38
rs759625706 Chr.5: 74738502-74738502 on Build GRCh38
rs747760119 Chr.5: 74738486-74738486 on Build GRCh38
rs772506292 Chr.5: 74738489-74738489 on Build GRCh38
rs765235690 Chr.5: 74738507-74738507 on Build GRCh38
rs897046003 Chr.5: 74738412-74738412 on Build GRCh38
rs928364160 Chr.5: 74738669-74738669 on Build GRCh38
rs561167614 Chr.5: 74738670-74738670 on Build GRCh38
rs752948580 Chr.5: 74738514-74738514 on Build GRCh38
rs779755199 Chr.5: 74738457-74738457 on Build GRCh38
rs776748857 Chr.5: 74738501-74738501 on Build GRCh38
rs779411230 Chr.5: 74738856-74738856 on Build GRCh38
rs866401948 Chr.5: 74738563-74738563 on Build GRCh38
rs780130081 Chr.5: 74738535-74738535 on Build GRCh38
rs112155465 Chr.5: 74738634-74738634 on Build GRCh38
rs1029102576 Chr.5: 74738662-74738662 on Build GRCh38
rs887883100 Chr.5: 74738463-74738463 on Build GRCh38
rs762693230 Chr.5: 74738661-74738661 on Build GRCh38
rs747881218 Chr.5: 74738583-74738583 on Build GRCh38
rs532616672 Chr.5: 74738778-74738778 on Build GRCh38
rs201204413 Chr.5: 74738493-74738493 on Build GRCh38
rs116582430 Chr.5: 74738750-74738750 on Build GRCh38
rs901906200 Chr.5: 74738817-74738817 on Build GRCh38
rs142425196 Chr.5: 74738404-74738404 on Build GRCh38
rs766791577 Chr.5: 74738495-74738495 on Build GRCh38
rs863224037 Chr.5: 74738409-74738409 on Build GRCh38
rs755985449 Chr.5: 74738442-74738442 on Build GRCh38
rs540786091 Chr.5: 74738743-74738743 on Build GRCh38
rs760387518 Chr.5: 74738666-74738666 on Build GRCh38
rs571673017 Chr.5: 74738403-74738403 on Build GRCh38
rs765432280 Chr.5: 74738398-74738398 on Build GRCh38
rs7732843 Chr.5: 74738420-74738420 on Build GRCh38
rs141886148 Chr.5: 74738620-74738620 on Build GRCh38
rs550738633 Chr.5: 74738810-74738810 on Build GRCh38
rs981331880 Chr.5: 74738763-74738763 on Build GRCh38
rs551952385 Chr.5: 74738804-74738804 on Build GRCh38
rs953178248 Chr.5: 74738693-74738693 on Build GRCh38
rs147925896 Chr.5: 74738534-74738534 on Build GRCh38
rs931197445 Chr.5: 74738746-74738746 on Build GRCh38
rs763038574 Chr.5: 74738523-74738523 on Build GRCh38
rs561517778 Chr.5: 74738680-74738680 on Build GRCh38
rs533208093 Chr.5: 74738761-74738763 on Build GRCh38
rs1036836926 Chr.5: 74738705-74738705 on Build GRCh38
rs752142132 Chr.5: 74738684-74738684 on Build GRCh38
rs778863487 Chr.5: 74738587-74738587 on Build GRCh38
rs773425115 Chr.5: 74738492-74738492 on Build GRCh38
rs747322521 Chr.5: 74738592-74738592 on Build GRCh38
rs747424095 Chr.5: 74738762-74738762 on Build GRCh38
rs780509666 Chr.5: 74738757-74738757 on Build GRCh38
rs759997637 Chr.5: 74738611-74738611 on Build GRCh38
rs769887272 Chr.5: 74738619-74738619 on Build GRCh38
rs374630200 Chr.5: 74738559-74738559 on Build GRCh38
rs1017803669 Chr.5: 74738482-74738482 on Build GRCh38
rs753758523 Chr.5: 74738557-74738557 on Build GRCh38
rs375996126 Chr.5: 74738464-74738464 on Build GRCh38
rs371753740 Chr.5: 74738764-74738766 on Build GRCh38
rs573484564 Chr.5: 74738702-74738702 on Build GRCh38
rs538761639 Chr.5: 74738663-74738663 on Build GRCh38
rs773010798 Chr.5: 74738499-74738500 on Build GRCh38
rs118133558 Chr.5: 74738628-74738628 on Build GRCh38
rs749758056 Chr.5: 74738614-74738614 on Build GRCh38
rs772191981 Chr.5: 74738433-74738433 on Build GRCh38
rs991542337 Chr.5: 74738816-74738816 on Build GRCh38
rs771592465 Chr.5: 74738584-74738584 on Build GRCh38
rs754563414 Chr.5: 74738565-74738565 on Build GRCh38
rs1027641865 Chr.5: 74738427-74738427 on Build GRCh38
rs866244233 Chr.5: 74738610-74738610 on Build GRCh38
rs200232238 Chr.5: 74738413-74738413 on Build GRCh38
rs981113569 Chr.5: 74738448-74738448 on Build GRCh38
rs143233751 Chr.5: 74738526-74738526 on Build GRCh38
rs931054729 Chr.5: 74738875-74738875 on Build GRCh38
rs777129105 Chr.5: 74738608-74738608 on Build GRCh38
rs182115360 Chr.5: 74738667-74738667 on Build GRCh38
rs186213016 Chr.5: 74738465-74738465 on Build GRCh38
rs531729405 Chr.5: 74738671-74738671 on Build GRCh38
rs958427363 Chr.5: 74738797-74738797 on Build GRCh38
rs368410845 Chr.5: 74738714-74738714 on Build GRCh38
rs199681606 Chr.5: 74738575-74738575 on Build GRCh38
rs536132088 Chr.5: 74738430-74738430 on Build GRCh38
rs998108555 Chr.5: 74738819-74738819 on Build GRCh38

View Full Product Details