Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81335629-81336137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT12
Entrez Gene ID
134526
Gene Name
acyl-CoA thioesterase 12
Gene Aliases
CACH-1, Cach, STARD15, THEAL
UniGene
Hs.591756
Gene Chromosome Location
Chr.5: 81309045-81394174 on Build GRCh38
Transcripts
NM_130767.2 XM_006714531.2 XM_006714532.3 XM_017009045.1 XM_017009046.1 XM_017009047.1 XM_017009048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6104657 c.1172G>T p.W391L Chr.5: 81335858-81335858 on Build GRCh38
5026361 c.1129-1G>C p.? Chr.5: 81335902-81335902 on Build GRCh38
7353976 c.1173G>T p.W391C Chr.5: 81335857-81335857 on Build GRCh38
4425333 c.1213C>A p.R405S Chr.5: 81335817-81335817 on Build GRCh38
6470546 c.1129-1G>A p.? Chr.5: 81335902-81335902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902145769 Chr.5: 81336078-81336078 on Build GRCh38
rs78872643 Chr.5: 81335962-81335962 on Build GRCh38
rs563003435 Chr.5: 81336061-81336061 on Build GRCh38
rs758382100 Chr.5: 81335903-81335903 on Build GRCh38
rs985244098 Chr.5: 81335940-81335940 on Build GRCh38
rs756221986 Chr.5: 81335754-81335754 on Build GRCh38
rs576051111 Chr.5: 81335915-81335915 on Build GRCh38
rs938282510 Chr.5: 81336010-81336010 on Build GRCh38
rs769064260 Chr.5: 81335945-81335945 on Build GRCh38
rs139985604 Chr.5: 81335656-81335656 on Build GRCh38
rs973521852 Chr.5: 81335966-81335966 on Build GRCh38
rs564306951 Chr.5: 81335817-81335817 on Build GRCh38
rs750592941 Chr.5: 81335873-81335873 on Build GRCh38
rs1003276589 Chr.5: 81335922-81335922 on Build GRCh38
rs770470580 Chr.5: 81335815-81335815 on Build GRCh38
rs116174844 Chr.5: 81336101-81336101 on Build GRCh38
rs769500159 Chr.5: 81336063-81336063 on Build GRCh38
rs552739984 Chr.5: 81335694-81335694 on Build GRCh38
rs778959500 Chr.5: 81335727-81335727 on Build GRCh38
rs764480644 Chr.5: 81335844-81335844 on Build GRCh38
rs976799516 Chr.5: 81336005-81336005 on Build GRCh38
rs369360280 Chr.5: 81335722-81335722 on Build GRCh38
rs776691407 Chr.5: 81335658-81335658 on Build GRCh38
rs756939603 Chr.5: 81335930-81335930 on Build GRCh38
rs756044212 Chr.5: 81335781-81335781 on Build GRCh38
rs781379551 Chr.5: 81335738-81335738 on Build GRCh38
rs201217225 Chr.5: 81335747-81335747 on Build GRCh38
rs149756748 Chr.5: 81336097-81336097 on Build GRCh38
rs375160700 Chr.5: 81335939-81335939 on Build GRCh38
rs560752102 Chr.5: 81336004-81336004 on Build GRCh38
rs147623252 Chr.5: 81335793-81335793 on Build GRCh38
rs552801317 Chr.5: 81336060-81336060 on Build GRCh38
rs113587855 Chr.5: 81335728-81335728 on Build GRCh38
rs780717291 Chr.5: 81335919-81335919 on Build GRCh38
rs906049540 Chr.5: 81335683-81335683 on Build GRCh38
rs1035848381 Chr.5: 81335685-81335685 on Build GRCh38
rs60755777 Chr.5: 81336058-81336058 on Build GRCh38
rs956709087 Chr.5: 81336034-81336034 on Build GRCh38
rs753650852 Chr.5: 81335918-81335922 on Build GRCh38
rs1012011775 Chr.5: 81335670-81335670 on Build GRCh38
rs141642570 Chr.5: 81335845-81335845 on Build GRCh38
rs1023352455 Chr.5: 81335682-81335682 on Build GRCh38
rs762188607 Chr.5: 81335869-81335869 on Build GRCh38
rs867504639 Chr.5: 81336036-81336038 on Build GRCh38
rs1027021394 Chr.5: 81335704-81335704 on Build GRCh38
rs377639300 Chr.5: 81335827-81335827 on Build GRCh38
rs189598373 Chr.5: 81335709-81335709 on Build GRCh38
rs36123812 Chr.5: 81336037-81336038 on Build GRCh38
rs376835484 Chr.5: 81335921-81335921 on Build GRCh38
rs10371 Chr.5: 81335823-81335823 on Build GRCh38
rs74794658 Chr.5: 81335700-81335700 on Build GRCh38
rs923806450 Chr.5: 81336007-81336007 on Build GRCh38
rs923749852 Chr.5: 81335781-81335781 on Build GRCh38
rs1042156100 Chr.5: 81336072-81336072 on Build GRCh38
rs180672704 Chr.5: 81335930-81335930 on Build GRCh38
rs372394141 Chr.5: 81335796-81335796 on Build GRCh38
rs756276958 Chr.5: 81335877-81335877 on Build GRCh38
rs762089378 Chr.5: 81335952-81335952 on Build GRCh38
rs745418107 Chr.5: 81335818-81335818 on Build GRCh38
rs6452399 Chr.5: 81335692-81335692 on Build GRCh38
rs752750653 Chr.5: 81335892-81335892 on Build GRCh38
rs112980342 Chr.5: 81335987-81335987 on Build GRCh38
rs745503512 Chr.5: 81335920-81335920 on Build GRCh38
rs752044430 Chr.5: 81335975-81335975 on Build GRCh38
rs1029549605 Chr.5: 81335938-81335938 on Build GRCh38
rs185248841 Chr.5: 81335950-81335950 on Build GRCh38
rs372983246 Chr.5: 81335724-81335724 on Build GRCh38
rs766555642 Chr.5: 81335890-81335890 on Build GRCh38
rs371123929 Chr.5: 81335776-81335776 on Build GRCh38
rs369520197 Chr.5: 81335929-81335929 on Build GRCh38
rs772051643 Chr.5: 81335792-81335792 on Build GRCh38
rs935299430 Chr.5: 81335791-81335791 on Build GRCh38
rs767343020 Chr.5: 81335684-81335684 on Build GRCh38
rs943885117 Chr.5: 81335766-81335766 on Build GRCh38
rs569244692 Chr.5: 81335695-81335695 on Build GRCh38
rs190848050 Chr.5: 81335980-81335980 on Build GRCh38
rs767663717 Chr.5: 81335871-81335871 on Build GRCh38
rs200014577 Chr.5: 81336037-81336037 on Build GRCh38
rs746716877 Chr.5: 81335808-81335808 on Build GRCh38
rs748042477 Chr.5: 81335789-81335789 on Build GRCh38
rs531671751 Chr.5: 81336096-81336096 on Build GRCh38
rs929341470 Chr.5: 81335983-81335983 on Build GRCh38
rs750652372 Chr.5: 81335823-81335823 on Build GRCh38
rs1006796261 Chr.5: 81336109-81336109 on Build GRCh38
rs746805766 Chr.5: 81335908-81335908 on Build GRCh38
rs1055291673 Chr.5: 81336019-81336019 on Build GRCh38
rs374660014 Chr.5: 81335826-81335826 on Build GRCh38
rs779823691 Chr.5: 81336036-81336037 on Build GRCh38
rs576654714 Chr.5: 81335816-81335816 on Build GRCh38
rs763839868 Chr.5: 81335798-81335798 on Build GRCh38

View Full Product Details