Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 82058417-82058931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG10
Entrez Gene ID
83734
Gene Name
autophagy related 10
Gene Aliases
APG10, APG10L, pp12616
UniGene
Hs.713698
Gene Chromosome Location
Chr.5: 81972021-82258502 on Build GRCh38
Transcripts
NM_001131028.1 NM_031482.4 XM_005248610.4 XM_005248611.4 XM_005248612.3 XM_011543660.2 XM_017009944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4476976 c.210C>T p.P70P Chr.5: 82058596-82058596 on Build GRCh38
6210546 c.109-3C>T p.? Chr.5: 82058492-82058492 on Build GRCh38
1132099 c.170C>G p.S57* Chr.5: 82058556-82058556 on Build GRCh38
1634348 c.140A>G p.H47R Chr.5: 82058526-82058526 on Build GRCh38
5646872 c.109G>T p.D37Y Chr.5: 82058495-82058495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780032864 Chr.5: 82058486-82058486 on Build GRCh38
rs965524652 Chr.5: 82058814-82058814 on Build GRCh38
rs763660766 Chr.5: 82058608-82058609 on Build GRCh38
rs770135272 Chr.5: 82058550-82058550 on Build GRCh38
rs1029007975 Chr.5: 82058827-82058827 on Build GRCh38
rs147821761 Chr.5: 82058592-82058592 on Build GRCh38
rs752949356 Chr.5: 82058470-82058470 on Build GRCh38
rs753799304 Chr.5: 82058489-82058489 on Build GRCh38
rs879807917 Chr.5: 82058648-82058648 on Build GRCh38
rs760287013 Chr.5: 82058604-82058605 on Build GRCh38
rs777639513 Chr.5: 82058623-82058623 on Build GRCh38
rs758954782 Chr.5: 82058610-82058611 on Build GRCh38
rs756894714 Chr.5: 82058691-82058691 on Build GRCh38
rs1031322834 Chr.5: 82058708-82058708 on Build GRCh38
rs568426556 Chr.5: 82058717-82058717 on Build GRCh38
rs777066432 Chr.5: 82058533-82058533 on Build GRCh38
rs747855524 Chr.5: 82058506-82058506 on Build GRCh38
rs1008597203 Chr.5: 82058776-82058776 on Build GRCh38
rs909633568 Chr.5: 82058675-82058675 on Build GRCh38
rs573579148 Chr.5: 82058898-82058898 on Build GRCh38
rs761937487 Chr.5: 82058587-82058587 on Build GRCh38
rs751866062 Chr.5: 82058624-82058625 on Build GRCh38
rs975566480 Chr.5: 82058817-82058817 on Build GRCh38
rs767646913 Chr.5: 82058590-82058590 on Build GRCh38
rs141370493 Chr.5: 82058614-82058614 on Build GRCh38
rs149472606 Chr.5: 82058567-82058567 on Build GRCh38
rs754812511 Chr.5: 82058494-82058494 on Build GRCh38
rs3734114 Chr.5: 82058570-82058570 on Build GRCh38
rs78103836 Chr.5: 82058819-82058819 on Build GRCh38
rs921490586 Chr.5: 82058495-82058495 on Build GRCh38
rs759888724 Chr.5: 82058457-82058457 on Build GRCh38
rs553341171 Chr.5: 82058711-82058711 on Build GRCh38
rs1000281187 Chr.5: 82058684-82058685 on Build GRCh38
rs887578920 Chr.5: 82058622-82058622 on Build GRCh38
rs891569754 Chr.5: 82058721-82058721 on Build GRCh38
rs370509861 Chr.5: 82058520-82058520 on Build GRCh38
rs763168464 Chr.5: 82058471-82058471 on Build GRCh38
rs754971963 Chr.5: 82058593-82058593 on Build GRCh38
rs530696023 Chr.5: 82058640-82058640 on Build GRCh38
rs756914785 Chr.5: 82058627-82058627 on Build GRCh38
rs1054510922 Chr.5: 82058851-82058851 on Build GRCh38
rs1044708704 Chr.5: 82058651-82058651 on Build GRCh38
rs751011711 Chr.5: 82058609-82058610 on Build GRCh38
rs376140952 Chr.5: 82058472-82058472 on Build GRCh38
rs777169540 Chr.5: 82058446-82058446 on Build GRCh38
rs770565421 Chr.5: 82058525-82058525 on Build GRCh38
rs952981137 Chr.5: 82058830-82058830 on Build GRCh38
rs552087833 Chr.5: 82058642-82058642 on Build GRCh38
rs941197957 Chr.5: 82058678-82058678 on Build GRCh38
rs755651793 Chr.5: 82058867-82058867 on Build GRCh38
rs558875151 Chr.5: 82058780-82058780 on Build GRCh38
rs765623929 Chr.5: 82058466-82058466 on Build GRCh38
rs147595059 Chr.5: 82058821-82058821 on Build GRCh38
rs570504781 Chr.5: 82058650-82058650 on Build GRCh38
rs752585136 Chr.5: 82058608-82058608 on Build GRCh38
rs373358022 Chr.5: 82058607-82058607 on Build GRCh38
rs185636498 Chr.5: 82058782-82058782 on Build GRCh38
rs569857456 Chr.5: 82058639-82058639 on Build GRCh38
rs534743101 Chr.5: 82058681-82058681 on Build GRCh38
rs759326059 Chr.5: 82058499-82058499 on Build GRCh38
rs746650579 Chr.5: 82058624-82058624 on Build GRCh38
rs1045690577 Chr.5: 82058774-82058774 on Build GRCh38
rs751662053 Chr.5: 82058473-82058473 on Build GRCh38
rs796847264 Chr.5: 82058578-82058578 on Build GRCh38
rs564856686 Chr.5: 82058508-82058508 on Build GRCh38
rs777541393 Chr.5: 82058515-82058515 on Build GRCh38
rs985187967 Chr.5: 82058665-82058665 on Build GRCh38
rs775956594 Chr.5: 82058559-82058559 on Build GRCh38
rs759289095 Chr.5: 82058602-82058603 on Build GRCh38
rs540174795 Chr.5: 82058866-82058866 on Build GRCh38
rs757305933 Chr.5: 82058474-82058474 on Build GRCh38
rs143946049 Chr.5: 82058569-82058569 on Build GRCh38
rs535967162 Chr.5: 82058742-82058742 on Build GRCh38
rs775838818 Chr.5: 82058641-82058641 on Build GRCh38
rs975626501 Chr.5: 82058489-82058490 on Build GRCh38
rs137936585 Chr.5: 82058542-82058542 on Build GRCh38
rs778772048 Chr.5: 82058497-82058497 on Build GRCh38
rs558533524 Chr.5: 82058888-82058888 on Build GRCh38
rs1001594993 Chr.5: 82058563-82058563 on Build GRCh38
rs1037313280 Chr.5: 82058682-82058682 on Build GRCh38
rs775138303 Chr.5: 82058603-82058604 on Build GRCh38
rs766825287 Chr.5: 82058612-82058613 on Build GRCh38
rs749693793 Chr.5: 82058649-82058649 on Build GRCh38
rs773993466 Chr.5: 82058563-82058582 on Build GRCh38

View Full Product Details