Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 83652713-83653218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN1
Entrez Gene ID
1404
Gene Name
hyaluronan and proteoglycan link protein 1
Gene Aliases
CRT1, CRTL1
UniGene
Hs.2799
Gene Chromosome Location
Chr.5: 83638198-83721210 on Build GRCh38
Transcripts
NM_001884.3 XM_011543168.2 XM_017009051.1 XM_017009052.1 XM_017009053.1 XM_017009054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956272 c.111C>T p.G37G Chr.5: 83652814-83652814 on Build GRCh38
1070975 c.188T>G p.F63C Chr.5: 83652737-83652737 on Build GRCh38
161452 c.108T>G p.N36K Chr.5: 83652817-83652817 on Build GRCh38
3618440 c.169G>A p.V57I Chr.5: 83652756-83652756 on Build GRCh38
1620618 c.179C>A p.P60Q Chr.5: 83652746-83652746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72771293 Chr.5: 83652997-83652997 on Build GRCh38
rs185835624 Chr.5: 83652844-83652844 on Build GRCh38
rs775525333 Chr.5: 83652840-83652840 on Build GRCh38
rs3777049 Chr.5: 83653051-83653051 on Build GRCh38
rs1010726439 Chr.5: 83653016-83653016 on Build GRCh38
rs959661599 Chr.5: 83653191-83653191 on Build GRCh38
rs528701880 Chr.5: 83653084-83653084 on Build GRCh38
rs758251315 Chr.5: 83652855-83652855 on Build GRCh38
rs950672104 Chr.5: 83652804-83652804 on Build GRCh38
rs753629809 Chr.5: 83652859-83652862 on Build GRCh38
rs775898414 Chr.5: 83652782-83652782 on Build GRCh38
rs991351621 Chr.5: 83652859-83652859 on Build GRCh38
rs35909957 Chr.5: 83652978-83652979 on Build GRCh38
rs765976542 Chr.5: 83652832-83652832 on Build GRCh38
rs754759453 Chr.5: 83652745-83652745 on Build GRCh38
rs1050729068 Chr.5: 83652985-83652985 on Build GRCh38
rs753508982 Chr.5: 83652838-83652838 on Build GRCh38
rs755660679 Chr.5: 83652831-83652831 on Build GRCh38
rs576797253 Chr.5: 83652758-83652758 on Build GRCh38
rs962883787 Chr.5: 83652761-83652761 on Build GRCh38
rs911415933 Chr.5: 83652808-83652808 on Build GRCh38
rs768137611 Chr.5: 83652817-83652817 on Build GRCh38
rs536033064 Chr.5: 83653076-83653076 on Build GRCh38
rs768968964 Chr.5: 83652851-83652851 on Build GRCh38
rs762232801 Chr.5: 83652813-83652813 on Build GRCh38
rs867485078 Chr.5: 83653081-83653081 on Build GRCh38
rs1029345725 Chr.5: 83653065-83653065 on Build GRCh38
rs907168930 Chr.5: 83652960-83652960 on Build GRCh38
rs75228023 Chr.5: 83653167-83653167 on Build GRCh38
rs1006621269 Chr.5: 83653033-83653033 on Build GRCh38
rs774866703 Chr.5: 83652809-83652809 on Build GRCh38
rs971311045 Chr.5: 83653121-83653121 on Build GRCh38
rs1042590298 Chr.5: 83652953-83652953 on Build GRCh38
rs542489423 Chr.5: 83652811-83652811 on Build GRCh38
rs985128318 Chr.5: 83653166-83653166 on Build GRCh38
rs531265982 Chr.5: 83652894-83652894 on Build GRCh38
rs116714072 Chr.5: 83652972-83652972 on Build GRCh38
rs770973771 Chr.5: 83652767-83652767 on Build GRCh38
rs530272123 Chr.5: 83652797-83652797 on Build GRCh38
rs336971 Chr.5: 83653036-83653036 on Build GRCh38
rs547235109 Chr.5: 83652795-83652795 on Build GRCh38
rs1035024834 Chr.5: 83653173-83653173 on Build GRCh38
rs1056306631 Chr.5: 83652974-83652974 on Build GRCh38
rs144483408 Chr.5: 83652769-83652769 on Build GRCh38
rs559408235 Chr.5: 83652777-83652777 on Build GRCh38
rs982110777 Chr.5: 83652815-83652815 on Build GRCh38
rs763374644 Chr.5: 83652793-83652793 on Build GRCh38
rs749922283 Chr.5: 83652830-83652830 on Build GRCh38
rs930210723 Chr.5: 83652952-83652952 on Build GRCh38
rs994018766 Chr.5: 83653082-83653082 on Build GRCh38
rs368012751 Chr.5: 83653097-83653097 on Build GRCh38
rs552522157 Chr.5: 83652897-83652897 on Build GRCh38
rs778859881 Chr.5: 83652751-83652751 on Build GRCh38
rs748039286 Chr.5: 83652853-83652853 on Build GRCh38
rs924215368 Chr.5: 83652971-83652971 on Build GRCh38
rs770115705 Chr.5: 83652780-83652780 on Build GRCh38
rs760495727 Chr.5: 83652839-83652840 on Build GRCh38
rs1025527052 Chr.5: 83653091-83653091 on Build GRCh38
rs763996139 Chr.5: 83652840-83652842 on Build GRCh38
rs777902946 Chr.5: 83652864-83652864 on Build GRCh38
rs767623176 Chr.5: 83653045-83653045 on Build GRCh38
rs748048150 Chr.5: 83652752-83652752 on Build GRCh38

View Full Product Details