Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 86620539-86621036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7C
Entrez Gene ID
1350
Gene Name
cytochrome c oxidase subunit 7C
Gene Aliases
-
UniGene
Hs.430075
Gene Chromosome Location
Chr.5: 86617967-86620766 on Build GRCh38
Transcripts
NM_001867.2 NR_037402.1

Gene Symbol
MIR3607
Entrez Gene ID
100500805
Gene Name
microRNA 3607
Gene Aliases
mir-3607
UniGene
Gene Chromosome Location
Chr.5: 86620497-86620575 on Build GRCh38
Transcripts
NM_001867.2 NR_037402.1

SNP Detail

SNP ID Chromosome Location
rs933190779 Chr.5: 86620758-86620758 on Build GRCh38
rs35118667 Chr.5: 86620584-86620585 on Build GRCh38
rs188162668 Chr.5: 86620754-86620754 on Build GRCh38
rs540525954 Chr.5: 86620986-86620987 on Build GRCh38
rs946823177 Chr.5: 86620683-86620683 on Build GRCh38
rs900358635 Chr.5: 86620748-86620748 on Build GRCh38
rs993766780 Chr.5: 86620848-86620848 on Build GRCh38
rs898808052 Chr.5: 86620824-86620824 on Build GRCh38
rs756265841 Chr.5: 86620778-86620778 on Build GRCh38
rs1042892172 Chr.5: 86620689-86620689 on Build GRCh38
rs548951630 Chr.5: 86620780-86620780 on Build GRCh38
rs774922459 Chr.5: 86620983-86620983 on Build GRCh38
rs184196417 Chr.5: 86620994-86620994 on Build GRCh38
rs915219131 Chr.5: 86620609-86620609 on Build GRCh38
rs927958400 Chr.5: 86620634-86620634 on Build GRCh38
rs551657381 Chr.5: 86620764-86620764 on Build GRCh38
rs754145925 Chr.5: 86620854-86620854 on Build GRCh38
rs973564244 Chr.5: 86620574-86620574 on Build GRCh38
rs1051599465 Chr.5: 86620763-86620763 on Build GRCh38
rs1023787201 Chr.5: 86621007-86621007 on Build GRCh38
rs989534215 Chr.5: 86620986-86620986 on Build GRCh38
rs151331151 Chr.5: 86620651-86620651 on Build GRCh38
rs1027020516 Chr.5: 86620851-86620851 on Build GRCh38
rs767881963 Chr.5: 86620595-86620595 on Build GRCh38
rs551029920 Chr.5: 86620960-86620960 on Build GRCh38
rs537624217 Chr.5: 86620572-86620574 on Build GRCh38
rs757837278 Chr.5: 86620948-86620948 on Build GRCh38
rs1037348558 Chr.5: 86620796-86620796 on Build GRCh38
rs756127021 Chr.5: 86620643-86620646 on Build GRCh38
rs898907784 Chr.5: 86620913-86620913 on Build GRCh38
rs192055396 Chr.5: 86620870-86620870 on Build GRCh38
rs759485682 Chr.5: 86620982-86620983 on Build GRCh38
rs561127382 Chr.5: 86620792-86620792 on Build GRCh38
rs988567714 Chr.5: 86620643-86620643 on Build GRCh38
rs959460346 Chr.5: 86620958-86620958 on Build GRCh38
rs142485335 Chr.5: 86620596-86620596 on Build GRCh38
rs947297977 Chr.5: 86620635-86620635 on Build GRCh38
rs61188008 Chr.5: 86620720-86620720 on Build GRCh38
rs778238764 Chr.5: 86620810-86620810 on Build GRCh38
rs759904327 Chr.5: 86620883-86620883 on Build GRCh38
rs750211286 Chr.5: 86620739-86620739 on Build GRCh38
rs890396197 Chr.5: 86620797-86620798 on Build GRCh38
rs752860963 Chr.5: 86620657-86620657 on Build GRCh38
rs758600747 Chr.5: 86620675-86620675 on Build GRCh38
rs527535382 Chr.5: 86620762-86620762 on Build GRCh38
rs767838894 Chr.5: 86620997-86620997 on Build GRCh38
rs760750723 Chr.5: 86620583-86620583 on Build GRCh38
rs777892298 Chr.5: 86620691-86620691 on Build GRCh38
rs554557972 Chr.5: 86620983-86620986 on Build GRCh38

View Full Product Details