Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95391223-95391622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM81B
Entrez Gene ID
153643
Gene Name
family with sequence similarity 81 member B
Gene Aliases
-
UniGene
Hs.276287
Gene Chromosome Location
Chr.5: 95384793-95450441 on Build GRCh38
Transcripts
NM_152548.2 XM_005271905.2 XM_005271906.4 XM_005271907.3 XM_011543207.1 XM_011543208.1 XM_017009118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3141591 c.115A>G p.M39V Chr.5: 95391504-95391504 on Build GRCh38
1582659 c.69C>G p.F23L Chr.5: 95391458-95391458 on Build GRCh38
3141592 c.122C>T p.S41L Chr.5: 95391511-95391511 on Build GRCh38
6660174 c.101G>T p.G34V Chr.5: 95391490-95391490 on Build GRCh38
3618812 c.16C>T p.L6F Chr.5: 95391405-95391405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774936097 Chr.5: 95391469-95391469 on Build GRCh38
rs770259043 Chr.5: 95391414-95391414 on Build GRCh38
rs746130949 Chr.5: 95391410-95391410 on Build GRCh38
rs756795842 Chr.5: 95391356-95391356 on Build GRCh38
rs764441237 Chr.5: 95391450-95391450 on Build GRCh38
rs13360706 Chr.5: 95391349-95391349 on Build GRCh38
rs747354260 Chr.5: 95391584-95391584 on Build GRCh38
rs1000182315 Chr.5: 95391334-95391334 on Build GRCh38
rs771061770 Chr.5: 95391405-95391405 on Build GRCh38
rs917057820 Chr.5: 95391361-95391361 on Build GRCh38
rs754811490 Chr.5: 95391534-95391534 on Build GRCh38
rs894497624 Chr.5: 95391559-95391559 on Build GRCh38
rs534418226 Chr.5: 95391488-95391488 on Build GRCh38
rs996905357 Chr.5: 95391271-95391271 on Build GRCh38
rs963775105 Chr.5: 95391333-95391333 on Build GRCh38
rs372506553 Chr.5: 95391505-95391505 on Build GRCh38
rs746999587 Chr.5: 95391402-95391402 on Build GRCh38
rs778577584 Chr.5: 95391488-95391488 on Build GRCh38
rs200016294 Chr.5: 95391440-95391440 on Build GRCh38
rs778663346 Chr.5: 95391535-95391535 on Build GRCh38
rs750048781 Chr.5: 95391518-95391518 on Build GRCh38
rs776053729 Chr.5: 95391420-95391420 on Build GRCh38
rs761180106 Chr.5: 95391489-95391489 on Build GRCh38
rs781468838 Chr.5: 95391542-95391542 on Build GRCh38
rs763417366 Chr.5: 95391351-95391351 on Build GRCh38
rs201749392 Chr.5: 95391453-95391453 on Build GRCh38
rs779867200 Chr.5: 95391380-95391380 on Build GRCh38
rs968662323 Chr.5: 95391297-95391297 on Build GRCh38
rs769088350 Chr.5: 95391562-95391562 on Build GRCh38
rs775859523 Chr.5: 95391560-95391560 on Build GRCh38
rs747899251 Chr.5: 95391443-95391443 on Build GRCh38
rs945867724 Chr.5: 95391434-95391434 on Build GRCh38
rs748145450 Chr.5: 95391539-95391539 on Build GRCh38
rs373741847 Chr.5: 95391362-95391362 on Build GRCh38
rs746069736 Chr.5: 95391541-95391541 on Build GRCh38
rs770207792 Chr.5: 95391558-95391558 on Build GRCh38
rs928583844 Chr.5: 95391501-95391501 on Build GRCh38
rs930281184 Chr.5: 95391442-95391443 on Build GRCh38
rs1028365936 Chr.5: 95391279-95391279 on Build GRCh38
rs562139540 Chr.5: 95391395-95391395 on Build GRCh38
rs553195188 Chr.5: 95391252-95391252 on Build GRCh38
rs541850371 Chr.5: 95391355-95391355 on Build GRCh38
rs755948817 Chr.5: 95391521-95391521 on Build GRCh38
rs191657761 Chr.5: 95391531-95391531 on Build GRCh38
rs764069181 Chr.5: 95391414-95391414 on Build GRCh38
rs767019989 Chr.5: 95391360-95391360 on Build GRCh38
rs755865577 Chr.5: 95391376-95391376 on Build GRCh38
rs767046810 Chr.5: 95391508-95391508 on Build GRCh38
rs369527523 Chr.5: 95391354-95391354 on Build GRCh38
rs1049788157 Chr.5: 95391566-95391566 on Build GRCh38
rs779314255 Chr.5: 95391530-95391530 on Build GRCh38
rs749203848 Chr.5: 95391384-95391384 on Build GRCh38
rs759928858 Chr.5: 95391341-95391341 on Build GRCh38
rs910900795 Chr.5: 95391275-95391275 on Build GRCh38
rs746073366 Chr.5: 95391551-95391551 on Build GRCh38
rs375243027 Chr.5: 95391430-95391430 on Build GRCh38
rs372813424 Chr.5: 95391340-95391340 on Build GRCh38
rs765725475 Chr.5: 95391344-95391344 on Build GRCh38
rs780437960 Chr.5: 95391413-95391413 on Build GRCh38
rs772251255 Chr.5: 95391561-95391561 on Build GRCh38
rs200301845 Chr.5: 95391476-95391476 on Build GRCh38

View Full Product Details